Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 54476227-54476733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMFB
Entrez Gene ID
2764
Gene Name
glia maturation factor beta
Gene Aliases
GMF
UniGene
Hs.151413
Gene Chromosome Location
Chr.14: 54474491-54489026 on Build GRCh38
Transcripts
NM_004124.2

SNP Detail

SNP ID Chromosome Location
rs944800949 Chr.14: 54476353-54476353 on Build GRCh38
rs976163798 Chr.14: 54476682-54476682 on Build GRCh38
rs563092627 Chr.14: 54476446-54476446 on Build GRCh38
rs565610741 Chr.14: 54476564-54476564 on Build GRCh38
rs532043085 Chr.14: 54476374-54476374 on Build GRCh38
rs200146109 Chr.14: 54476653-54476653 on Build GRCh38
rs545385619 Chr.14: 54476519-54476519 on Build GRCh38
rs952240469 Chr.14: 54476314-54476314 on Build GRCh38
rs945293970 Chr.14: 54476419-54476420 on Build GRCh38
rs189772591 Chr.14: 54476588-54476588 on Build GRCh38
rs761212284 Chr.14: 54476344-54476344 on Build GRCh38
rs543129009 Chr.14: 54476384-54476384 on Build GRCh38
rs545770216 Chr.14: 54476316-54476316 on Build GRCh38
rs1043636289 Chr.14: 54476428-54476428 on Build GRCh38
rs761820871 Chr.14: 54476673-54476673 on Build GRCh38
rs972252351 Chr.14: 54476282-54476282 on Build GRCh38
rs539147699 Chr.14: 54476319-54476319 on Build GRCh38
rs910665269 Chr.14: 54476327-54476327 on Build GRCh38
rs896805062 Chr.14: 54476557-54476557 on Build GRCh38
rs995117914 Chr.14: 54476584-54476584 on Build GRCh38
rs1013295831 Chr.14: 54476532-54476532 on Build GRCh38
rs572522750 Chr.14: 54476356-54476356 on Build GRCh38
rs952291298 Chr.14: 54476619-54476619 on Build GRCh38
rs576684186 Chr.14: 54476494-54476494 on Build GRCh38
rs1015680879 Chr.14: 54476543-54476543 on Build GRCh38
rs986744849 Chr.14: 54476623-54476623 on Build GRCh38
rs937852537 Chr.14: 54476462-54476465 on Build GRCh38
rs554164424 Chr.14: 54476324-54476324 on Build GRCh38
rs1027790314 Chr.14: 54476291-54476292 on Build GRCh38
rs1017733294 Chr.14: 54476648-54476648 on Build GRCh38
rs368138563 Chr.14: 54476380-54476380 on Build GRCh38
rs1027893012 Chr.14: 54476601-54476601 on Build GRCh38

View Full Product Details