Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 54479672-54480218 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMFB
Entrez Gene ID
2764
Gene Name
glia maturation factor beta
Gene Aliases
GMF
UniGene
Hs.151413
Gene Chromosome Location
Chr.14: 54474491-54489026 on Build GRCh38
Transcripts
NM_004124.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6311129 c.353C>A p.T118N Chr.14: 54479790-54479790 on Build GRCh38
6672005 c.356+1G>A p.? Chr.14: 54479786-54479786 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs376113581 Chr.14: 54479874-54479874 on Build GRCh38
rs750001227 Chr.14: 54479755-54479755 on Build GRCh38
rs777192530 Chr.14: 54480147-54480147 on Build GRCh38
rs202188385 Chr.14: 54479878-54479878 on Build GRCh38
rs1026311203 Chr.14: 54479918-54479918 on Build GRCh38
rs748444440 Chr.14: 54479800-54479800 on Build GRCh38
rs887361146 Chr.14: 54480036-54480036 on Build GRCh38
rs948689184 Chr.14: 54479707-54479707 on Build GRCh38
rs145015405 Chr.14: 54479858-54479858 on Build GRCh38
rs757286777 Chr.14: 54479750-54479750 on Build GRCh38
rs938351758 Chr.14: 54479801-54479801 on Build GRCh38
rs149936512 Chr.14: 54479805-54479805 on Build GRCh38
rs558593428 Chr.14: 54479922-54479922 on Build GRCh38
rs979919664 Chr.14: 54480069-54480069 on Build GRCh38
rs764070065 Chr.14: 54479744-54479744 on Build GRCh38
rs997411824 Chr.14: 54480176-54480176 on Build GRCh38
rs950869154 Chr.14: 54479968-54479968 on Build GRCh38
rs769212920 Chr.14: 54479982-54479982 on Build GRCh38
rs185319696 Chr.14: 54479945-54479945 on Build GRCh38
rs909269491 Chr.14: 54480010-54480010 on Build GRCh38
rs765243173 Chr.14: 54479889-54479889 on Build GRCh38
rs982949994 Chr.14: 54479706-54479706 on Build GRCh38
rs906931753 Chr.14: 54479963-54479963 on Build GRCh38
rs560572081 Chr.14: 54480108-54480108 on Build GRCh38
rs10139223 Chr.14: 54480192-54480192 on Build GRCh38
rs907104786 Chr.14: 54479757-54479757 on Build GRCh38
rs192472698 Chr.14: 54479877-54479877 on Build GRCh38
rs749750001 Chr.14: 54479814-54479814 on Build GRCh38
rs553104864 Chr.14: 54479749-54479749 on Build GRCh38
rs1022482979 Chr.14: 54479876-54479876 on Build GRCh38
rs1017541176 Chr.14: 54480095-54480095 on Build GRCh38
rs778164384 Chr.14: 54479807-54479807 on Build GRCh38
rs190172573 Chr.14: 54480140-54480140 on Build GRCh38
rs375494190 Chr.14: 54479752-54479752 on Build GRCh38
rs377694702 Chr.14: 54479768-54479768 on Build GRCh38
rs367684737 Chr.14: 54479887-54479887 on Build GRCh38
rs543369570 Chr.14: 54479921-54479921 on Build GRCh38
rs376093428 Chr.14: 54479904-54479904 on Build GRCh38
rs1044175684 Chr.14: 54479930-54479935 on Build GRCh38
rs976088890 Chr.14: 54479914-54479914 on Build GRCh38
rs1004394107 Chr.14: 54480044-54480044 on Build GRCh38
rs776076153 Chr.14: 54479867-54479868 on Build GRCh38
rs948577437 Chr.14: 54480026-54480026 on Build GRCh38
rs938591139 Chr.14: 54480129-54480129 on Build GRCh38
rs73273169 Chr.14: 54479870-54479870 on Build GRCh38
rs984675740 Chr.14: 54479700-54479700 on Build GRCh38
rs374721333 Chr.14: 54480138-54480138 on Build GRCh38
rs1017025724 Chr.14: 54479735-54479735 on Build GRCh38
rs541383272 Chr.14: 54480008-54480008 on Build GRCh38
rs370982356 Chr.14: 54479769-54479769 on Build GRCh38
rs541936338 Chr.14: 54480194-54480194 on Build GRCh38
rs760043845 Chr.14: 54479941-54479941 on Build GRCh38
rs761298925 Chr.14: 54479903-54479903 on Build GRCh38
rs745994511 Chr.14: 54479867-54479867 on Build GRCh38
rs928563046 Chr.14: 54480089-54480089 on Build GRCh38
rs909037039 Chr.14: 54479704-54479704 on Build GRCh38
rs372364778 Chr.14: 54480031-54480031 on Build GRCh38

View Full Product Details