Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 54480748-54481257 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMFB
Entrez Gene ID
2764
Gene Name
glia maturation factor beta
Gene Aliases
GMF
UniGene
Hs.151413
Gene Chromosome Location
Chr.14: 54474491-54489026 on Build GRCh38
Transcripts
NM_004124.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6442415 c.278C>T p.P93L Chr.14: 54480879-54480879 on Build GRCh38
433141 c.226C>T p.Q76* Chr.14: 54480931-54480931 on Build GRCh38
956298 c.250T>C p.Y84H Chr.14: 54480907-54480907 on Build GRCh38
1748891 c.235G>A p.D79N Chr.14: 54480922-54480922 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1010033995 Chr.14: 54481125-54481125 on Build GRCh38
rs1020109286 Chr.14: 54481126-54481126 on Build GRCh38
rs898751988 Chr.14: 54480777-54480783 on Build GRCh38
rs905850025 Chr.14: 54481047-54481047 on Build GRCh38
rs779481745 Chr.14: 54480882-54480882 on Build GRCh38
rs1035554848 Chr.14: 54481029-54481029 on Build GRCh38
rs757229143 Chr.14: 54481022-54481022 on Build GRCh38
rs759163667 Chr.14: 54480979-54480979 on Build GRCh38
rs778878807 Chr.14: 54481078-54481078 on Build GRCh38
rs1049937488 Chr.14: 54480814-54480814 on Build GRCh38
rs60913976 Chr.14: 54480825-54480825 on Build GRCh38
rs1047856656 Chr.14: 54481026-54481026 on Build GRCh38
rs376955215 Chr.14: 54481044-54481044 on Build GRCh38
rs1053276079 Chr.14: 54481178-54481178 on Build GRCh38
rs980404620 Chr.14: 54481014-54481014 on Build GRCh38
rs768857709 Chr.14: 54480994-54480997 on Build GRCh38
rs375266091 Chr.14: 54481034-54481034 on Build GRCh38
rs754086967 Chr.14: 54480990-54480990 on Build GRCh38
rs181364600 Chr.14: 54481117-54481117 on Build GRCh38
rs747204561 Chr.14: 54480905-54480905 on Build GRCh38
rs377744361 Chr.14: 54480914-54480914 on Build GRCh38
rs745887555 Chr.14: 54480886-54480886 on Build GRCh38
rs754689184 Chr.14: 54480828-54480828 on Build GRCh38
rs767217701 Chr.14: 54480980-54480980 on Build GRCh38
rs753875555 Chr.14: 54481012-54481012 on Build GRCh38
rs539921069 Chr.14: 54481196-54481196 on Build GRCh38
rs769549222 Chr.14: 54480934-54480934 on Build GRCh38
rs55773293 Chr.14: 54481015-54481015 on Build GRCh38
rs373005080 Chr.14: 54480847-54480847 on Build GRCh38
rs772270133 Chr.14: 54480896-54480896 on Build GRCh38
rs907669338 Chr.14: 54481048-54481048 on Build GRCh38
rs757652768 Chr.14: 54480850-54480850 on Build GRCh38
rs777964033 Chr.14: 54480838-54480838 on Build GRCh38
rs988581719 Chr.14: 54480784-54480784 on Build GRCh38
rs762934830 Chr.14: 54480944-54480944 on Build GRCh38
rs764100175 Chr.14: 54481115-54481115 on Build GRCh38
rs59955412 Chr.14: 54480783-54480783 on Build GRCh38
rs1006468087 Chr.14: 54481041-54481041 on Build GRCh38
rs186398443 Chr.14: 54481123-54481123 on Build GRCh38
rs757657067 Chr.14: 54480996-54480996 on Build GRCh38
rs541974082 Chr.14: 54481104-54481104 on Build GRCh38
rs978685806 Chr.14: 54481213-54481213 on Build GRCh38
rs750965291 Chr.14: 54481009-54481009 on Build GRCh38
rs769048645 Chr.14: 54480908-54480908 on Build GRCh38
rs913144420 Chr.14: 54480845-54480845 on Build GRCh38
rs895866774 Chr.14: 54481124-54481124 on Build GRCh38
rs941736705 Chr.14: 54481056-54481056 on Build GRCh38
rs973101196 Chr.14: 54481077-54481077 on Build GRCh38
rs558838905 Chr.14: 54481219-54481219 on Build GRCh38
rs978868899 Chr.14: 54480871-54480871 on Build GRCh38
rs891876023 Chr.14: 54480947-54480947 on Build GRCh38
rs773214575 Chr.14: 54480936-54480936 on Build GRCh38
rs751105605 Chr.14: 54480962-54480962 on Build GRCh38
rs779207592 Chr.14: 54481004-54481004 on Build GRCh38
rs969079397 Chr.14: 54481197-54481197 on Build GRCh38
rs944221138 Chr.14: 54481156-54481156 on Build GRCh38
rs761700818 Chr.14: 54480926-54480926 on Build GRCh38
rs780352554 Chr.14: 54480902-54480902 on Build GRCh38
rs371154404 Chr.14: 54480952-54480952 on Build GRCh38

View Full Product Details