Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 55352277-55352769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO34
Entrez Gene ID
55030
Gene Name
F-box protein 34
Gene Aliases
CGI-301, Fbx34
UniGene
Hs.525348
Gene Chromosome Location
Chr.14: 55271303-55355222 on Build GRCh38
Transcripts
NM_017943.3 NM_152231.1 XM_005267791.4 XM_006720185.3 XM_017021391.1 XM_017021392.1 XM_017021393.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5740105 c.1969G>A p.A657T Chr.14: 55352359-55352359 on Build GRCh38
6662210 c.1908G>T p.K636N Chr.14: 55352298-55352298 on Build GRCh38
7162708 c.1943G>T p.R648L Chr.14: 55352333-55352333 on Build GRCh38
1748896 c.1943G>A p.R648Q Chr.14: 55352333-55352333 on Build GRCh38
3719622 c.1947G>T p.W649C Chr.14: 55352337-55352337 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs942071592 Chr.14: 55352737-55352737 on Build GRCh38
rs765201566 Chr.14: 55352474-55352474 on Build GRCh38
rs767959423 Chr.14: 55352328-55352328 on Build GRCh38
rs753888835 Chr.14: 55352340-55352340 on Build GRCh38
rs764638239 Chr.14: 55352445-55352445 on Build GRCh38
rs767552943 Chr.14: 55352391-55352391 on Build GRCh38
rs925850398 Chr.14: 55352599-55352599 on Build GRCh38
rs10138395 Chr.14: 55352501-55352501 on Build GRCh38
rs1047594682 Chr.14: 55352704-55352704 on Build GRCh38
rs929115137 Chr.14: 55352643-55352643 on Build GRCh38
rs966828794 Chr.14: 55352584-55352584 on Build GRCh38
rs1025615778 Chr.14: 55352394-55352394 on Build GRCh38
rs369899790 Chr.14: 55352365-55352365 on Build GRCh38
rs761125107 Chr.14: 55352433-55352433 on Build GRCh38
rs766190732 Chr.14: 55352415-55352415 on Build GRCh38
rs371099912 Chr.14: 55352539-55352539 on Build GRCh38
rs746109098 Chr.14: 55352356-55352356 on Build GRCh38
rs758567096 Chr.14: 55352477-55352477 on Build GRCh38
rs747999360 Chr.14: 55352541-55352541 on Build GRCh38
rs951471330 Chr.14: 55352399-55352399 on Build GRCh38
rs145830018 Chr.14: 55352358-55352358 on Build GRCh38
rs552238438 Chr.14: 55352636-55352636 on Build GRCh38
rs957078316 Chr.14: 55352593-55352593 on Build GRCh38
rs772722852 Chr.14: 55352740-55352740 on Build GRCh38
rs1017603455 Chr.14: 55352476-55352476 on Build GRCh38
rs945705295 Chr.14: 55352585-55352585 on Build GRCh38
rs138667502 Chr.14: 55352555-55352555 on Build GRCh38
rs762670037 Chr.14: 55352401-55352401 on Build GRCh38
rs374114341 Chr.14: 55352554-55352554 on Build GRCh38
rs887239445 Chr.14: 55352714-55352714 on Build GRCh38
rs1005765130 Chr.14: 55352439-55352439 on Build GRCh38
rs535237309 Chr.14: 55352333-55352333 on Build GRCh38
rs201837605 Chr.14: 55352376-55352376 on Build GRCh38
rs746799004 Chr.14: 55352534-55352534 on Build GRCh38
rs781157283 Chr.14: 55352540-55352540 on Build GRCh38
rs1047822783 Chr.14: 55352651-55352651 on Build GRCh38
rs114576128 Chr.14: 55352600-55352600 on Build GRCh38
rs559347198 Chr.14: 55352739-55352739 on Build GRCh38
rs771347407 Chr.14: 55352588-55352588 on Build GRCh38
rs769796413 Chr.14: 55352542-55352542 on Build GRCh38
rs376757183 Chr.14: 55352458-55352458 on Build GRCh38
rs772815143 Chr.14: 55352398-55352398 on Build GRCh38
rs192116680 Chr.14: 55352654-55352654 on Build GRCh38
rs948633931 Chr.14: 55352604-55352604 on Build GRCh38
rs764763112 Chr.14: 55352335-55352335 on Build GRCh38
rs745951101 Chr.14: 55352379-55352379 on Build GRCh38
rs267604008 Chr.14: 55352347-55352347 on Build GRCh38
rs904315742 Chr.14: 55352349-55352349 on Build GRCh38
rs779804049 Chr.14: 55352369-55352369 on Build GRCh38
rs757131448 Chr.14: 55352608-55352609 on Build GRCh38
rs770704574 Chr.14: 55352572-55352572 on Build GRCh38
rs1047772301 Chr.14: 55352467-55352467 on Build GRCh38
rs981277132 Chr.14: 55352632-55352632 on Build GRCh38
rs1043374114 Chr.14: 55352308-55352308 on Build GRCh38
rs757297376 Chr.14: 55352346-55352346 on Build GRCh38
rs148730342 Chr.14: 55352514-55352514 on Build GRCh38
rs746866580 Chr.14: 55352370-55352370 on Build GRCh38
rs901026926 Chr.14: 55352650-55352650 on Build GRCh38
rs373736848 Chr.14: 55352388-55352388 on Build GRCh38
rs753159194 Chr.14: 55352332-55352332 on Build GRCh38
rs142724770 Chr.14: 55352621-55352621 on Build GRCh38
rs754323973 Chr.14: 55352456-55352456 on Build GRCh38
rs768578981 Chr.14: 55352374-55352374 on Build GRCh38
rs369569876 Chr.14: 55352727-55352727 on Build GRCh38
rs759901831 Chr.14: 55352323-55352323 on Build GRCh38
rs189204235 Chr.14: 55352587-55352587 on Build GRCh38
rs928138195 Chr.14: 55352533-55352533 on Build GRCh38

View Full Product Details