Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 55366261-55366774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG14
Entrez Gene ID
22863
Gene Name
autophagy related 14
Gene Aliases
ATG14L, BARKOR, KIAA0831
UniGene
Hs.414809
Gene Chromosome Location
Chr.14: 55366391-55411858 on Build GRCh38
Transcripts
NM_014924.4 XM_011536563.2

SNP Detail

SNP ID Chromosome Location
rs982096483 Chr.14: 55366562-55366562 on Build GRCh38
rs918061879 Chr.14: 55366298-55366298 on Build GRCh38
rs930375460 Chr.14: 55366574-55366574 on Build GRCh38
rs1056657174 Chr.14: 55366297-55366297 on Build GRCh38
rs568487775 Chr.14: 55366593-55366593 on Build GRCh38
rs79228435 Chr.14: 55366612-55366612 on Build GRCh38
rs1008388052 Chr.14: 55366666-55366666 on Build GRCh38
rs568355925 Chr.14: 55366501-55366501 on Build GRCh38
rs368943916 Chr.14: 55366361-55366361 on Build GRCh38
rs146530522 Chr.14: 55366301-55366301 on Build GRCh38
rs537903034 Chr.14: 55366352-55366352 on Build GRCh38
rs150148557 Chr.14: 55366632-55366632 on Build GRCh38
rs968116672 Chr.14: 55366440-55366440 on Build GRCh38
rs140876174 Chr.14: 55366380-55366380 on Build GRCh38
rs1050599120 Chr.14: 55366624-55366624 on Build GRCh38
rs994481716 Chr.14: 55366456-55366456 on Build GRCh38
rs546503675 Chr.14: 55366401-55366401 on Build GRCh38
rs1000930624 Chr.14: 55366721-55366724 on Build GRCh38
rs545488950 Chr.14: 55366333-55366333 on Build GRCh38
rs562698824 Chr.14: 55366304-55366304 on Build GRCh38
rs10692792 Chr.14: 55366642-55366643 on Build GRCh38
rs386777723 Chr.14: 55366640-55366642 on Build GRCh38
rs1033648828 Chr.14: 55366491-55366491 on Build GRCh38
rs745365088 Chr.14: 55366526-55366526 on Build GRCh38
rs919003799 Chr.14: 55366564-55366564 on Build GRCh38
rs2297816 Chr.14: 55366320-55366320 on Build GRCh38
rs529138759 Chr.14: 55366494-55366494 on Build GRCh38
rs938207190 Chr.14: 55366291-55366291 on Build GRCh38
rs965829898 Chr.14: 55366367-55366368 on Build GRCh38
rs932116572 Chr.14: 55366616-55366616 on Build GRCh38
rs1009810175 Chr.14: 55366656-55366656 on Build GRCh38
rs923935134 Chr.14: 55366582-55366582 on Build GRCh38
rs114720589 Chr.14: 55366442-55366442 on Build GRCh38
rs768450777 Chr.14: 55366702-55366702 on Build GRCh38
rs754331351 Chr.14: 55366452-55366452 on Build GRCh38
rs1007401529 Chr.14: 55366357-55366357 on Build GRCh38
rs757765868 Chr.14: 55366507-55366507 on Build GRCh38
rs746693048 Chr.14: 55366691-55366691 on Build GRCh38
rs889512298 Chr.14: 55366634-55366634 on Build GRCh38
rs996376284 Chr.14: 55366324-55366324 on Build GRCh38
rs1026886495 Chr.14: 55366498-55366498 on Build GRCh38
rs867761814 Chr.14: 55366727-55366727 on Build GRCh38
rs557692545 Chr.14: 55366388-55366388 on Build GRCh38
rs10628679 Chr.14: 55366645-55366646 on Build GRCh38
rs990631804 Chr.14: 55366515-55366515 on Build GRCh38
rs752714204 Chr.14: 55366718-55366718 on Build GRCh38
rs757968298 Chr.14: 55366606-55366606 on Build GRCh38
rs1049216526 Chr.14: 55366581-55366582 on Build GRCh38
rs767839364 Chr.14: 55366592-55366592 on Build GRCh38
rs868190142 Chr.14: 55366550-55366550 on Build GRCh38
rs1019708205 Chr.14: 55366437-55366437 on Build GRCh38
rs778333697 Chr.14: 55366516-55366516 on Build GRCh38
rs940171391 Chr.14: 55366529-55366530 on Build GRCh38
rs971874678 Chr.14: 55366548-55366548 on Build GRCh38
rs993815741 Chr.14: 55366717-55366717 on Build GRCh38
rs375405950 Chr.14: 55366674-55366674 on Build GRCh38
rs779723161 Chr.14: 55366629-55366629 on Build GRCh38
rs550858319 Chr.14: 55366583-55366583 on Build GRCh38
rs1054028011 Chr.14: 55366598-55366598 on Build GRCh38
rs971227714 Chr.14: 55366553-55366553 on Build GRCh38
rs1040252326 Chr.14: 55366605-55366606 on Build GRCh38
rs889039366 Chr.14: 55366339-55366339 on Build GRCh38

View Full Product Details