Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 57202515-57203056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC5
Entrez Gene ID
10640
Gene Name
exocyst complex component 5
Gene Aliases
HSEC10, PRO1912, SEC10, SEC10L1, SEC10P
UniGene
Hs.743987
Gene Chromosome Location
Chr.14: 57202476-57268899 on Build GRCh38
Transcripts
NM_006544.3

SNP Detail

SNP ID Chromosome Location
rs1056958223 Chr.14: 57202779-57202780 on Build GRCh38
rs539013150 Chr.14: 57202864-57202864 on Build GRCh38
rs898920577 Chr.14: 57202841-57202841 on Build GRCh38
rs548037383 Chr.14: 57202654-57202654 on Build GRCh38
rs1028872789 Chr.14: 57202601-57202601 on Build GRCh38
rs10138118 Chr.14: 57202982-57202982 on Build GRCh38
rs530959862 Chr.14: 57202851-57202852 on Build GRCh38
rs940372443 Chr.14: 57202814-57202814 on Build GRCh38
rs571703249 Chr.14: 57202851-57202851 on Build GRCh38
rs987044692 Chr.14: 57202613-57202613 on Build GRCh38
rs11851015 Chr.14: 57202815-57202815 on Build GRCh38
rs1008509614 Chr.14: 57202882-57202882 on Build GRCh38
rs114074295 Chr.14: 57202651-57202651 on Build GRCh38
rs979999104 Chr.14: 57202736-57202736 on Build GRCh38
rs995695426 Chr.14: 57202580-57202580 on Build GRCh38
rs368226654 Chr.14: 57202571-57202571 on Build GRCh38
rs547114610 Chr.14: 57202559-57202559 on Build GRCh38
rs905662712 Chr.14: 57202835-57202846 on Build GRCh38
rs889940522 Chr.14: 57202852-57202852 on Build GRCh38
rs1019656321 Chr.14: 57202620-57202620 on Build GRCh38
rs565434127 Chr.14: 57202623-57202623 on Build GRCh38
rs752739416 Chr.14: 57202548-57202549 on Build GRCh38
rs372023709 Chr.14: 57202785-57202785 on Build GRCh38
rs968864668 Chr.14: 57202954-57202954 on Build GRCh38
rs569934263 Chr.14: 57202794-57202794 on Build GRCh38
rs898879356 Chr.14: 57202544-57202544 on Build GRCh38
rs1019897850 Chr.14: 57202890-57202890 on Build GRCh38
rs954361319 Chr.14: 57202604-57202604 on Build GRCh38
rs956247551 Chr.14: 57202912-57202912 on Build GRCh38
rs988788592 Chr.14: 57202641-57202641 on Build GRCh38
rs938640978 Chr.14: 57202778-57202779 on Build GRCh38
rs764366295 Chr.14: 57202610-57202610 on Build GRCh38
rs142382358 Chr.14: 57202931-57202931 on Build GRCh38
rs940762825 Chr.14: 57202548-57202548 on Build GRCh38
rs192216546 Chr.14: 57202939-57202939 on Build GRCh38
rs1022041910 Chr.14: 57202943-57202943 on Build GRCh38
rs550166393 Chr.14: 57202840-57202840 on Build GRCh38
rs777961907 Chr.14: 57202688-57202688 on Build GRCh38
rs11552404 Chr.14: 57202570-57202570 on Build GRCh38
rs927171243 Chr.14: 57203014-57203014 on Build GRCh38
rs554356730 Chr.14: 57202918-57202918 on Build GRCh38
rs927110760 Chr.14: 57202777-57202778 on Build GRCh38
rs886553485 Chr.14: 57202540-57202540 on Build GRCh38

View Full Product Details