Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 57206078-57206614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC5
Entrez Gene ID
10640
Gene Name
exocyst complex component 5
Gene Aliases
HSEC10, PRO1912, SEC10, SEC10L1, SEC10P
UniGene
Hs.743987
Gene Chromosome Location
Chr.14: 57202476-57268899 on Build GRCh38
Transcripts
NM_006544.3 XM_005267272.3

SNP Detail

SNP ID Chromosome Location
rs923599954 Chr.14: 57206149-57206149 on Build GRCh38
rs970552041 Chr.14: 57206373-57206373 on Build GRCh38
rs190404950 Chr.14: 57206338-57206338 on Build GRCh38
rs546902237 Chr.14: 57206432-57206432 on Build GRCh38
rs550322594 Chr.14: 57206115-57206115 on Build GRCh38
rs888734875 Chr.14: 57206573-57206573 on Build GRCh38
rs1023446020 Chr.14: 57206362-57206362 on Build GRCh38
rs1054225998 Chr.14: 57206566-57206567 on Build GRCh38
rs751027329 Chr.14: 57206433-57206433 on Build GRCh38
rs552460155 Chr.14: 57206579-57206580 on Build GRCh38
rs10706566 Chr.14: 57206383-57206383 on Build GRCh38
rs779298821 Chr.14: 57206262-57206262 on Build GRCh38
rs1041053033 Chr.14: 57206210-57206210 on Build GRCh38
rs572343022 Chr.14: 57206565-57206565 on Build GRCh38
rs373149793 Chr.14: 57206508-57206508 on Build GRCh38
rs1030675532 Chr.14: 57206393-57206393 on Build GRCh38
rs929642787 Chr.14: 57206196-57206196 on Build GRCh38
rs1048377281 Chr.14: 57206531-57206531 on Build GRCh38
rs555509675 Chr.14: 57206175-57206175 on Build GRCh38
rs377164569 Chr.14: 57206174-57206174 on Build GRCh38
rs369341461 Chr.14: 57206271-57206271 on Build GRCh38
rs893915894 Chr.14: 57206354-57206354 on Build GRCh38
rs985963759 Chr.14: 57206463-57206463 on Build GRCh38
rs1032341740 Chr.14: 57206351-57206351 on Build GRCh38
rs942456523 Chr.14: 57206453-57206454 on Build GRCh38
rs749021863 Chr.14: 57206184-57206185 on Build GRCh38
rs951291661 Chr.14: 57206408-57206408 on Build GRCh38
rs944132014 Chr.14: 57206475-57206475 on Build GRCh38
rs757986630 Chr.14: 57206253-57206253 on Build GRCh38
rs900659746 Chr.14: 57206264-57206264 on Build GRCh38
rs528521902 Chr.14: 57206426-57206426 on Build GRCh38
rs931889085 Chr.14: 57206444-57206444 on Build GRCh38
rs34380808 Chr.14: 57206463-57206463 on Build GRCh38
rs981879777 Chr.14: 57206380-57206381 on Build GRCh38
rs138867006 Chr.14: 57206193-57206193 on Build GRCh38
rs1023413009 Chr.14: 57206106-57206106 on Build GRCh38
rs970505381 Chr.14: 57206357-57206357 on Build GRCh38
rs568908051 Chr.14: 57206138-57206138 on Build GRCh38
rs562277388 Chr.14: 57206501-57206501 on Build GRCh38
rs897111869 Chr.14: 57206223-57206223 on Build GRCh38
rs902802793 Chr.14: 57206533-57206533 on Build GRCh38
rs944120561 Chr.14: 57206205-57206205 on Build GRCh38
rs902736687 Chr.14: 57206331-57206331 on Build GRCh38
rs572931730 Chr.14: 57206164-57206164 on Build GRCh38
rs888682558 Chr.14: 57206286-57206286 on Build GRCh38
rs956545718 Chr.14: 57206418-57206418 on Build GRCh38
rs45586331 Chr.14: 57206254-57206254 on Build GRCh38
rs573866469 Chr.14: 57206182-57206182 on Build GRCh38
rs911684802 Chr.14: 57206474-57206474 on Build GRCh38
rs7141911 Chr.14: 57206153-57206153 on Build GRCh38
rs1052037157 Chr.14: 57206309-57206309 on Build GRCh38
rs754556015 Chr.14: 57206442-57206442 on Build GRCh38
rs893786966 Chr.14: 57206568-57206568 on Build GRCh38
rs929990643 Chr.14: 57206526-57206526 on Build GRCh38
rs117870712 Chr.14: 57206183-57206183 on Build GRCh38
rs1012449657 Chr.14: 57206287-57206287 on Build GRCh38
rs796756463 Chr.14: 57206393-57206393 on Build GRCh38
rs1023416652 Chr.14: 57206161-57206161 on Build GRCh38
rs981819565 Chr.14: 57206366-57206366 on Build GRCh38
rs1023686702 Chr.14: 57206580-57206580 on Build GRCh38
rs1041496138 Chr.14: 57206476-57206476 on Build GRCh38
rs528382169 Chr.14: 57206150-57206150 on Build GRCh38
rs922325108 Chr.14: 57206477-57206477 on Build GRCh38

View Full Product Details