Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 58200031-58200533 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTR10
Entrez Gene ID
55860
Gene Name
actin-related protein 10 homolog
Gene Aliases
ACTR11, Arp10, Arp11, HARP11
UniGene
Hs.509451
Gene Chromosome Location
Chr.14: 58200115-58235636 on Build GRCh38
Transcripts
NM_018477.2 XM_011536960.1 XM_011536961.1 XM_011536962.2 XM_011536963.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4659161 c.21G>A p.L7L Chr.14: 58200238-58200238 on Build GRCh38
1607776 c.34G>A p.E12K Chr.14: 58200251-58200251 on Build GRCh38
4593637 c.35A>G p.E12G Chr.14: 58200252-58200252 on Build GRCh38
1283243 c.45delG p.V16fs*2 Chr.14: 58200262-58200262 on Build GRCh38
238565 c.77A>G p.K26R Chr.14: 58200294-58200294 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs572361971 Chr.14: 58200268-58200268 on Build GRCh38
rs370132859 Chr.14: 58200277-58200277 on Build GRCh38
rs952457397 Chr.14: 58200139-58200139 on Build GRCh38
rs983349594 Chr.14: 58200339-58200339 on Build GRCh38
rs778042043 Chr.14: 58200197-58200197 on Build GRCh38
rs768827180 Chr.14: 58200326-58200326 on Build GRCh38
rs140960545 Chr.14: 58200294-58200294 on Build GRCh38
rs749793150 Chr.14: 58200169-58200169 on Build GRCh38
rs944818509 Chr.14: 58200504-58200504 on Build GRCh38
rs756463513 Chr.14: 58200194-58200194 on Build GRCh38
rs894844038 Chr.14: 58200455-58200455 on Build GRCh38
rs765840778 Chr.14: 58200249-58200249 on Build GRCh38
rs757909069 Chr.14: 58200187-58200187 on Build GRCh38
rs938937573 Chr.14: 58200421-58200421 on Build GRCh38
rs76669080 Chr.14: 58200252-58200252 on Build GRCh38
rs1019166703 Chr.14: 58200110-58200110 on Build GRCh38
rs149639887 Chr.14: 58200350-58200350 on Build GRCh38
rs61975916 Chr.14: 58200164-58200164 on Build GRCh38
rs529665709 Chr.14: 58200491-58200491 on Build GRCh38
rs764596951 Chr.14: 58200372-58200372 on Build GRCh38
rs369639625 Chr.14: 58200175-58200175 on Build GRCh38
rs900771732 Chr.14: 58200114-58200114 on Build GRCh38
rs774626407 Chr.14: 58200172-58200172 on Build GRCh38
rs762920561 Chr.14: 58200338-58200338 on Build GRCh38
rs765061227 Chr.14: 58200346-58200347 on Build GRCh38
rs187781198 Chr.14: 58200206-58200206 on Build GRCh38
rs764713122 Chr.14: 58200183-58200183 on Build GRCh38
rs1009322563 Chr.14: 58200105-58200105 on Build GRCh38
rs199915707 Chr.14: 58200178-58200178 on Build GRCh38
rs750002001 Chr.14: 58200185-58200185 on Build GRCh38
rs765092950 Chr.14: 58200230-58200230 on Build GRCh38
rs1014774163 Chr.14: 58200502-58200502 on Build GRCh38
rs776916390 Chr.14: 58200327-58200327 on Build GRCh38
rs761538904 Chr.14: 58200332-58200332 on Build GRCh38
rs1055386530 Chr.14: 58200082-58200082 on Build GRCh38
rs1027808344 Chr.14: 58200136-58200136 on Build GRCh38
rs778914739 Chr.14: 58200461-58200461 on Build GRCh38
rs762403667 Chr.14: 58200235-58200235 on Build GRCh38
rs541080712 Chr.14: 58200468-58200468 on Build GRCh38
rs768475040 Chr.14: 58200215-58200215 on Build GRCh38
rs112877891 Chr.14: 58200094-58200094 on Build GRCh38
rs529955430 Chr.14: 58200102-58200102 on Build GRCh38
rs1003820049 Chr.14: 58200264-58200264 on Build GRCh38
rs773722240 Chr.14: 58200168-58200168 on Build GRCh38
rs753775802 Chr.14: 58200179-58200179 on Build GRCh38
rs761786320 Chr.14: 58200180-58200180 on Build GRCh38
rs75449460 Chr.14: 58200254-58200254 on Build GRCh38
rs907438424 Chr.14: 58200419-58200419 on Build GRCh38
rs761735074 Chr.14: 58200222-58200222 on Build GRCh38
rs571631584 Chr.14: 58200195-58200195 on Build GRCh38
rs996865668 Chr.14: 58200293-58200293 on Build GRCh38
rs1056052294 Chr.14: 58200428-58200428 on Build GRCh38
rs532105233 Chr.14: 58200454-58200454 on Build GRCh38
rs553869088 Chr.14: 58200262-58200262 on Build GRCh38
rs769599984 Chr.14: 58200333-58200333 on Build GRCh38
rs779509782 Chr.14: 58200286-58200286 on Build GRCh38
rs990285498 Chr.14: 58200389-58200389 on Build GRCh38
rs750376218 Chr.14: 58200234-58200234 on Build GRCh38
rs946505291 Chr.14: 58200440-58200440 on Build GRCh38
rs373681203 Chr.14: 58200199-58200199 on Build GRCh38
rs937829992 Chr.14: 58200081-58200081 on Build GRCh38
rs191136748 Chr.14: 58200309-58200309 on Build GRCh38
rs965964732 Chr.14: 58200352-58200352 on Build GRCh38
rs11557247 Chr.14: 58200275-58200275 on Build GRCh38
rs772245659 Chr.14: 58200295-58200295 on Build GRCh38
rs772298780 Chr.14: 58200209-58200209 on Build GRCh38
rs757353955 Chr.14: 58200416-58200416 on Build GRCh38
rs915163222 Chr.14: 58200432-58200432 on Build GRCh38
rs776347173 Chr.14: 58200216-58200216 on Build GRCh38
rs775585156 Chr.14: 58200176-58200176 on Build GRCh38
rs932057004 Chr.14: 58200373-58200373 on Build GRCh38
rs1042402032 Chr.14: 58200097-58200097 on Build GRCh38
rs548520557 Chr.14: 58200495-58200495 on Build GRCh38
rs754183660 Chr.14: 58200274-58200274 on Build GRCh38
rs754334299 Chr.14: 58200460-58200460 on Build GRCh38
rs1012035360 Chr.14: 58200284-58200284 on Build GRCh38
rs569889056 Chr.14: 58200162-58200162 on Build GRCh38
rs184552003 Chr.14: 58200405-58200405 on Build GRCh38
rs1040802881 Chr.14: 58200278-58200278 on Build GRCh38
rs556730507 Chr.14: 58200192-58200192 on Build GRCh38
rs373158150 Chr.14: 58200411-58200411 on Build GRCh38
rs779564612 Chr.14: 58200202-58200202 on Build GRCh38
rs983876007 Chr.14: 58200140-58200140 on Build GRCh38
rs773094756 Chr.14: 58200337-58200337 on Build GRCh38
rs976047939 Chr.14: 58200370-58200370 on Build GRCh38
rs977940150 Chr.14: 58200447-58200447 on Build GRCh38
rs961436114 Chr.14: 58200367-58200367 on Build GRCh38
rs113130132 Chr.14: 58200363-58200363 on Build GRCh38
rs548458939 Chr.14: 58200132-58200132 on Build GRCh38
rs760466397 Chr.14: 58200177-58200177 on Build GRCh38
rs754640516 Chr.14: 58200253-58200253 on Build GRCh38
rs879834624 Chr.14: 58200466-58200466 on Build GRCh38
rs924033012 Chr.14: 58200471-58200471 on Build GRCh38
rs375794354 Chr.14: 58200213-58200213 on Build GRCh38
rs775739906 Chr.14: 58200210-58200210 on Build GRCh38
rs371641363 Chr.14: 58200490-58200490 on Build GRCh38

View Full Product Details