Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 58641473-58642014 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DACT1
Entrez Gene ID
51339
Gene Name
dishevelled binding antagonist of beta catenin 1
Gene Aliases
DAPPER, DAPPER1, DPR1, FRODO, HDPR1, THYEX3
UniGene
Hs.48950
Gene Chromosome Location
Chr.14: 58634068-58648321 on Build GRCh38
Transcripts
NM_001079520.1 NM_016651.5 NR_046093.1 NR_046095.1 XM_006720167.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1370273 c.483T>G p.F161L Chr.14: 58641596-58641596 on Build GRCh38
5784005 c.596C>T p.A199V Chr.14: 58641709-58641709 on Build GRCh38
7173850 c.527C>G p.S176C Chr.14: 58641640-58641640 on Build GRCh38
1726493 c.621C>G p.C207W Chr.14: 58641734-58641734 on Build GRCh38
2246305 c.495T>C p.S165S Chr.14: 58641608-58641608 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1000561370 Chr.14: 58641858-58641858 on Build GRCh38
rs942068352 Chr.14: 58641647-58641647 on Build GRCh38
rs201729366 Chr.14: 58641695-58641695 on Build GRCh38
rs986283767 Chr.14: 58641939-58641939 on Build GRCh38
rs35083114 Chr.14: 58641879-58641880 on Build GRCh38
rs947698733 Chr.14: 58641537-58641537 on Build GRCh38
rs765864644 Chr.14: 58641798-58641798 on Build GRCh38
rs963940129 Chr.14: 58641961-58641961 on Build GRCh38
rs1044994807 Chr.14: 58641799-58641799 on Build GRCh38
rs167481 Chr.14: 58641960-58641960 on Build GRCh38
rs763904196 Chr.14: 58641660-58641660 on Build GRCh38
rs775719546 Chr.14: 58641653-58641653 on Build GRCh38
rs571908843 Chr.14: 58641918-58641919 on Build GRCh38
rs181833601 Chr.14: 58641710-58641710 on Build GRCh38
rs529546661 Chr.14: 58641911-58641911 on Build GRCh38
rs956642389 Chr.14: 58641514-58641514 on Build GRCh38
rs765280159 Chr.14: 58641789-58641789 on Build GRCh38
rs544184336 Chr.14: 58641519-58641519 on Build GRCh38
rs745610660 Chr.14: 58641845-58641845 on Build GRCh38
rs745956242 Chr.14: 58641744-58641744 on Build GRCh38
rs142445185 Chr.14: 58641714-58641714 on Build GRCh38
rs778173028 Chr.14: 58641584-58641584 on Build GRCh38
rs147305451 Chr.14: 58641886-58641886 on Build GRCh38
rs1036297888 Chr.14: 58641631-58641631 on Build GRCh38
rs138731030 Chr.14: 58641513-58641513 on Build GRCh38
rs771827981 Chr.14: 58641907-58641907 on Build GRCh38
rs919217146 Chr.14: 58641978-58641979 on Build GRCh38
rs746797846 Chr.14: 58641535-58641535 on Build GRCh38
rs916323794 Chr.14: 58641912-58641912 on Build GRCh38
rs761689496 Chr.14: 58641786-58641786 on Build GRCh38
rs753556749 Chr.14: 58641683-58641683 on Build GRCh38
rs143942985 Chr.14: 58641698-58641698 on Build GRCh38
rs758191914 Chr.14: 58641542-58641542 on Build GRCh38
rs757924168 Chr.14: 58641707-58641707 on Build GRCh38
rs370417663 Chr.14: 58641790-58641790 on Build GRCh38
rs772334316 Chr.14: 58641644-58641644 on Build GRCh38
rs772103272 Chr.14: 58641757-58641757 on Build GRCh38
rs762582282 Chr.14: 58641792-58641792 on Build GRCh38
rs144708680 Chr.14: 58641734-58641734 on Build GRCh38
rs149436015 Chr.14: 58641920-58641921 on Build GRCh38
rs756457653 Chr.14: 58641726-58641726 on Build GRCh38
rs984541041 Chr.14: 58641946-58641946 on Build GRCh38
rs750013177 Chr.14: 58641705-58641705 on Build GRCh38
rs371861288 Chr.14: 58641558-58641558 on Build GRCh38
rs774451791 Chr.14: 58641613-58641613 on Build GRCh38
rs954951367 Chr.14: 58641589-58641589 on Build GRCh38
rs771491889 Chr.14: 58641736-58641736 on Build GRCh38
rs929394932 Chr.14: 58641982-58641982 on Build GRCh38
rs901646194 Chr.14: 58641646-58641646 on Build GRCh38
rs550532642 Chr.14: 58641925-58641925 on Build GRCh38
rs528728898 Chr.14: 58641877-58641877 on Build GRCh38
rs199525555 Chr.14: 58641764-58641764 on Build GRCh38
rs967370707 Chr.14: 58641807-58641807 on Build GRCh38
rs377198014 Chr.14: 58641721-58641721 on Build GRCh38
rs1036383387 Chr.14: 58641862-58641862 on Build GRCh38
rs768733057 Chr.14: 58641768-58641768 on Build GRCh38
rs111529266 Chr.14: 58641672-58641672 on Build GRCh38
rs1004762150 Chr.14: 58641608-58641608 on Build GRCh38
rs749752608 Chr.14: 58641593-58641593 on Build GRCh38
rs191649775 Chr.14: 58641709-58641709 on Build GRCh38
rs759605025 Chr.14: 58641640-58641640 on Build GRCh38
rs748475527 Chr.14: 58641570-58641570 on Build GRCh38
rs757506328 Chr.14: 58641574-58641574 on Build GRCh38
rs879295610 Chr.14: 58641863-58641863 on Build GRCh38
rs776557560 Chr.14: 58641781-58641781 on Build GRCh38
rs923407392 Chr.14: 58641575-58641575 on Build GRCh38
rs779840984 Chr.14: 58641557-58641557 on Build GRCh38
rs910784617 Chr.14: 58641645-58641645 on Build GRCh38
rs940349259 Chr.14: 58641975-58641975 on Build GRCh38
rs769444915 Chr.14: 58641881-58641882 on Build GRCh38
rs997786026 Chr.14: 58641684-58641684 on Build GRCh38
rs540517662 Chr.14: 58641880-58641880 on Build GRCh38
rs749582603 Chr.14: 58641735-58641735 on Build GRCh38
rs780622148 Chr.14: 58641649-58641649 on Build GRCh38
rs199913166 Chr.14: 58641730-58641730 on Build GRCh38
rs1038087724 Chr.14: 58641666-58641666 on Build GRCh38
rs779376717 Chr.14: 58641738-58641738 on Build GRCh38
rs556194564 Chr.14: 58641617-58641617 on Build GRCh38
rs771403994 Chr.14: 58641600-58641600 on Build GRCh38

View Full Product Details