Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 59322802-59323296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAAM1
Entrez Gene ID
23002
Gene Name
dishevelled associated activator of morphogenesis 1
Gene Aliases
-
UniGene
Hs.19156
Gene Chromosome Location
Chr.14: 59188611-59371405 on Build GRCh38
Transcripts
NM_001270520.1 NM_014992.2 XM_005267430.1 XM_005267431.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6336167 c.561G>T p.A187A Chr.14: 59323012-59323012 on Build GRCh38
2246377 c.527G>A p.R176Q Chr.14: 59322978-59322978 on Build GRCh38
5740111 c.665C>T p.A222V Chr.14: 59323116-59323116 on Build GRCh38
7140244 c.460G>T p.D154Y Chr.14: 59322911-59322911 on Build GRCh38
5758858 c.649A>C p.I217L Chr.14: 59323100-59323100 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs890045593 Chr.14: 59322946-59322946 on Build GRCh38
rs761811745 Chr.14: 59322978-59322978 on Build GRCh38
rs777319376 Chr.14: 59322923-59322923 on Build GRCh38
rs763917911 Chr.14: 59323216-59323216 on Build GRCh38
rs902156580 Chr.14: 59323193-59323193 on Build GRCh38
rs749844960 Chr.14: 59323153-59323153 on Build GRCh38
rs760420558 Chr.14: 59323135-59323135 on Build GRCh38
rs758073917 Chr.14: 59323156-59323156 on Build GRCh38
rs748395859 Chr.14: 59323077-59323077 on Build GRCh38
rs1017044466 Chr.14: 59322991-59322991 on Build GRCh38
rs749616487 Chr.14: 59323264-59323264 on Build GRCh38
rs747355323 Chr.14: 59322937-59322937 on Build GRCh38
rs751860849 Chr.14: 59322981-59322981 on Build GRCh38
rs770367484 Chr.14: 59322956-59322956 on Build GRCh38
rs748543467 Chr.14: 59323164-59323164 on Build GRCh38
rs756037306 Chr.14: 59322918-59322918 on Build GRCh38
rs754756459 Chr.14: 59322888-59322888 on Build GRCh38
rs770457227 Chr.14: 59323093-59323093 on Build GRCh38
rs141883625 Chr.14: 59323207-59323207 on Build GRCh38
rs756705889 Chr.14: 59323040-59323040 on Build GRCh38
rs767407813 Chr.14: 59322994-59322994 on Build GRCh38
rs776405966 Chr.14: 59323221-59323221 on Build GRCh38
rs531992927 Chr.14: 59323180-59323180 on Build GRCh38
rs748734798 Chr.14: 59322925-59322925 on Build GRCh38
rs749794234 Chr.14: 59323102-59323102 on Build GRCh38
rs17096074 Chr.14: 59323174-59323174 on Build GRCh38
rs531300580 Chr.14: 59323138-59323138 on Build GRCh38
rs980120198 Chr.14: 59323172-59323172 on Build GRCh38
rs775874471 Chr.14: 59322825-59322825 on Build GRCh38
rs763284090 Chr.14: 59322867-59322867 on Build GRCh38
rs200378702 Chr.14: 59323262-59323262 on Build GRCh38
rs756593611 Chr.14: 59323167-59323167 on Build GRCh38
rs149324121 Chr.14: 59322962-59322962 on Build GRCh38
rs761753130 Chr.14: 59323230-59323230 on Build GRCh38
rs750637290 Chr.14: 59323012-59323012 on Build GRCh38
rs201077027 Chr.14: 59323160-59323160 on Build GRCh38
rs991000995 Chr.14: 59322917-59322917 on Build GRCh38
rs771178796 Chr.14: 59323111-59323111 on Build GRCh38
rs771611001 Chr.14: 59323059-59323059 on Build GRCh38
rs376496086 Chr.14: 59322842-59322842 on Build GRCh38
rs969334442 Chr.14: 59323140-59323140 on Build GRCh38
rs771717982 Chr.14: 59323233-59323233 on Build GRCh38
rs776830715 Chr.14: 59322950-59322950 on Build GRCh38
rs749978966 Chr.14: 59323242-59323242 on Build GRCh38
rs946478824 Chr.14: 59323154-59323154 on Build GRCh38
rs199631642 Chr.14: 59323270-59323270 on Build GRCh38
rs542286199 Chr.14: 59323253-59323253 on Build GRCh38
rs774506910 Chr.14: 59323194-59323194 on Build GRCh38
rs745574074 Chr.14: 59323045-59323045 on Build GRCh38
rs775357562 Chr.14: 59323130-59323130 on Build GRCh38
rs564090602 Chr.14: 59323139-59323139 on Build GRCh38
rs757156688 Chr.14: 59323150-59323150 on Build GRCh38
rs778249260 Chr.14: 59323168-59323168 on Build GRCh38
rs773808956 Chr.14: 59323098-59323098 on Build GRCh38
rs372804956 Chr.14: 59322911-59322911 on Build GRCh38
rs377002769 Chr.14: 59323117-59323117 on Build GRCh38
rs781630768 Chr.14: 59323076-59323076 on Build GRCh38
rs375119832 Chr.14: 59322885-59322885 on Build GRCh38
rs757790150 Chr.14: 59323268-59323268 on Build GRCh38
rs753375909 Chr.14: 59323035-59323035 on Build GRCh38
rs895420481 Chr.14: 59323228-59323228 on Build GRCh38
rs369329990 Chr.14: 59322910-59322910 on Build GRCh38
rs8022614 Chr.14: 59323009-59323009 on Build GRCh38
rs199730602 Chr.14: 59323010-59323010 on Build GRCh38
rs770572293 Chr.14: 59322929-59322929 on Build GRCh38
rs567041153 Chr.14: 59323208-59323208 on Build GRCh38
rs534551526 Chr.14: 59323255-59323255 on Build GRCh38
rs369988349 Chr.14: 59322858-59322858 on Build GRCh38
rs778262988 Chr.14: 59323042-59323042 on Build GRCh38
rs138885144 Chr.14: 59322940-59322940 on Build GRCh38
rs763089193 Chr.14: 59322977-59322977 on Build GRCh38
rs111578221 Chr.14: 59323236-59323236 on Build GRCh38
rs372952716 Chr.14: 59322992-59322992 on Build GRCh38
rs778495301 Chr.14: 59322932-59322932 on Build GRCh38
rs879260041 Chr.14: 59323107-59323107 on Build GRCh38
rs745984536 Chr.14: 59323272-59323272 on Build GRCh38
rs891989534 Chr.14: 59322875-59322875 on Build GRCh38
rs752154826 Chr.14: 59322851-59322851 on Build GRCh38
rs759856885 Chr.14: 59323118-59323118 on Build GRCh38
rs767573792 Chr.14: 59323120-59323120 on Build GRCh38
rs8021976 Chr.14: 59322868-59322868 on Build GRCh38
rs1003237514 Chr.14: 59323200-59323200 on Build GRCh38
rs35092700 Chr.14: 59322977-59322978 on Build GRCh38
rs375808320 Chr.14: 59322953-59322953 on Build GRCh38
rs866648375 Chr.14: 59323146-59323146 on Build GRCh38

View Full Product Details