Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 59324336-59324883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAAM1
Entrez Gene ID
23002
Gene Name
dishevelled associated activator of morphogenesis 1
Gene Aliases
-
UniGene
Hs.19156
Gene Chromosome Location
Chr.14: 59188611-59371405 on Build GRCh38
Transcripts
NM_001270520.1 NM_014992.2 XM_005267430.1 XM_005267431.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3496614 c.913C>T p.R305C Chr.14: 59324378-59324378 on Build GRCh38
3370008 c.917A>G p.Y306C Chr.14: 59324382-59324382 on Build GRCh38
2246383 c.947T>C p.V316A Chr.14: 59324412-59324412 on Build GRCh38
2246384 c.969C>T p.H323H Chr.14: 59324434-59324434 on Build GRCh38
4390500 c.988A>T p.R330W Chr.14: 59324453-59324453 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150648068 Chr.14: 59324378-59324378 on Build GRCh38
rs995927609 Chr.14: 59324796-59324796 on Build GRCh38
rs1022053558 Chr.14: 59324379-59324379 on Build GRCh38
rs149203032 Chr.14: 59324728-59324728 on Build GRCh38
rs1030895124 Chr.14: 59324808-59324808 on Build GRCh38
rs756342829 Chr.14: 59324414-59324414 on Build GRCh38
rs773480310 Chr.14: 59324501-59324501 on Build GRCh38
rs933464928 Chr.14: 59324657-59324657 on Build GRCh38
rs760303668 Chr.14: 59324481-59324481 on Build GRCh38
rs541967890 Chr.14: 59324691-59324691 on Build GRCh38
rs371946350 Chr.14: 59324534-59324534 on Build GRCh38
rs749049016 Chr.14: 59324432-59324432 on Build GRCh38
rs998896212 Chr.14: 59324790-59324790 on Build GRCh38
rs774385213 Chr.14: 59324441-59324441 on Build GRCh38
rs777552379 Chr.14: 59324846-59324846 on Build GRCh38
rs780499191 Chr.14: 59324365-59324365 on Build GRCh38
rs775062758 Chr.14: 59324477-59324477 on Build GRCh38
rs908866806 Chr.14: 59324605-59324605 on Build GRCh38
rs78937873 Chr.14: 59324512-59324512 on Build GRCh38
rs558287135 Chr.14: 59324582-59324582 on Build GRCh38
rs150115749 Chr.14: 59324502-59324502 on Build GRCh38
rs999849152 Chr.14: 59324433-59324433 on Build GRCh38
rs887195745 Chr.14: 59324837-59324837 on Build GRCh38
rs1004383013 Chr.14: 59324858-59324858 on Build GRCh38
rs940392276 Chr.14: 59324623-59324623 on Build GRCh38
rs1053510805 Chr.14: 59324661-59324661 on Build GRCh38
rs781215515 Chr.14: 59324409-59324409 on Build GRCh38
rs1050554544 Chr.14: 59324686-59324686 on Build GRCh38
rs890364822 Chr.14: 59324736-59324736 on Build GRCh38
rs202088104 Chr.14: 59324475-59324475 on Build GRCh38
rs1027503801 Chr.14: 59324472-59324472 on Build GRCh38
rs1007460154 Chr.14: 59324766-59324766 on Build GRCh38
rs375190688 Chr.14: 59324767-59324767 on Build GRCh38
rs751660141 Chr.14: 59324392-59324392 on Build GRCh38
rs371132871 Chr.14: 59324434-59324434 on Build GRCh38
rs926330763 Chr.14: 59324634-59324634 on Build GRCh38
rs992006326 Chr.14: 59324506-59324506 on Build GRCh38
rs1023465159 Chr.14: 59324857-59324857 on Build GRCh38
rs748383513 Chr.14: 59324411-59324411 on Build GRCh38
rs896423016 Chr.14: 59324819-59324819 on Build GRCh38
rs140088700 Chr.14: 59324408-59324408 on Build GRCh38
rs972712119 Chr.14: 59324633-59324633 on Build GRCh38
rs10141362 Chr.14: 59324710-59324710 on Build GRCh38
rs763745522 Chr.14: 59324489-59324489 on Build GRCh38
rs745780967 Chr.14: 59324474-59324474 on Build GRCh38
rs752612001 Chr.14: 59324641-59324641 on Build GRCh38
rs756149159 Chr.14: 59324630-59324630 on Build GRCh38
rs1051586056 Chr.14: 59324725-59324725 on Build GRCh38
rs182730720 Chr.14: 59324775-59324775 on Build GRCh38
rs777438140 Chr.14: 59324416-59324416 on Build GRCh38

View Full Product Details