Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 60118092-60118587 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCNX4
Entrez Gene ID
64430
Gene Name
pecanex homolog 4 (Drosophila)
Gene Aliases
C14orf135, FBP2, PCNXL4
UniGene
Hs.509499
Gene Chromosome Location
Chr.14: 60091276-60142976 on Build GRCh38
Transcripts
NM_022495.5 XM_005268009.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4561260 c.877G>T p.V293F Chr.14: 60118329-60118329 on Build GRCh38
3754027 c.961A>G p.K321E Chr.14: 60118413-60118413 on Build GRCh38
4443860 c.1024G>A p.V342M Chr.14: 60118476-60118476 on Build GRCh38
3496637 c.1597C>T p.R533C Chr.14: 60118347-60118347 on Build GRCh38
1238341 c.1676G>A p.R559Q Chr.14: 60118426-60118426 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544281582 Chr.14: 60118193-60118193 on Build GRCh38
rs751038052 Chr.14: 60118347-60118347 on Build GRCh38
rs773266496 Chr.14: 60118341-60118341 on Build GRCh38
rs745721887 Chr.14: 60118423-60118423 on Build GRCh38
rs748449922 Chr.14: 60118476-60118476 on Build GRCh38
rs889295960 Chr.14: 60118462-60118462 on Build GRCh38
rs367714052 Chr.14: 60118317-60118317 on Build GRCh38
rs756259505 Chr.14: 60118504-60118504 on Build GRCh38
rs1054676754 Chr.14: 60118129-60118129 on Build GRCh38
rs308998 Chr.14: 60118413-60118413 on Build GRCh38
rs367684975 Chr.14: 60118559-60118559 on Build GRCh38
rs760234133 Chr.14: 60118452-60118452 on Build GRCh38
rs747288236 Chr.14: 60118326-60118326 on Build GRCh38
rs574566736 Chr.14: 60118389-60118389 on Build GRCh38
rs540767851 Chr.14: 60118306-60118306 on Build GRCh38
rs75318366 Chr.14: 60118434-60118434 on Build GRCh38
rs370693942 Chr.14: 60118468-60118468 on Build GRCh38
rs751000940 Chr.14: 60118375-60118375 on Build GRCh38
rs201294965 Chr.14: 60118431-60118431 on Build GRCh38
rs891191865 Chr.14: 60118225-60118225 on Build GRCh38
rs769687066 Chr.14: 60118406-60118406 on Build GRCh38
rs549238617 Chr.14: 60118348-60118348 on Build GRCh38
rs374537948 Chr.14: 60118471-60118471 on Build GRCh38
rs1039749793 Chr.14: 60118428-60118428 on Build GRCh38
rs770651679 Chr.14: 60118515-60118515 on Build GRCh38
rs144345462 Chr.14: 60118564-60118564 on Build GRCh38
rs747116922 Chr.14: 60118383-60118383 on Build GRCh38
rs372544161 Chr.14: 60118262-60118266 on Build GRCh38
rs766616903 Chr.14: 60118303-60118304 on Build GRCh38
rs760419187 Chr.14: 60118536-60118536 on Build GRCh38
rs769419691 Chr.14: 60118339-60118339 on Build GRCh38
rs912533453 Chr.14: 60118421-60118421 on Build GRCh38
rs967034628 Chr.14: 60118280-60118280 on Build GRCh38
rs78784092 Chr.14: 60118117-60118117 on Build GRCh38
rs766784651 Chr.14: 60118547-60118547 on Build GRCh38
rs750726110 Chr.14: 60118456-60118456 on Build GRCh38
rs114135575 Chr.14: 60118508-60118508 on Build GRCh38
rs1021802354 Chr.14: 60118254-60118254 on Build GRCh38
rs761724318 Chr.14: 60118334-60118334 on Build GRCh38
rs377696401 Chr.14: 60118382-60118382 on Build GRCh38
rs990365375 Chr.14: 60118231-60118231 on Build GRCh38
rs186619189 Chr.14: 60118355-60118355 on Build GRCh38
rs749056022 Chr.14: 60118513-60118513 on Build GRCh38
rs560297842 Chr.14: 60118314-60118314 on Build GRCh38
rs758813454 Chr.14: 60118376-60118376 on Build GRCh38
rs757512982 Chr.14: 60118313-60118313 on Build GRCh38
rs199774597 Chr.14: 60118278-60118278 on Build GRCh38
rs760596087 Chr.14: 60118409-60118410 on Build GRCh38
rs865978934 Chr.14: 60118381-60118381 on Build GRCh38
rs369608124 Chr.14: 60118408-60118408 on Build GRCh38
rs190033446 Chr.14: 60118555-60118555 on Build GRCh38
rs192298073 Chr.14: 60118385-60118385 on Build GRCh38
rs946043294 Chr.14: 60118209-60118209 on Build GRCh38
rs112724029 Chr.14: 60118118-60118119 on Build GRCh38
rs899629097 Chr.14: 60118237-60118237 on Build GRCh38
rs1008242521 Chr.14: 60118228-60118228 on Build GRCh38
rs778147686 Chr.14: 60118294-60118294 on Build GRCh38
rs763886002 Chr.14: 60118556-60118557 on Build GRCh38
rs928432829 Chr.14: 60118324-60118324 on Build GRCh38
rs200008756 Chr.14: 60118525-60118525 on Build GRCh38
rs775486238 Chr.14: 60118366-60118367 on Build GRCh38
rs769395621 Chr.14: 60118117-60118118 on Build GRCh38
rs758749937 Chr.14: 60118460-60118460 on Build GRCh38
rs774273762 Chr.14: 60118516-60118516 on Build GRCh38
rs773383771 Chr.14: 60118281-60118281 on Build GRCh38
rs201321269 Chr.14: 60118475-60118475 on Build GRCh38
rs371462329 Chr.14: 60118563-60118563 on Build GRCh38
rs201437114 Chr.14: 60118190-60118196 on Build GRCh38
rs765963274 Chr.14: 60118345-60118345 on Build GRCh38
rs779144130 Chr.14: 60118369-60118369 on Build GRCh38
rs767177480 Chr.14: 60118351-60118351 on Build GRCh38
rs777530172 Chr.14: 60118257-60118257 on Build GRCh38
rs564562636 Chr.14: 60118244-60118244 on Build GRCh38
rs774220888 Chr.14: 60118426-60118426 on Build GRCh38
rs75497201 Chr.14: 60118118-60118118 on Build GRCh38
rs765687054 Chr.14: 60118453-60118453 on Build GRCh38
rs763095385 Chr.14: 60118288-60118290 on Build GRCh38
rs575679780 Chr.14: 60118145-60118145 on Build GRCh38
rs995748266 Chr.14: 60118238-60118238 on Build GRCh38
rs189013229 Chr.14: 60118470-60118470 on Build GRCh38
rs780834392 Chr.14: 60118544-60118544 on Build GRCh38
rs367609970 Chr.14: 60118419-60118426 on Build GRCh38
rs775193195 Chr.14: 60118441-60118441 on Build GRCh38
rs763686044 Chr.14: 60118540-60118540 on Build GRCh38
rs555713799 Chr.14: 60118124-60118124 on Build GRCh38
rs374433271 Chr.14: 60118342-60118342 on Build GRCh38
rs868420819 Chr.14: 60118478-60118478 on Build GRCh38
rs781056587 Chr.14: 60118500-60118500 on Build GRCh38
rs780272829 Chr.14: 60118325-60118325 on Build GRCh38
rs998007163 Chr.14: 60118447-60118447 on Build GRCh38
rs139085229 Chr.14: 60118422-60118426 on Build GRCh38
rs578026729 Chr.14: 60118263-60118263 on Build GRCh38
rs776952726 Chr.14: 60118327-60118327 on Build GRCh38
rs772263632 Chr.14: 60118319-60118319 on Build GRCh38
rs757648965 Chr.14: 60118358-60118358 on Build GRCh38
rs754601579 Chr.14: 60118285-60118285 on Build GRCh38
rs940698015 Chr.14: 60118422-60118422 on Build GRCh38
rs753777581 Chr.14: 60118264-60118264 on Build GRCh38
rs953695141 Chr.14: 60118205-60118205 on Build GRCh38

View Full Product Details