Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 64268412-64268933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESR2
Entrez Gene ID
2100
Gene Name
estrogen receptor 2
Gene Aliases
ER-BETA, ESR-BETA, ESRB, ESTRB, Erb, NR3A2
UniGene
Hs.660607
Gene Chromosome Location
Chr.14: 64226712-64338631 on Build GRCh38
Transcripts
NM_001040275.1 NM_001214902.1 NM_001271876.1 NM_001271877.1 NM_001291712.1 NM_001291723.1 NM_001437.2 NR_073496.1 NR_073497.1 XM_017021079.1 XM_017021080.1 XM_017021081.1 XM_017021082.1 XM_017021083.1 XM_017021084.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3815096 c.631G>A p.E211K Chr.14: 64268816-64268816 on Build GRCh38
5362397 c.584A>C p.K195T Chr.14: 64268863-64268863 on Build GRCh38
6656410 c.565A>C p.N189H Chr.14: 64268882-64268882 on Build GRCh38
5000399 c.547T>G p.Y183D Chr.14: 64268900-64268900 on Build GRCh38
6656412 c.565A>C p.N189H Chr.14: 64268882-64268882 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2228481 Chr.14: 64268817-64268817 on Build GRCh38
rs184492441 Chr.14: 64268771-64268771 on Build GRCh38
rs754945292 Chr.14: 64268887-64268887 on Build GRCh38
rs765370070 Chr.14: 64268800-64268800 on Build GRCh38
rs761261331 Chr.14: 64268838-64268838 on Build GRCh38
rs910614673 Chr.14: 64268438-64268438 on Build GRCh38
rs889002608 Chr.14: 64268680-64268680 on Build GRCh38
rs141513906 Chr.14: 64268496-64268496 on Build GRCh38
rs572425023 Chr.14: 64268466-64268466 on Build GRCh38
rs752555445 Chr.14: 64268910-64268910 on Build GRCh38
rs1056297317 Chr.14: 64268655-64268655 on Build GRCh38
rs1007892425 Chr.14: 64268690-64268690 on Build GRCh38
rs968057666 Chr.14: 64268692-64268692 on Build GRCh38
rs1051729034 Chr.14: 64268843-64268843 on Build GRCh38
rs980925917 Chr.14: 64268527-64268527 on Build GRCh38
rs750554682 Chr.14: 64268802-64268802 on Build GRCh38
rs768839285 Chr.14: 64268855-64268855 on Build GRCh38
rs748369077 Chr.14: 64268596-64268596 on Build GRCh38
rs986225210 Chr.14: 64268437-64268437 on Build GRCh38
rs139479509 Chr.14: 64268865-64268865 on Build GRCh38
rs751392863 Chr.14: 64268877-64268877 on Build GRCh38
rs968416904 Chr.14: 64268442-64268442 on Build GRCh38
rs530138782 Chr.14: 64268529-64268529 on Build GRCh38
rs753951395 Chr.14: 64268788-64268788 on Build GRCh38
rs767173223 Chr.14: 64268897-64268897 on Build GRCh38
rs772568096 Chr.14: 64268834-64268834 on Build GRCh38
rs541573392 Chr.14: 64268491-64268491 on Build GRCh38
rs776143898 Chr.14: 64268764-64268764 on Build GRCh38
rs552474098 Chr.14: 64268677-64268677 on Build GRCh38
rs747686242 Chr.14: 64268819-64268819 on Build GRCh38
rs919374831 Chr.14: 64268832-64268832 on Build GRCh38
rs773394073 Chr.14: 64268869-64268869 on Build GRCh38
rs918210062 Chr.14: 64268489-64268489 on Build GRCh38
rs750091675 Chr.14: 64268904-64268906 on Build GRCh38
rs368896463 Chr.14: 64268675-64268675 on Build GRCh38
rs1009494091 Chr.14: 64268674-64268674 on Build GRCh38
rs145278854 Chr.14: 64268867-64268867 on Build GRCh38
rs781052752 Chr.14: 64268818-64268818 on Build GRCh38
rs556956556 Chr.14: 64268816-64268816 on Build GRCh38
rs985074529 Chr.14: 64268811-64268811 on Build GRCh38
rs180722269 Chr.14: 64268551-64268551 on Build GRCh38
rs563819798 Chr.14: 64268597-64268597 on Build GRCh38
rs992818214 Chr.14: 64268479-64268479 on Build GRCh38
rs775804915 Chr.14: 64268772-64268772 on Build GRCh38
rs1003365702 Chr.14: 64268609-64268609 on Build GRCh38
rs780025522 Chr.14: 64268760-64268761 on Build GRCh38
rs778719275 Chr.14: 64268539-64268539 on Build GRCh38
rs371856990 Chr.14: 64268828-64268828 on Build GRCh38
rs904386265 Chr.14: 64268715-64268715 on Build GRCh38
rs998587399 Chr.14: 64268753-64268753 on Build GRCh38
rs760053106 Chr.14: 64268858-64268858 on Build GRCh38
rs1021883717 Chr.14: 64268440-64268440 on Build GRCh38
rs552876311 Chr.14: 64268878-64268878 on Build GRCh38
rs2228481 Chr.14: 64268817-64268817 on Build GRCh38
rs1017729034 Chr.14: 64268703-64268703 on Build GRCh38
rs941002434 Chr.14: 64268560-64268560 on Build GRCh38
rs201208686 Chr.14: 64268672-64268672 on Build GRCh38
rs746965258 Chr.14: 64268792-64268794 on Build GRCh38
rs539747077 Chr.14: 64268857-64268857 on Build GRCh38
rs772586318 Chr.14: 64268759-64268759 on Build GRCh38
rs929573616 Chr.14: 64268467-64268467 on Build GRCh38
rs1044811411 Chr.14: 64268571-64268571 on Build GRCh38
rs892311182 Chr.14: 64268886-64268886 on Build GRCh38
rs548338296 Chr.14: 64268449-64268449 on Build GRCh38
rs942093033 Chr.14: 64268595-64268595 on Build GRCh38
rs534381951 Chr.14: 64268741-64268741 on Build GRCh38
rs776039661 Chr.14: 64268835-64268835 on Build GRCh38
rs3742614 Chr.14: 64268778-64268778 on Build GRCh38
rs746498138 Chr.14: 64268713-64268713 on Build GRCh38
rs761893224 Chr.14: 64268861-64268861 on Build GRCh38
rs374870812 Chr.14: 64268868-64268868 on Build GRCh38
rs909370443 Chr.14: 64268824-64268824 on Build GRCh38
rs74808355 Chr.14: 64268814-64268814 on Build GRCh38
rs368924653 Chr.14: 64268827-64268827 on Build GRCh38
rs763191319 Chr.14: 64268874-64268874 on Build GRCh38
rs550457044 Chr.14: 64268553-64268553 on Build GRCh38
rs760613053 Chr.14: 64268457-64268460 on Build GRCh38
rs532886837 Chr.14: 64268658-64268658 on Build GRCh38
rs766405281 Chr.14: 64268876-64268876 on Build GRCh38
rs75350477 Chr.14: 64268701-64268701 on Build GRCh38
rs770100746 Chr.14: 64268909-64268909 on Build GRCh38
rs959718396 Chr.14: 64268469-64268469 on Build GRCh38
rs369253565 Chr.14: 64268872-64268872 on Build GRCh38
rs748841139 Chr.14: 64268833-64268833 on Build GRCh38
rs761014695 Chr.14: 64268776-64268776 on Build GRCh38

View Full Product Details