Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 64471848-64472399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP5
Entrez Gene ID
9495
Gene Name
A-kinase anchoring protein 5
Gene Aliases
AKAP75, AKAP79, H21
UniGene
Hs.656683
Gene Chromosome Location
Chr.14: 64465499-64474503 on Build GRCh38
Transcripts
NM_004857.3 NM_001304508.1 XM_006720250.3

Gene Symbol
ZBTB25
Entrez Gene ID
7597
Gene Name
zinc finger and BTB domain containing 25
Gene Aliases
C14orf51, KUP, ZNF46
UniGene
Hs.654571
Gene Chromosome Location
Chr.14: 64447172-64505213 on Build GRCh38
Transcripts
NM_004857.3 NM_001304508.1 XM_006720250.3

SNP Detail

SNP ID Chromosome Location
rs530428219 Chr.14: 64472023-64472023 on Build GRCh38
rs1006515761 Chr.14: 64472179-64472179 on Build GRCh38
rs986167010 Chr.14: 64471882-64471882 on Build GRCh38
rs777206208 Chr.14: 64472371-64472371 on Build GRCh38
rs922630183 Chr.14: 64472260-64472260 on Build GRCh38
rs1000036268 Chr.14: 64472171-64472171 on Build GRCh38
rs1057330251 Chr.14: 64472041-64472041 on Build GRCh38
rs550397356 Chr.14: 64472333-64472333 on Build GRCh38
rs541433478 Chr.14: 64471955-64471955 on Build GRCh38
rs775389394 Chr.14: 64472025-64472025 on Build GRCh38
rs898044940 Chr.14: 64472223-64472223 on Build GRCh38
rs964111600 Chr.14: 64472189-64472189 on Build GRCh38
rs924483108 Chr.14: 64471998-64471998 on Build GRCh38
rs910046051 Chr.14: 64472331-64472331 on Build GRCh38
rs550868969 Chr.14: 64472225-64472225 on Build GRCh38
rs1001166263 Chr.14: 64472026-64472026 on Build GRCh38
rs769134019 Chr.14: 64472070-64472070 on Build GRCh38
rs61985664 Chr.14: 64472268-64472268 on Build GRCh38
rs888092456 Chr.14: 64472174-64472174 on Build GRCh38
rs895711742 Chr.14: 64472086-64472086 on Build GRCh38
rs976730709 Chr.14: 64471914-64471914 on Build GRCh38
rs531113472 Chr.14: 64472098-64472098 on Build GRCh38
rs1008038365 Chr.14: 64472251-64472251 on Build GRCh38
rs561384421 Chr.14: 64471968-64471968 on Build GRCh38
rs1036606224 Chr.14: 64472206-64472206 on Build GRCh38
rs930248868 Chr.14: 64471967-64471967 on Build GRCh38
rs951620798 Chr.14: 64472273-64472273 on Build GRCh38
rs983470151 Chr.14: 64472311-64472311 on Build GRCh38
rs980970085 Chr.14: 64472365-64472365 on Build GRCh38
rs968683569 Chr.14: 64472366-64472366 on Build GRCh38
rs968630574 Chr.14: 64471986-64471986 on Build GRCh38
rs953502046 Chr.14: 64472250-64472250 on Build GRCh38
rs370156561 Chr.14: 64472349-64472349 on Build GRCh38
rs938538031 Chr.14: 64472032-64472032 on Build GRCh38
rs761198926 Chr.14: 64472145-64472147 on Build GRCh38
rs1030892663 Chr.14: 64472240-64472240 on Build GRCh38
rs376346635 Chr.14: 64472005-64472005 on Build GRCh38
rs145434363 Chr.14: 64471919-64471919 on Build GRCh38
rs200963387 Chr.14: 64472309-64472309 on Build GRCh38
rs977306774 Chr.14: 64471992-64471992 on Build GRCh38
rs762683750 Chr.14: 64472138-64472138 on Build GRCh38
rs910067864 Chr.14: 64471971-64471971 on Build GRCh38
rs1018742778 Chr.14: 64472186-64472186 on Build GRCh38
rs74058133 Chr.14: 64471932-64471932 on Build GRCh38

View Full Product Details