Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 64472621-64473122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP5
Entrez Gene ID
9495
Gene Name
A-kinase anchoring protein 5
Gene Aliases
AKAP75, AKAP79, H21
UniGene
Hs.656683
Gene Chromosome Location
Chr.14: 64465499-64474503 on Build GRCh38
Transcripts
NM_004857.3 NM_001304508.1 XM_006720250.3

Gene Symbol
ZBTB25
Entrez Gene ID
7597
Gene Name
zinc finger and BTB domain containing 25
Gene Aliases
C14orf51, KUP, ZNF46
UniGene
Hs.654571
Gene Chromosome Location
Chr.14: 64447172-64505213 on Build GRCh38
Transcripts
NM_004857.3 NM_001304508.1 XM_006720250.3

SNP Detail

SNP ID Chromosome Location
rs754143235 Chr.14: 64472738-64472738 on Build GRCh38
rs935958215 Chr.14: 64472824-64472824 on Build GRCh38
rs933626919 Chr.14: 64472919-64472919 on Build GRCh38
rs887198801 Chr.14: 64472950-64472951 on Build GRCh38
rs1022834925 Chr.14: 64472733-64472733 on Build GRCh38
rs539271621 Chr.14: 64472759-64472759 on Build GRCh38
rs1016953850 Chr.14: 64472992-64472992 on Build GRCh38
rs899712292 Chr.14: 64472869-64472871 on Build GRCh38
rs570341903 Chr.14: 64472750-64472750 on Build GRCh38
rs779703546 Chr.14: 64472647-64472647 on Build GRCh38
rs889339310 Chr.14: 64472952-64472952 on Build GRCh38
rs970286564 Chr.14: 64472797-64472797 on Build GRCh38
rs922249086 Chr.14: 64472894-64472894 on Build GRCh38
rs558594128 Chr.14: 64472887-64472887 on Build GRCh38
rs960015122 Chr.14: 64472693-64472693 on Build GRCh38
rs557350754 Chr.14: 64472948-64472949 on Build GRCh38
rs998832201 Chr.14: 64472681-64472681 on Build GRCh38
rs909636558 Chr.14: 64472848-64472848 on Build GRCh38
rs541023606 Chr.14: 64473091-64473091 on Build GRCh38
rs943542417 Chr.14: 64473020-64473020 on Build GRCh38
rs917005470 Chr.14: 64472697-64472697 on Build GRCh38
rs1004591082 Chr.14: 64472979-64472979 on Build GRCh38
rs184656281 Chr.14: 64472897-64472897 on Build GRCh38
rs1041397326 Chr.14: 64472771-64472771 on Build GRCh38
rs983919846 Chr.14: 64472808-64472808 on Build GRCh38
rs555478747 Chr.14: 64472859-64472860 on Build GRCh38
rs993133699 Chr.14: 64472715-64472715 on Build GRCh38
rs904577768 Chr.14: 64472663-64472663 on Build GRCh38
rs939729372 Chr.14: 64472861-64472861 on Build GRCh38
rs1052438472 Chr.14: 64472950-64472950 on Build GRCh38
rs1031606462 Chr.14: 64472684-64472684 on Build GRCh38
rs1043182155 Chr.14: 64473069-64473069 on Build GRCh38
rs1029660855 Chr.14: 64472938-64472938 on Build GRCh38
rs945669524 Chr.14: 64472724-64472724 on Build GRCh38
rs916219420 Chr.14: 64472649-64472650 on Build GRCh38

View Full Product Details