Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 64934900-64935398 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHURC1
Entrez Gene ID
91612
Gene Name
churchill domain containing 1
Gene Aliases
C14orf52, My015, chch
UniGene
Hs.731537
Gene Chromosome Location
Chr.14: 64914361-64935368 on Build GRCh38
Transcripts
NM_001204063.1 NM_001204064.1 NM_145165.3 NM_001202558.1 NM_001202559.1

Gene Symbol
CHURC1-FNTB
Entrez Gene ID
100529261
Gene Name
CHURC1-FNTB readthrough
Gene Aliases
-
UniGene
Hs.325531
Gene Chromosome Location
Chr.14: 64914361-65062655 on Build GRCh38
Transcripts
NM_001204063.1 NM_001204064.1 NM_145165.3 NM_001202558.1 NM_001202559.1

SNP Detail

SNP ID Chromosome Location
rs1036396244 Chr.14: 64935329-64935330 on Build GRCh38
rs890444603 Chr.14: 64935324-64935328 on Build GRCh38
rs1025402329 Chr.14: 64935319-64935319 on Build GRCh38
rs190168419 Chr.14: 64935119-64935119 on Build GRCh38
rs571455687 Chr.14: 64934951-64934951 on Build GRCh38
rs922155361 Chr.14: 64935307-64935307 on Build GRCh38
rs990865711 Chr.14: 64935238-64935238 on Build GRCh38
rs955238614 Chr.14: 64934970-64934970 on Build GRCh38
rs551127374 Chr.14: 64934950-64934950 on Build GRCh38
rs994062685 Chr.14: 64935315-64935315 on Build GRCh38
rs561355240 Chr.14: 64935034-64935034 on Build GRCh38
rs572258287 Chr.14: 64935370-64935371 on Build GRCh38
rs895035233 Chr.14: 64935348-64935349 on Build GRCh38
rs915432768 Chr.14: 64935256-64935256 on Build GRCh38
rs1015867330 Chr.14: 64935185-64935196 on Build GRCh38
rs901302574 Chr.14: 64934976-64934976 on Build GRCh38
rs1054036292 Chr.14: 64935103-64935103 on Build GRCh38
rs995417334 Chr.14: 64935206-64935206 on Build GRCh38
rs563180413 Chr.14: 64935286-64935289 on Build GRCh38
rs541511823 Chr.14: 64935015-64935015 on Build GRCh38
rs759220679 Chr.14: 64935025-64935025 on Build GRCh38
rs1025454566 Chr.14: 64935115-64935115 on Build GRCh38
rs187064221 Chr.14: 64935087-64935087 on Build GRCh38
rs959979167 Chr.14: 64935097-64935097 on Build GRCh38
rs990591646 Chr.14: 64935032-64935032 on Build GRCh38
rs964393466 Chr.14: 64935177-64935177 on Build GRCh38
rs10138667 Chr.14: 64935252-64935252 on Build GRCh38
rs1005594909 Chr.14: 64935346-64935346 on Build GRCh38
rs897000902 Chr.14: 64935342-64935345 on Build GRCh38
rs1021269335 Chr.14: 64935039-64935039 on Build GRCh38
rs1052228409 Chr.14: 64935322-64935322 on Build GRCh38
rs1026824357 Chr.14: 64935214-64935214 on Build GRCh38
rs1043209566 Chr.14: 64935299-64935299 on Build GRCh38
rs10532184 Chr.14: 64935338-64935342 on Build GRCh38
rs976818207 Chr.14: 64935192-64935192 on Build GRCh38
rs563062967 Chr.14: 64935134-64935134 on Build GRCh38
rs895717767 Chr.14: 64935311-64935311 on Build GRCh38
rs922838025 Chr.14: 64935212-64935212 on Build GRCh38
rs753162409 Chr.14: 64935078-64935078 on Build GRCh38
rs911190695 Chr.14: 64935148-64935148 on Build GRCh38
rs1018772728 Chr.14: 64935350-64935350 on Build GRCh38
rs533044387 Chr.14: 64935133-64935133 on Build GRCh38
rs548693498 Chr.14: 64935309-64935309 on Build GRCh38
rs1036011676 Chr.14: 64935092-64935092 on Build GRCh38
rs995260389 Chr.14: 64935345-64935346 on Build GRCh38
rs147183761 Chr.14: 64935303-64935303 on Build GRCh38
rs6745 Chr.14: 64934977-64934977 on Build GRCh38
rs781448382 Chr.14: 64935302-64935302 on Build GRCh38
rs890509430 Chr.14: 64935169-64935169 on Build GRCh38
rs952555220 Chr.14: 64935328-64935328 on Build GRCh38
rs1026875157 Chr.14: 64935346-64935346 on Build GRCh38
rs970910655 Chr.14: 64935271-64935271 on Build GRCh38
rs947093488 Chr.14: 64935281-64935281 on Build GRCh38
rs961192197 Chr.14: 64935204-64935204 on Build GRCh38
rs776216920 Chr.14: 64934978-64934978 on Build GRCh38
rs147039056 Chr.14: 64935083-64935083 on Build GRCh38
rs977867694 Chr.14: 64935277-64935277 on Build GRCh38
rs546795562 Chr.14: 64935223-64935223 on Build GRCh38
rs935599419 Chr.14: 64935229-64935229 on Build GRCh38
rs1042919104 Chr.14: 64935050-64935050 on Build GRCh38
rs572913427 Chr.14: 64935004-64935004 on Build GRCh38
rs10146885 Chr.14: 64935101-64935101 on Build GRCh38
rs1004956395 Chr.14: 64935183-64935183 on Build GRCh38

View Full Product Details