Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 65004163-65004649 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHURC1-FNTB
Entrez Gene ID
100529261
Gene Name
CHURC1-FNTB readthrough
Gene Aliases
-
UniGene
Hs.325531
Gene Chromosome Location
Chr.14: 64914361-65062655 on Build GRCh38
Transcripts
NM_001202558.1 NM_001202559.1 NM_002028.3

Gene Symbol
FNTB
Entrez Gene ID
2342
Gene Name
farnesyltransferase, CAAX box, beta
Gene Aliases
FPTB
UniGene
Hs.325531
Gene Chromosome Location
Chr.14: 64986789-65062652 on Build GRCh38
Transcripts
NM_001202558.1 NM_001202559.1 NM_002028.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3419964 c.162G>T p.K54N Chr.14: 65004266-65004266 on Build GRCh38
3497202 c.174C>T p.V58V Chr.14: 65004278-65004278 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs368879213 Chr.14: 65004215-65004215 on Build GRCh38
rs868751342 Chr.14: 65004290-65004290 on Build GRCh38
rs772690370 Chr.14: 65004282-65004282 on Build GRCh38
rs2296323 Chr.14: 65004468-65004468 on Build GRCh38
rs762014629 Chr.14: 65004233-65004233 on Build GRCh38
rs763052121 Chr.14: 65004246-65004246 on Build GRCh38
rs1037314116 Chr.14: 65004349-65004349 on Build GRCh38
rs777301110 Chr.14: 65004270-65004270 on Build GRCh38
rs760932142 Chr.14: 65004225-65004225 on Build GRCh38
rs890254294 Chr.14: 65004393-65004393 on Build GRCh38
rs774404178 Chr.14: 65004611-65004611 on Build GRCh38
rs930167829 Chr.14: 65004201-65004201 on Build GRCh38
rs1034347953 Chr.14: 65004432-65004432 on Build GRCh38
rs755744657 Chr.14: 65004267-65004267 on Build GRCh38
rs200384517 Chr.14: 65004314-65004314 on Build GRCh38
rs759399892 Chr.14: 65004355-65004355 on Build GRCh38
rs192068491 Chr.14: 65004581-65004581 on Build GRCh38
rs776925757 Chr.14: 65004231-65004231 on Build GRCh38
rs564543218 Chr.14: 65004535-65004535 on Build GRCh38
rs771638373 Chr.14: 65004204-65004204 on Build GRCh38
rs910081324 Chr.14: 65004518-65004518 on Build GRCh38
rs532297504 Chr.14: 65004209-65004209 on Build GRCh38
rs766215092 Chr.14: 65004251-65004251 on Build GRCh38
rs746426903 Chr.14: 65004211-65004211 on Build GRCh38
rs1010085210 Chr.14: 65004395-65004395 on Build GRCh38
rs375096435 Chr.14: 65004348-65004348 on Build GRCh38
rs996048056 Chr.14: 65004373-65004373 on Build GRCh38
rs777015815 Chr.14: 65004304-65004304 on Build GRCh38
rs774119852 Chr.14: 65004340-65004340 on Build GRCh38
rs769257165 Chr.14: 65004198-65004198 on Build GRCh38
rs762886547 Chr.14: 65004334-65004334 on Build GRCh38
rs927984724 Chr.14: 65004472-65004472 on Build GRCh38
rs185874927 Chr.14: 65004257-65004257 on Build GRCh38
rs766346583 Chr.14: 65004338-65004338 on Build GRCh38
rs768861678 Chr.14: 65004303-65004303 on Build GRCh38
rs765358068 Chr.14: 65004235-65004235 on Build GRCh38
rs201402631 Chr.14: 65004551-65004551 on Build GRCh38
rs775985377 Chr.14: 65004224-65004224 on Build GRCh38
rs751384349 Chr.14: 65004252-65004252 on Build GRCh38
rs375425851 Chr.14: 65004210-65004210 on Build GRCh38
rs951604490 Chr.14: 65004512-65004512 on Build GRCh38
rs779423198 Chr.14: 65004205-65004205 on Build GRCh38
rs1042902302 Chr.14: 65004590-65004590 on Build GRCh38
rs145125744 Chr.14: 65004417-65004417 on Build GRCh38
rs543187024 Chr.14: 65004403-65004403 on Build GRCh38
rs796392425 Chr.14: 65004466-65004466 on Build GRCh38
rs746240901 Chr.14: 65004525-65004525 on Build GRCh38
rs780363660 Chr.14: 65004292-65004292 on Build GRCh38
rs879832073 Chr.14: 65004600-65004600 on Build GRCh38
rs889704103 Chr.14: 65004196-65004196 on Build GRCh38
rs958697492 Chr.14: 65004450-65004450 on Build GRCh38
rs752302231 Chr.14: 65004262-65004262 on Build GRCh38
rs982100553 Chr.14: 65004469-65004469 on Build GRCh38
rs777843194 Chr.14: 65004431-65004431 on Build GRCh38
rs917520132 Chr.14: 65004496-65004496 on Build GRCh38
rs771277535 Chr.14: 65004368-65004368 on Build GRCh38
rs1020764866 Chr.14: 65004402-65004402 on Build GRCh38
rs535858782 Chr.14: 65004238-65004238 on Build GRCh38
rs748787797 Chr.14: 65004275-65004275 on Build GRCh38
rs762892819 Chr.14: 65004567-65004567 on Build GRCh38
rs1051690909 Chr.14: 65004390-65004390 on Build GRCh38
rs560735586 Chr.14: 65004321-65004321 on Build GRCh38
rs982949844 Chr.14: 65004513-65004513 on Build GRCh38
rs768934987 Chr.14: 65004226-65004226 on Build GRCh38
rs78424070 Chr.14: 65004533-65004533 on Build GRCh38
rs770136544 Chr.14: 65004327-65004327 on Build GRCh38
rs941806881 Chr.14: 65004317-65004317 on Build GRCh38
rs574189581 Chr.14: 65004356-65004356 on Build GRCh38
rs762174175 Chr.14: 65004319-65004319 on Build GRCh38
rs1000310971 Chr.14: 65004404-65004404 on Build GRCh38

View Full Product Details