Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 65044169-65044691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHURC1-FNTB
Entrez Gene ID
100529261
Gene Name
CHURC1-FNTB readthrough
Gene Aliases
-
UniGene
Hs.325531
Gene Chromosome Location
Chr.14: 64914361-65062655 on Build GRCh38
Transcripts
NM_001202558.1 NM_001202559.1 NM_002028.3 NM_001271069.1 NM_197957.3 NR_039855.1

Gene Symbol
FNTB
Entrez Gene ID
2342
Gene Name
farnesyltransferase, CAAX box, beta
Gene Aliases
FPTB
UniGene
Hs.325531
Gene Chromosome Location
Chr.14: 64986789-65062652 on Build GRCh38
Transcripts
NM_001202558.1 NM_001202559.1 NM_002028.3 NM_001271069.1 NM_197957.3 NR_039855.1

Gene Symbol
MAX
Entrez Gene ID
4149
Gene Name
MYC associated factor X
Gene Aliases
bHLHd4
UniGene
Hs.285354
Gene Chromosome Location
Chr.14: 65006101-65102695 on Build GRCh38
Transcripts
NM_001202558.1 NM_001202559.1 NM_002028.3 NM_001271069.1 NM_197957.3 NR_039855.1

Gene Symbol
MIR4706
Entrez Gene ID
100616490
Gene Name
microRNA 4706
Gene Aliases
mir-4706
UniGene
Gene Chromosome Location
Chr.14: 65044688-65044769 on Build GRCh38
Transcripts
NM_001202558.1 NM_001202559.1 NM_002028.3 NM_001271069.1 NM_197957.3 NR_039855.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
166963 c.848G>A p.R283Q Chr.14: 65044336-65044336 on Build GRCh38
3419965 c.949G>A p.A317T Chr.14: 65044437-65044437 on Build GRCh38
957092 c.939C>T p.R313R Chr.14: 65044427-65044427 on Build GRCh38
5093703 c.948C>T p.H316H Chr.14: 65044436-65044436 on Build GRCh38
4747394 c.912_916delGGCGG p.G306fs*13 Chr.14: 65044400-65044404 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1034077816 Chr.14: 65044297-65044297 on Build GRCh38
rs201482556 Chr.14: 65044283-65044283 on Build GRCh38
rs761059627 Chr.14: 65044359-65044359 on Build GRCh38
rs891948705 Chr.14: 65044550-65044550 on Build GRCh38
rs987847608 Chr.14: 65044254-65044254 on Build GRCh38
rs772958122 Chr.14: 65044428-65044428 on Build GRCh38
rs766520679 Chr.14: 65044483-65044483 on Build GRCh38
rs755920502 Chr.14: 65044664-65044664 on Build GRCh38
rs968176786 Chr.14: 65044572-65044572 on Build GRCh38
rs996388187 Chr.14: 65044223-65044223 on Build GRCh38
rs754152276 Chr.14: 65044362-65044362 on Build GRCh38
rs780923374 Chr.14: 65044490-65044490 on Build GRCh38
rs768166051 Chr.14: 65044327-65044327 on Build GRCh38
rs376882084 Chr.14: 65044425-65044425 on Build GRCh38
rs764500262 Chr.14: 65044437-65044437 on Build GRCh38
rs746211414 Chr.14: 65044262-65044263 on Build GRCh38
rs958916922 Chr.14: 65044311-65044311 on Build GRCh38
rs764555353 Chr.14: 65044360-65044360 on Build GRCh38
rs2296321 Chr.14: 65044248-65044248 on Build GRCh38
rs769572355 Chr.14: 65044427-65044427 on Build GRCh38
rs1022488147 Chr.14: 65044260-65044260 on Build GRCh38
rs112323207 Chr.14: 65044522-65044522 on Build GRCh38
rs890416773 Chr.14: 65044545-65044545 on Build GRCh38
rs370347794 Chr.14: 65044436-65044436 on Build GRCh38
rs887887316 Chr.14: 65044435-65044435 on Build GRCh38
rs980815296 Chr.14: 65044306-65044306 on Build GRCh38
rs574024095 Chr.14: 65044663-65044663 on Build GRCh38
rs182475561 Chr.14: 65044457-65044457 on Build GRCh38
rs776268523 Chr.14: 65044294-65044294 on Build GRCh38
rs538376680 Chr.14: 65044551-65044551 on Build GRCh38
rs560423090 Chr.14: 65044249-65044249 on Build GRCh38
rs770610468 Chr.14: 65044433-65044433 on Build GRCh38
rs777069619 Chr.14: 65044455-65044455 on Build GRCh38
rs757537203 Chr.14: 65044367-65044367 on Build GRCh38
rs754841039 Chr.14: 65044419-65044419 on Build GRCh38
rs750508990 Chr.14: 65044402-65044402 on Build GRCh38
rs1052014970 Chr.14: 65044544-65044547 on Build GRCh38
rs960392513 Chr.14: 65044613-65044613 on Build GRCh38
rs555462654 Chr.14: 65044479-65044479 on Build GRCh38
rs143595505 Chr.14: 65044588-65044588 on Build GRCh38
rs189142573 Chr.14: 65044594-65044594 on Build GRCh38
rs200008673 Chr.14: 65044489-65044489 on Build GRCh38
rs750596724 Chr.14: 65044466-65044466 on Build GRCh38
rs531471652 Chr.14: 65044299-65044299 on Build GRCh38
rs377596512 Chr.14: 65044488-65044488 on Build GRCh38
rs769556051 Chr.14: 65044313-65044313 on Build GRCh38
rs35551013 Chr.14: 65044577-65044578 on Build GRCh38
rs142942714 Chr.14: 65044426-65044426 on Build GRCh38
rs752570490 Chr.14: 65044492-65044492 on Build GRCh38
rs557967841 Chr.14: 65044555-65044555 on Build GRCh38
rs763139733 Chr.14: 65044474-65044474 on Build GRCh38
rs765475648 Chr.14: 65044463-65044463 on Build GRCh38
rs573391527 Chr.14: 65044371-65044371 on Build GRCh38
rs553722789 Chr.14: 65044636-65044636 on Build GRCh38
rs372687079 Chr.14: 65044358-65044358 on Build GRCh38
rs375760309 Chr.14: 65044326-65044326 on Build GRCh38
rs117195243 Chr.14: 65044525-65044525 on Build GRCh38
rs138494513 Chr.14: 65044403-65044403 on Build GRCh38
rs772861260 Chr.14: 65044322-65044322 on Build GRCh38
rs758348521 Chr.14: 65044268-65044268 on Build GRCh38
rs779880861 Chr.14: 65044406-65044406 on Build GRCh38
rs773699138 Chr.14: 65044434-65044434 on Build GRCh38
rs746813710 Chr.14: 65044286-65044286 on Build GRCh38
rs777164875 Chr.14: 65044248-65044249 on Build GRCh38
rs45565742 Chr.14: 65044253-65044253 on Build GRCh38
rs747017414 Chr.14: 65044414-65044414 on Build GRCh38
rs562723220 Chr.14: 65044292-65044292 on Build GRCh38
rs748827539 Chr.14: 65044430-65044430 on Build GRCh38
rs975484730 Chr.14: 65044516-65044516 on Build GRCh38

View Full Product Details