Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 69569736-69570246 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC177
Entrez Gene ID
56936
Gene Name
coiled-coil domain containing 177
Gene Aliases
C14orf162, PLPL
UniGene
Hs.458319
Gene Chromosome Location
Chr.14: 69569814-69574883 on Build GRCh38
Transcripts
NM_001271507.1

SNP Detail

SNP ID Chromosome Location
rs772330115 Chr.14: 69569949-69569949 on Build GRCh38
rs548040589 Chr.14: 69570187-69570187 on Build GRCh38
rs905149889 Chr.14: 69569968-69569968 on Build GRCh38
rs547957922 Chr.14: 69570102-69570102 on Build GRCh38
rs544007703 Chr.14: 69569879-69569879 on Build GRCh38
rs145076280 Chr.14: 69569886-69569886 on Build GRCh38
rs993710021 Chr.14: 69569796-69569796 on Build GRCh38
rs796839012 Chr.14: 69569869-69569870 on Build GRCh38
rs552649742 Chr.14: 69569861-69569862 on Build GRCh38
rs988700666 Chr.14: 69570109-69570109 on Build GRCh38
rs938037187 Chr.14: 69570194-69570194 on Build GRCh38
rs964432647 Chr.14: 69569934-69569934 on Build GRCh38
rs531781533 Chr.14: 69569770-69569770 on Build GRCh38
rs114783194 Chr.14: 69570163-69570163 on Build GRCh38
rs1005852665 Chr.14: 69569865-69569865 on Build GRCh38
rs565038692 Chr.14: 69569828-69569828 on Build GRCh38
rs923071408 Chr.14: 69570044-69570044 on Build GRCh38
rs908379672 Chr.14: 69569856-69569856 on Build GRCh38
rs397965613 Chr.14: 69569871-69569872 on Build GRCh38
rs982581763 Chr.14: 69569827-69569827 on Build GRCh38
rs1000805579 Chr.14: 69570183-69570184 on Build GRCh38
rs185123841 Chr.14: 69570167-69570167 on Build GRCh38
rs952371261 Chr.14: 69569811-69569811 on Build GRCh38
rs58198153 Chr.14: 69569860-69569861 on Build GRCh38
rs745457539 Chr.14: 69569837-69569839 on Build GRCh38
rs776943914 Chr.14: 69570204-69570204 on Build GRCh38
rs760632895 Chr.14: 69570134-69570134 on Build GRCh38
rs113429861 Chr.14: 69569859-69569860 on Build GRCh38
rs61980689 Chr.14: 69569873-69569873 on Build GRCh38
rs1056461990 Chr.14: 69570209-69570209 on Build GRCh38
rs76981262 Chr.14: 69569871-69569871 on Build GRCh38
rs934091425 Chr.14: 69569826-69569826 on Build GRCh38
rs778214904 Chr.14: 69569967-69569967 on Build GRCh38
rs75966360 Chr.14: 69569871-69569871 on Build GRCh38
rs1026977704 Chr.14: 69569805-69569805 on Build GRCh38
rs1038284762 Chr.14: 69569945-69569946 on Build GRCh38
rs922660955 Chr.14: 69569812-69569815 on Build GRCh38
rs869260274 Chr.14: 69569868-69569870 on Build GRCh38
rs192739261 Chr.14: 69569988-69569988 on Build GRCh38
rs372017984 Chr.14: 69570191-69570191 on Build GRCh38
rs903358647 Chr.14: 69570178-69570178 on Build GRCh38
rs947331745 Chr.14: 69570147-69570147 on Build GRCh38

View Full Product Details