Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 69573299-69573802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC177
Entrez Gene ID
56936
Gene Name
coiled-coil domain containing 177
Gene Aliases
C14orf162, PLPL
UniGene
Hs.458319
Gene Chromosome Location
Chr.14: 69569814-69574883 on Build GRCh38
Transcripts
NM_001271507.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7271985 c.64G>A p.E22K Chr.14: 69573559-69573559 on Build GRCh38
7271986 c.64G>A p.E22K Chr.14: 69573559-69573559 on Build GRCh38
4150831 c.206C>G p.S69C Chr.14: 69573417-69573417 on Build GRCh38
4148213 c.60G>C p.G20G Chr.14: 69573563-69573563 on Build GRCh38
4148212 c.117T>C p.A39A Chr.14: 69573506-69573506 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs990081394 Chr.14: 69573505-69573505 on Build GRCh38
rs1032148814 Chr.14: 69573705-69573705 on Build GRCh38
rs572854793 Chr.14: 69573772-69573772 on Build GRCh38
rs951963358 Chr.14: 69573427-69573427 on Build GRCh38
rs971900890 Chr.14: 69573368-69573368 on Build GRCh38
rs762252992 Chr.14: 69573493-69573493 on Build GRCh38
rs138905072 Chr.14: 69573642-69573642 on Build GRCh38
rs1050026269 Chr.14: 69573598-69573598 on Build GRCh38
rs540809779 Chr.14: 69573708-69573708 on Build GRCh38
rs563526840 Chr.14: 69573524-69573524 on Build GRCh38
rs984615503 Chr.14: 69573656-69573656 on Build GRCh38
rs1018975914 Chr.14: 69573566-69573566 on Build GRCh38
rs6573892 Chr.14: 69573506-69573506 on Build GRCh38
rs554652060 Chr.14: 69573744-69573744 on Build GRCh38
rs966560736 Chr.14: 69573468-69573468 on Build GRCh38
rs113077435 Chr.14: 69573674-69573674 on Build GRCh38
rs940171453 Chr.14: 69573549-69573549 on Build GRCh38
rs889508352 Chr.14: 69573395-69573395 on Build GRCh38
rs966554675 Chr.14: 69573351-69573351 on Build GRCh38
rs770020903 Chr.14: 69573614-69573614 on Build GRCh38
rs1043998714 Chr.14: 69573544-69573544 on Build GRCh38
rs1028003853 Chr.14: 69573386-69573386 on Build GRCh38
rs550871201 Chr.14: 69573712-69573712 on Build GRCh38
rs928747498 Chr.14: 69573541-69573541 on Build GRCh38
rs117298196 Chr.14: 69573563-69573563 on Build GRCh38
rs576968208 Chr.14: 69573574-69573574 on Build GRCh38
rs558596005 Chr.14: 69573661-69573661 on Build GRCh38
rs951970978 Chr.14: 69573647-69573647 on Build GRCh38
rs1052574099 Chr.14: 69573483-69573494 on Build GRCh38
rs1032180394 Chr.14: 69573488-69573488 on Build GRCh38
rs915870243 Chr.14: 69573455-69573455 on Build GRCh38
rs768123327 Chr.14: 69573499-69573499 on Build GRCh38
rs1019020791 Chr.14: 69573347-69573347 on Build GRCh38
rs981048008 Chr.14: 69573721-69573721 on Build GRCh38
rs914365877 Chr.14: 69573755-69573755 on Build GRCh38
rs889342320 Chr.14: 69573774-69573774 on Build GRCh38
rs1019811144 Chr.14: 69573464-69573464 on Build GRCh38
rs544861271 Chr.14: 69573510-69573510 on Build GRCh38
rs539351154 Chr.14: 69573694-69573694 on Build GRCh38
rs766878645 Chr.14: 69573482-69573483 on Build GRCh38
rs183327144 Chr.14: 69573690-69573690 on Build GRCh38
rs569479375 Chr.14: 69573715-69573715 on Build GRCh38
rs528099391 Chr.14: 69573596-69573596 on Build GRCh38
rs190804679 Chr.14: 69573576-69573576 on Build GRCh38
rs776070246 Chr.14: 69573768-69573768 on Build GRCh38
rs530519555 Chr.14: 69573538-69573538 on Build GRCh38
rs1040795769 Chr.14: 69573666-69573666 on Build GRCh38
rs889394168 Chr.14: 69573560-69573560 on Build GRCh38
rs902263308 Chr.14: 69573675-69573675 on Build GRCh38
rs745909065 Chr.14: 69573585-69573585 on Build GRCh38
rs988956732 Chr.14: 69573753-69573753 on Build GRCh38
rs957956714 Chr.14: 69573494-69573494 on Build GRCh38
rs977934397 Chr.14: 69573477-69573477 on Build GRCh38
rs781109542 Chr.14: 69573582-69573582 on Build GRCh38
rs889443880 Chr.14: 69573601-69573601 on Build GRCh38
rs948754567 Chr.14: 69573511-69573511 on Build GRCh38
rs1007958701 Chr.14: 69573435-69573435 on Build GRCh38
rs773774456 Chr.14: 69573697-69573698 on Build GRCh38
rs919087998 Chr.14: 69573419-69573419 on Build GRCh38
rs1026433778 Chr.14: 69573605-69573605 on Build GRCh38
rs560787174 Chr.14: 69573725-69573725 on Build GRCh38
rs1024004158 Chr.14: 69573762-69573762 on Build GRCh38
rs753258186 Chr.14: 69573502-69573502 on Build GRCh38
rs934423339 Chr.14: 69573483-69573488 on Build GRCh38
rs928357986 Chr.14: 69573471-69573471 on Build GRCh38

View Full Product Details