Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 72893965-72894463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPF3
Entrez Gene ID
8110
Gene Name
double PHD fingers 3
Gene Aliases
BAF45C, CERD4
UniGene
Hs.162868
Gene Chromosome Location
Chr.14: 72609032-72894116 on Build GRCh38
Transcripts
NM_001280542.1 NM_001280543.1 NM_012074.4 XM_017021670.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6076122 c.24C>A p.P8P Chr.14: 72894065-72894065 on Build GRCh38
7198987 c.16C>A p.H6N Chr.14: 72894073-72894073 on Build GRCh38
7198986 c.16C>A p.H6N Chr.14: 72894073-72894073 on Build GRCh38
5991978 c.32+6_32+9delTAAG p.? Chr.14: 72894048-72894051 on Build GRCh38
6076121 c.24C>A p.P8P Chr.14: 72894065-72894065 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs77901786 Chr.14: 72894001-72894001 on Build GRCh38
rs760133909 Chr.14: 72894099-72894099 on Build GRCh38
rs937274578 Chr.14: 72894259-72894259 on Build GRCh38
rs568916042 Chr.14: 72894196-72894196 on Build GRCh38
rs1047019222 Chr.14: 72894198-72894198 on Build GRCh38
rs752543658 Chr.14: 72894125-72894125 on Build GRCh38
rs368069682 Chr.14: 72894353-72894353 on Build GRCh38
rs537700641 Chr.14: 72894205-72894205 on Build GRCh38
rs78898062 Chr.14: 72894134-72894134 on Build GRCh38
rs188783513 Chr.14: 72894359-72894359 on Build GRCh38
rs1002990802 Chr.14: 72894226-72894226 on Build GRCh38
rs967588144 Chr.14: 72894255-72894255 on Build GRCh38
rs1034832327 Chr.14: 72894228-72894228 on Build GRCh38
rs1009074357 Chr.14: 72894171-72894171 on Build GRCh38
rs977055995 Chr.14: 72894145-72894145 on Build GRCh38
rs184255907 Chr.14: 72894124-72894124 on Build GRCh38
rs769090716 Chr.14: 72894080-72894080 on Build GRCh38
rs775931215 Chr.14: 72894071-72894071 on Build GRCh38
rs2215902 Chr.14: 72894219-72894219 on Build GRCh38
rs933045653 Chr.14: 72894175-72894175 on Build GRCh38
rs1030037555 Chr.14: 72894282-72894282 on Build GRCh38
rs368403776 Chr.14: 72894067-72894067 on Build GRCh38
rs538775775 Chr.14: 72894181-72894181 on Build GRCh38
rs1011727057 Chr.14: 72894238-72894238 on Build GRCh38
rs1033302553 Chr.14: 72894357-72894357 on Build GRCh38
rs971855637 Chr.14: 72894236-72894236 on Build GRCh38
rs913350945 Chr.14: 72894221-72894221 on Build GRCh38
rs766420993 Chr.14: 72894105-72894106 on Build GRCh38
rs1038272634 Chr.14: 72894275-72894275 on Build GRCh38
rs375157776 Chr.14: 72894337-72894337 on Build GRCh38
rs971479124 Chr.14: 72894069-72894069 on Build GRCh38
rs2215901 Chr.14: 72894241-72894241 on Build GRCh38
rs751478121 Chr.14: 72894131-72894131 on Build GRCh38
rs937222154 Chr.14: 72894082-72894082 on Build GRCh38
rs894665744 Chr.14: 72894237-72894237 on Build GRCh38
rs986281908 Chr.14: 72894176-72894176 on Build GRCh38
rs567292164 Chr.14: 72894002-72894003 on Build GRCh38
rs3061027 Chr.14: 72894116-72894117 on Build GRCh38
rs386381750 Chr.14: 72894113-72894114 on Build GRCh38
rs1037607205 Chr.14: 72894187-72894187 on Build GRCh38
rs763980962 Chr.14: 72894128-72894128 on Build GRCh38
rs758394087 Chr.14: 72894030-72894030 on Build GRCh38
rs910276716 Chr.14: 72894304-72894304 on Build GRCh38
rs575559757 Chr.14: 72894418-72894418 on Build GRCh38
rs777751055 Chr.14: 72894037-72894037 on Build GRCh38
rs757331758 Chr.14: 72894058-72894058 on Build GRCh38
rs781304681 Chr.14: 72894064-72894064 on Build GRCh38
rs796368632 Chr.14: 72894327-72894327 on Build GRCh38
rs923021081 Chr.14: 72894152-72894152 on Build GRCh38
rs758168945 Chr.14: 72894127-72894127 on Build GRCh38
rs754789485 Chr.14: 72894007-72894007 on Build GRCh38
rs753831760 Chr.14: 72894114-72894115 on Build GRCh38
rs913290106 Chr.14: 72893994-72893994 on Build GRCh38
rs368821011 Chr.14: 72894138-72894138 on Build GRCh38
rs5809598 Chr.14: 72894111-72894112 on Build GRCh38
rs3814865 Chr.14: 72894389-72894389 on Build GRCh38
rs1038020539 Chr.14: 72894111-72894111 on Build GRCh38
rs776157358 Chr.14: 72894110-72894110 on Build GRCh38
rs180963706 Chr.14: 72894084-72894084 on Build GRCh38
rs370550264 Chr.14: 72893999-72893999 on Build GRCh38
rs201332544 Chr.14: 72894044-72894044 on Build GRCh38
rs765863595 Chr.14: 72894105-72894105 on Build GRCh38
rs781276050 Chr.14: 72894136-72894136 on Build GRCh38
rs763199046 Chr.14: 72894056-72894056 on Build GRCh38
rs3814866 Chr.14: 72894313-72894313 on Build GRCh38
rs898532137 Chr.14: 72893983-72893983 on Build GRCh38
rs545138367 Chr.14: 72894369-72894369 on Build GRCh38
rs954519347 Chr.14: 72894283-72894283 on Build GRCh38
rs1019491449 Chr.14: 72894174-72894174 on Build GRCh38
rs990428349 Chr.14: 72894133-72894133 on Build GRCh38
rs973184754 Chr.14: 72894332-72894332 on Build GRCh38
rs76743039 Chr.14: 72894003-72894003 on Build GRCh38
rs907193359 Chr.14: 72894209-72894209 on Build GRCh38
rs949954285 Chr.14: 72894003-72894003 on Build GRCh38
rs988808419 Chr.14: 72894206-72894206 on Build GRCh38
rs749667758 Chr.14: 72894072-72894072 on Build GRCh38
rs549010693 Chr.14: 72894022-72894022 on Build GRCh38
rs33956553 Chr.14: 72894112-72894113 on Build GRCh38
rs977087304 Chr.14: 72894280-72894280 on Build GRCh38
rs574141316 Chr.14: 72894293-72894293 on Build GRCh38
rs910420993 Chr.14: 72894179-72894179 on Build GRCh38
rs115339146 Chr.14: 72894192-72894192 on Build GRCh38
rs752631670 Chr.14: 72894025-72894025 on Build GRCh38

View Full Product Details