Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 72940062-72940555 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF4
Entrez Gene ID
26094
Gene Name
DDB1 and CUL4 associated factor 4
Gene Aliases
WDR21, WDR21A
UniGene
Hs.331491
Gene Chromosome Location
Chr.14: 72926332-72962098 on Build GRCh38
Transcripts
NM_001163508.1 NM_001163509.1 NM_015604.3 NM_181340.2 NM_181341.2 XM_017021201.1 XM_017021202.1 XM_017021203.1 XM_017021204.1 XM_017021205.1 XM_017021206.1 XM_017021207.1 XM_017021208.1 XM_017021209.1 XM_017021210.1 XM_017021211.1 XM_017021212.1 XM_017021213.1 XM_017021214.1 XM_017021215.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4543668 c.341G>A p.R114K Chr.14: 72940367-72940367 on Build GRCh38
5362350 c.267delC p.L91fs*1 Chr.14: 72940293-72940293 on Build GRCh38
6205656 c.289C>T p.R97W Chr.14: 72940315-72940315 on Build GRCh38
6640494 c.229C>T p.R77C Chr.14: 72940255-72940255 on Build GRCh38
4430654 c.230G>A p.R77H Chr.14: 72940256-72940256 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766892392 Chr.14: 72940334-72940334 on Build GRCh38
rs1014956758 Chr.14: 72940470-72940470 on Build GRCh38
rs938818308 Chr.14: 72940106-72940106 on Build GRCh38
rs149121448 Chr.14: 72940369-72940369 on Build GRCh38
rs184655877 Chr.14: 72940297-72940297 on Build GRCh38
rs1010599888 Chr.14: 72940314-72940314 on Build GRCh38
rs750237205 Chr.14: 72940429-72940429 on Build GRCh38
rs775796928 Chr.14: 72940178-72940178 on Build GRCh38
rs761953600 Chr.14: 72940402-72940402 on Build GRCh38
rs757090548 Chr.14: 72940359-72940359 on Build GRCh38
rs193096472 Chr.14: 72940217-72940217 on Build GRCh38
rs996543093 Chr.14: 72940138-72940138 on Build GRCh38
rs568085633 Chr.14: 72940193-72940193 on Build GRCh38
rs569209028 Chr.14: 72940496-72940496 on Build GRCh38
rs115868434 Chr.14: 72940147-72940147 on Build GRCh38
rs774567508 Chr.14: 72940400-72940400 on Build GRCh38
rs770806933 Chr.14: 72940295-72940295 on Build GRCh38
rs772330861 Chr.14: 72940407-72940407 on Build GRCh38
rs529303867 Chr.14: 72940209-72940209 on Build GRCh38
rs577287808 Chr.14: 72940531-72940546 on Build GRCh38
rs979477585 Chr.14: 72940317-72940317 on Build GRCh38
rs781212988 Chr.14: 72940296-72940296 on Build GRCh38
rs878888326 Chr.14: 72940103-72940103 on Build GRCh38
rs749413805 Chr.14: 72940370-72940370 on Build GRCh38
rs762573869 Chr.14: 72940414-72940414 on Build GRCh38
rs768111454 Chr.14: 72940255-72940255 on Build GRCh38
rs752304788 Chr.14: 72940335-72940335 on Build GRCh38
rs916106571 Chr.14: 72940119-72940119 on Build GRCh38
rs745672293 Chr.14: 72940362-72940362 on Build GRCh38
rs781533598 Chr.14: 72940398-72940398 on Build GRCh38
rs142034868 Chr.14: 72940424-72940424 on Build GRCh38
rs748756541 Chr.14: 72940402-72940403 on Build GRCh38
rs992405184 Chr.14: 72940264-72940264 on Build GRCh38
rs149878526 Chr.14: 72940324-72940324 on Build GRCh38
rs746450387 Chr.14: 72940236-72940236 on Build GRCh38
rs764482723 Chr.14: 72940288-72940288 on Build GRCh38
rs768918283 Chr.14: 72940302-72940302 on Build GRCh38
rs758181662 Chr.14: 72940265-72940265 on Build GRCh38
rs779881989 Chr.14: 72940204-72940205 on Build GRCh38
rs774528760 Chr.14: 72940308-72940308 on Build GRCh38
rs370016560 Chr.14: 72940256-72940256 on Build GRCh38
rs768326831 Chr.14: 72940361-72940361 on Build GRCh38
rs970070820 Chr.14: 72940525-72940525 on Build GRCh38
rs564364944 Chr.14: 72940471-72940471 on Build GRCh38
rs752242788 Chr.14: 72940260-72940260 on Build GRCh38
rs139409447 Chr.14: 72940301-72940301 on Build GRCh38
rs1008185786 Chr.14: 72940101-72940101 on Build GRCh38
rs768685218 Chr.14: 72940388-72940388 on Build GRCh38
rs565965915 Chr.14: 72940252-72940252 on Build GRCh38
rs1015072944 Chr.14: 72940225-72940225 on Build GRCh38
rs1023901552 Chr.14: 72940515-72940515 on Build GRCh38
rs573519299 Chr.14: 72940412-72940412 on Build GRCh38
rs1051460484 Chr.14: 72940119-72940141 on Build GRCh38
rs189217066 Chr.14: 72940315-72940315 on Build GRCh38
rs916785452 Chr.14: 72940293-72940293 on Build GRCh38
rs376438962 Chr.14: 72940181-72940181 on Build GRCh38
rs545969923 Chr.14: 72940454-72940454 on Build GRCh38
rs757950361 Chr.14: 72940344-72940344 on Build GRCh38
rs867497261 Chr.14: 72940263-72940263 on Build GRCh38
rs931372635 Chr.14: 72940130-72940130 on Build GRCh38
rs751983056 Chr.14: 72940197-72940197 on Build GRCh38
rs763642317 Chr.14: 72940416-72940416 on Build GRCh38
rs61988546 Chr.14: 72940413-72940413 on Build GRCh38
rs373770788 Chr.14: 72940345-72940345 on Build GRCh38
rs762199428 Chr.14: 72940309-72940309 on Build GRCh38
rs952199436 Chr.14: 72940152-72940152 on Build GRCh38
rs200682888 Chr.14: 72940420-72940420 on Build GRCh38
rs746838937 Chr.14: 72940281-72940281 on Build GRCh38
rs539247726 Chr.14: 72940259-72940259 on Build GRCh38
rs750942030 Chr.14: 72940316-72940316 on Build GRCh38
rs140507721 Chr.14: 72940251-72940251 on Build GRCh38
rs202091513 Chr.14: 72940240-72940240 on Build GRCh38
rs780275291 Chr.14: 72940510-72940510 on Build GRCh38
rs532381874 Chr.14: 72940157-72940157 on Build GRCh38
rs879167896 Chr.14: 72940137-72940137 on Build GRCh38
rs145062084 Chr.14: 72940346-72940346 on Build GRCh38
rs1039632374 Chr.14: 72940131-72940131 on Build GRCh38
rs773502810 Chr.14: 72940409-72940409 on Build GRCh38
rs931546716 Chr.14: 72940377-72940377 on Build GRCh38
rs777545165 Chr.14: 72940278-72940278 on Build GRCh38
rs769678074 Chr.14: 72940365-72940365 on Build GRCh38
rs754527096 Chr.14: 72940264-72940265 on Build GRCh38

View Full Product Details