Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73658673-73659119 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAL1
Entrez Gene ID
83544
Gene Name
dynein axonemal light chain 1
Gene Aliases
C14orf168, CILD16
UniGene
Hs.271270
Gene Chromosome Location
Chr.14: 73644875-73703728 on Build GRCh38
Transcripts
NM_001201366.1 NM_031427.3 XM_017021679.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5848498 c.43G>A p.E15K Chr.14: 73658847-73658847 on Build GRCh38
5920666 c.106C>T p.P36S Chr.14: 73658910-73658910 on Build GRCh38
433341 c.86A>C p.K29T Chr.14: 73658890-73658890 on Build GRCh38
3497831 c.48G>A p.E16E Chr.14: 73658852-73658852 on Build GRCh38
6216991 c.129C>G p.S43S Chr.14: 73658933-73658933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748272991 Chr.14: 73658981-73658981 on Build GRCh38
rs115264278 Chr.14: 73658736-73658736 on Build GRCh38
rs1032650543 Chr.14: 73658705-73658705 on Build GRCh38
rs561525805 Chr.14: 73658714-73658714 on Build GRCh38
rs927254566 Chr.14: 73658809-73658809 on Build GRCh38
rs780217508 Chr.14: 73658811-73658811 on Build GRCh38
rs914371151 Chr.14: 73658701-73658701 on Build GRCh38
rs373365623 Chr.14: 73658923-73658923 on Build GRCh38
rs550967876 Chr.14: 73658883-73658883 on Build GRCh38
rs1051225356 Chr.14: 73658916-73658916 on Build GRCh38
rs777992586 Chr.14: 73658989-73658989 on Build GRCh38
rs573177503 Chr.14: 73659071-73659071 on Build GRCh38
rs750381368 Chr.14: 73658942-73658942 on Build GRCh38
rs774964160 Chr.14: 73658885-73658885 on Build GRCh38
rs567434187 Chr.14: 73659004-73659004 on Build GRCh38
rs767561779 Chr.14: 73659046-73659046 on Build GRCh38
rs536071808 Chr.14: 73659023-73659023 on Build GRCh38
rs377571208 Chr.14: 73658966-73658966 on Build GRCh38
rs759537801 Chr.14: 73658979-73658979 on Build GRCh38
rs369887147 Chr.14: 73658840-73658840 on Build GRCh38
rs368774074 Chr.14: 73658980-73658980 on Build GRCh38
rs536740472 Chr.14: 73658955-73658955 on Build GRCh38
rs756001510 Chr.14: 73658946-73658946 on Build GRCh38
rs1038344349 Chr.14: 73658832-73658832 on Build GRCh38
rs17129144 Chr.14: 73658731-73658731 on Build GRCh38
rs146077001 Chr.14: 73658974-73658974 on Build GRCh38
rs756232574 Chr.14: 73658803-73658803 on Build GRCh38
rs888816583 Chr.14: 73658844-73658844 on Build GRCh38
rs772287450 Chr.14: 73658863-73658863 on Build GRCh38
rs202161399 Chr.14: 73658936-73658936 on Build GRCh38
rs1026238121 Chr.14: 73659024-73659024 on Build GRCh38
rs570969955 Chr.14: 73658913-73658913 on Build GRCh38
rs773793708 Chr.14: 73658873-73658873 on Build GRCh38
rs772487723 Chr.14: 73658984-73658984 on Build GRCh38
rs971534896 Chr.14: 73659091-73659091 on Build GRCh38
rs762760445 Chr.14: 73658727-73658727 on Build GRCh38
rs988874757 Chr.14: 73659032-73659032 on Build GRCh38
rs980329304 Chr.14: 73658790-73658790 on Build GRCh38
rs376441562 Chr.14: 73659070-73659070 on Build GRCh38
rs528051790 Chr.14: 73658816-73658816 on Build GRCh38
rs747605624 Chr.14: 73659002-73659002 on Build GRCh38
rs748496499 Chr.14: 73658843-73658843 on Build GRCh38
rs75788431 Chr.14: 73659094-73659094 on Build GRCh38
rs371347472 Chr.14: 73658715-73658715 on Build GRCh38
rs749588974 Chr.14: 73658812-73658812 on Build GRCh38
rs762639902 Chr.14: 73658906-73658906 on Build GRCh38
rs763705136 Chr.14: 73658910-73658910 on Build GRCh38
rs879441623 Chr.14: 73659010-73659010 on Build GRCh38

View Full Product Details