Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73698289-73698792 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAL1
Entrez Gene ID
83544
Gene Name
dynein axonemal light chain 1
Gene Aliases
C14orf168, CILD16
UniGene
Hs.271270
Gene Chromosome Location
Chr.14: 73644875-73703728 on Build GRCh38
Transcripts
NM_001201366.1 NM_031427.3

SNP Detail

SNP ID Chromosome Location
rs951154942 Chr.14: 73698529-73698529 on Build GRCh38
rs188175429 Chr.14: 73698678-73698678 on Build GRCh38
rs552415043 Chr.14: 73698515-73698515 on Build GRCh38
rs939580242 Chr.14: 73698372-73698372 on Build GRCh38
rs943845802 Chr.14: 73698708-73698708 on Build GRCh38
rs886050700 Chr.14: 73698382-73698382 on Build GRCh38
rs560647853 Chr.14: 73698337-73698337 on Build GRCh38
rs992338864 Chr.14: 73698568-73698568 on Build GRCh38
rs4903140 Chr.14: 73698398-73698398 on Build GRCh38
rs970048423 Chr.14: 73698591-73698591 on Build GRCh38
rs1009292709 Chr.14: 73698469-73698469 on Build GRCh38
rs755563143 Chr.14: 73698655-73698655 on Build GRCh38
rs1039254535 Chr.14: 73698371-73698371 on Build GRCh38
rs908075309 Chr.14: 73698356-73698356 on Build GRCh38
rs886050702 Chr.14: 73698497-73698497 on Build GRCh38
rs746527785 Chr.14: 73698506-73698507 on Build GRCh38
rs1015080593 Chr.14: 73698554-73698554 on Build GRCh38
rs550654899 Chr.14: 73698530-73698530 on Build GRCh38
rs944955694 Chr.14: 73698347-73698347 on Build GRCh38
rs886050703 Chr.14: 73698512-73698512 on Build GRCh38
rs984039953 Chr.14: 73698339-73698339 on Build GRCh38
rs916813924 Chr.14: 73698584-73698584 on Build GRCh38
rs530824546 Chr.14: 73698526-73698527 on Build GRCh38
rs989122795 Chr.14: 73698691-73698691 on Build GRCh38
rs974217601 Chr.14: 73698325-73698325 on Build GRCh38
rs951207222 Chr.14: 73698341-73698341 on Build GRCh38
rs960891820 Chr.14: 73698564-73698564 on Build GRCh38
rs199669674 Chr.14: 73698520-73698521 on Build GRCh38
rs891000034 Chr.14: 73698484-73698484 on Build GRCh38
rs567528655 Chr.14: 73698526-73698526 on Build GRCh38
rs974863443 Chr.14: 73698654-73698654 on Build GRCh38
rs758786439 Chr.14: 73698349-73698349 on Build GRCh38
rs550751048 Chr.14: 73698521-73698521 on Build GRCh38
rs376746869 Chr.14: 73698733-73698733 on Build GRCh38
rs994869302 Chr.14: 73698521-73698524 on Build GRCh38
rs886050701 Chr.14: 73698394-73698394 on Build GRCh38
rs143263241 Chr.14: 73698483-73698483 on Build GRCh38
rs770668475 Chr.14: 73698542-73698546 on Build GRCh38
rs1042170928 Chr.14: 73698745-73698745 on Build GRCh38
rs1044132538 Chr.14: 73698419-73698419 on Build GRCh38
rs537848577 Chr.14: 73698707-73698707 on Build GRCh38
rs1052496434 Chr.14: 73698695-73698695 on Build GRCh38
rs925313273 Chr.14: 73698667-73698667 on Build GRCh38
rs765689656 Chr.14: 73698527-73698530 on Build GRCh38
rs966275409 Chr.14: 73698545-73698545 on Build GRCh38
rs939224850 Chr.14: 73698765-73698765 on Build GRCh38
rs995862264 Chr.14: 73698755-73698755 on Build GRCh38
rs916569041 Chr.14: 73698394-73698394 on Build GRCh38
rs551997152 Chr.14: 73698468-73698468 on Build GRCh38
rs141575996 Chr.14: 73698736-73698736 on Build GRCh38
rs565209134 Chr.14: 73698486-73698486 on Build GRCh38
rs935423083 Chr.14: 73698478-73698478 on Build GRCh38
rs995143615 Chr.14: 73698513-73698513 on Build GRCh38
rs979424868 Chr.14: 73698603-73698603 on Build GRCh38
rs148365447 Chr.14: 73698495-73698495 on Build GRCh38
rs992432149 Chr.14: 73698380-73698380 on Build GRCh38
rs1018177289 Chr.14: 73698503-73698503 on Build GRCh38
rs899762216 Chr.14: 73698506-73698506 on Build GRCh38
rs886050699 Chr.14: 73698341-73698341 on Build GRCh38
rs752083265 Chr.14: 73698587-73698587 on Build GRCh38

View Full Product Details