Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73717561-73718059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMSAN1
Entrez Gene ID
91748
Gene Name
ELM2 and Myb/SANT domain containing 1
Gene Aliases
C14orf117, C14orf43, LSR68, MIDEAS, c14_5541
UniGene
Hs.594157
Gene Chromosome Location
Chr.14: 73715122-73790285 on Build GRCh38
Transcripts
NM_001043318.2 NM_194278.3 XM_005268204.1 XM_005268205.1 XM_005268206.1

SNP Detail

SNP ID Chromosome Location
rs386364184 Chr.14: 73717938-73717939 on Build GRCh38
rs1013897645 Chr.14: 73717619-73717619 on Build GRCh38
rs1017482094 Chr.14: 73717755-73717755 on Build GRCh38
rs79648433 Chr.14: 73717902-73717902 on Build GRCh38
rs769982381 Chr.14: 73717668-73717668 on Build GRCh38
rs573681804 Chr.14: 73717598-73717598 on Build GRCh38
rs971552669 Chr.14: 73717874-73717874 on Build GRCh38
rs760049592 Chr.14: 73717949-73717949 on Build GRCh38
rs917407261 Chr.14: 73717590-73717590 on Build GRCh38
rs559448460 Chr.14: 73717941-73717941 on Build GRCh38
rs11454926 Chr.14: 73717934-73717935 on Build GRCh38
rs976244975 Chr.14: 73717958-73717958 on Build GRCh38
rs926287061 Chr.14: 73717604-73717604 on Build GRCh38
rs78795976 Chr.14: 73717901-73717901 on Build GRCh38
rs925023274 Chr.14: 73717705-73717705 on Build GRCh38
rs749570587 Chr.14: 73717798-73717798 on Build GRCh38
rs1009263528 Chr.14: 73717714-73717714 on Build GRCh38
rs985010852 Chr.14: 73718007-73718007 on Build GRCh38
rs913350477 Chr.14: 73717811-73717811 on Build GRCh38
rs528213877 Chr.14: 73717691-73717691 on Build GRCh38
rs553729105 Chr.14: 73717874-73717874 on Build GRCh38
rs150690902 Chr.14: 73717837-73717837 on Build GRCh38
rs7157499 Chr.14: 73717836-73717836 on Build GRCh38
rs950392385 Chr.14: 73717594-73717594 on Build GRCh38
rs966596077 Chr.14: 73717943-73717943 on Build GRCh38
rs373771757 Chr.14: 73717937-73717938 on Build GRCh38
rs956693463 Chr.14: 73717761-73717761 on Build GRCh38
rs542693997 Chr.14: 73717923-73717923 on Build GRCh38
rs528195269 Chr.14: 73718000-73718000 on Build GRCh38
rs969031401 Chr.14: 73717635-73717635 on Build GRCh38
rs940716261 Chr.14: 73718035-73718035 on Build GRCh38
rs536273237 Chr.14: 73717697-73717697 on Build GRCh38
rs944908859 Chr.14: 73717825-73717825 on Build GRCh38
rs1005338386 Chr.14: 73717861-73717861 on Build GRCh38
rs528874464 Chr.14: 73717919-73717919 on Build GRCh38
rs944532435 Chr.14: 73718024-73718024 on Build GRCh38
rs1023571679 Chr.14: 73717630-73717630 on Build GRCh38
rs386381774 Chr.14: 73717935-73717936 on Build GRCh38
rs887786936 Chr.14: 73717845-73717845 on Build GRCh38
rs776119971 Chr.14: 73718010-73718010 on Build GRCh38
rs573426983 Chr.14: 73717864-73717864 on Build GRCh38
rs868146685 Chr.14: 73717657-73717657 on Build GRCh38
rs773273765 Chr.14: 73717694-73717694 on Build GRCh38
rs988910625 Chr.14: 73717851-73717851 on Build GRCh38
rs748164789 Chr.14: 73717589-73717589 on Build GRCh38
rs980761384 Chr.14: 73717884-73717884 on Build GRCh38
rs386778797 Chr.14: 73717834-73717836 on Build GRCh38
rs1048044688 Chr.14: 73717692-73717692 on Build GRCh38
rs937618824 Chr.14: 73717617-73717617 on Build GRCh38
rs1000523992 Chr.14: 73717907-73717907 on Build GRCh38
rs551523676 Chr.14: 73718012-73718012 on Build GRCh38
rs571365983 Chr.14: 73718029-73718029 on Build GRCh38
rs546681024 Chr.14: 73717727-73717727 on Build GRCh38
rs553045578 Chr.14: 73717699-73717699 on Build GRCh38
rs10138390 Chr.14: 73717827-73717827 on Build GRCh38
rs575865954 Chr.14: 73717885-73717885 on Build GRCh38
rs1013449821 Chr.14: 73717899-73717899 on Build GRCh38
rs371572885 Chr.14: 73717984-73717984 on Build GRCh38
rs569519268 Chr.14: 73717962-73717962 on Build GRCh38
rs1031974603 Chr.14: 73717909-73717909 on Build GRCh38
rs890292679 Chr.14: 73717709-73717709 on Build GRCh38
rs559445098 Chr.14: 73717712-73717712 on Build GRCh38
rs922214643 Chr.14: 73717834-73717834 on Build GRCh38

View Full Product Details