Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73718286-73718779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMSAN1
Entrez Gene ID
91748
Gene Name
ELM2 and Myb/SANT domain containing 1
Gene Aliases
C14orf117, C14orf43, LSR68, MIDEAS, c14_5541
UniGene
Hs.594157
Gene Chromosome Location
Chr.14: 73715122-73790285 on Build GRCh38
Transcripts
NM_001043318.2 NM_194278.3 XM_005268204.1 XM_005268205.1 XM_005268206.1

SNP Detail

SNP ID Chromosome Location
rs188141243 Chr.14: 73718485-73718485 on Build GRCh38
rs921665483 Chr.14: 73718558-73718558 on Build GRCh38
rs200363514 Chr.14: 73718559-73718559 on Build GRCh38
rs140206807 Chr.14: 73718321-73718321 on Build GRCh38
rs767655157 Chr.14: 73718596-73718603 on Build GRCh38
rs1053664225 Chr.14: 73718620-73718620 on Build GRCh38
rs945278537 Chr.14: 73718639-73718639 on Build GRCh38
rs1018645264 Chr.14: 73718519-73718519 on Build GRCh38
rs73297460 Chr.14: 73718644-73718644 on Build GRCh38
rs946025951 Chr.14: 73718579-73718579 on Build GRCh38
rs1016927140 Chr.14: 73718486-73718486 on Build GRCh38
rs931366023 Chr.14: 73718689-73718689 on Build GRCh38
rs772551379 Chr.14: 73718525-73718525 on Build GRCh38
rs539060896 Chr.14: 73718668-73718668 on Build GRCh38
rs900837392 Chr.14: 73718432-73718432 on Build GRCh38
rs1001654785 Chr.14: 73718412-73718412 on Build GRCh38
rs558763909 Chr.14: 73718691-73718691 on Build GRCh38
rs928660848 Chr.14: 73718614-73718614 on Build GRCh38
rs953535566 Chr.14: 73718480-73718480 on Build GRCh38
rs1049452208 Chr.14: 73718696-73718696 on Build GRCh38
rs974465016 Chr.14: 73718549-73718549 on Build GRCh38
rs926400390 Chr.14: 73718563-73718582 on Build GRCh38
rs76837839 Chr.14: 73718342-73718342 on Build GRCh38
rs888237261 Chr.14: 73718707-73718707 on Build GRCh38
rs1047265421 Chr.14: 73718696-73718697 on Build GRCh38
rs913800371 Chr.14: 73718628-73718628 on Build GRCh38
rs73297459 Chr.14: 73718417-73718417 on Build GRCh38
rs1046053265 Chr.14: 73718598-73718598 on Build GRCh38
rs1006339920 Chr.14: 73718732-73718732 on Build GRCh38
rs917883859 Chr.14: 73718518-73718518 on Build GRCh38
rs1025803672 Chr.14: 73718450-73718450 on Build GRCh38
rs569381925 Chr.14: 73718757-73718757 on Build GRCh38
rs887264955 Chr.14: 73718729-73718729 on Build GRCh38
rs1009644309 Chr.14: 73718410-73718410 on Build GRCh38
rs926721397 Chr.14: 73718587-73718587 on Build GRCh38
rs913121899 Chr.14: 73718360-73718360 on Build GRCh38
rs941772398 Chr.14: 73718565-73718584 on Build GRCh38
rs567183493 Chr.14: 73718602-73718602 on Build GRCh38
rs1028043545 Chr.14: 73718457-73718457 on Build GRCh38
rs372370578 Chr.14: 73718687-73718687 on Build GRCh38
rs530613798 Chr.14: 73718442-73718442 on Build GRCh38
rs980821927 Chr.14: 73718568-73718568 on Build GRCh38
rs117016658 Chr.14: 73718384-73718384 on Build GRCh38
rs566543159 Chr.14: 73718661-73718661 on Build GRCh38
rs183464538 Chr.14: 73718381-73718381 on Build GRCh38
rs972736539 Chr.14: 73718515-73718515 on Build GRCh38
rs545658956 Chr.14: 73718385-73718385 on Build GRCh38
rs913040208 Chr.14: 73718572-73718572 on Build GRCh38
rs996529716 Chr.14: 73718448-73718448 on Build GRCh38
rs577459665 Chr.14: 73718495-73718495 on Build GRCh38
rs79547755 Chr.14: 73718642-73718642 on Build GRCh38
rs905002059 Chr.14: 73718309-73718309 on Build GRCh38
rs575029259 Chr.14: 73718347-73718350 on Build GRCh38

View Full Product Details