Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73932173-73932630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM161B
Entrez Gene ID
145483
Gene Name
family with sequence similarity 161 member B
Gene Aliases
C14orf44, c14_5547
UniGene
Hs.550547
Gene Chromosome Location
Chr.14: 73932031-73950414 on Build GRCh38
Transcripts
XM_011536475.1 NM_001242924.1 NM_001242926.1 NM_001242927.1 NM_001242928.1 NM_021188.2 NR_040251.1

Gene Symbol
ZNF410
Entrez Gene ID
57862
Gene Name
zinc finger protein 410
Gene Aliases
APA-1, APA1
UniGene
Hs.270869
Gene Chromosome Location
Chr.14: 73886615-73932288 on Build GRCh38
Transcripts
XM_011536475.1 NM_001242924.1 NM_001242926.1 NM_001242927.1 NM_001242928.1 NM_021188.2 NR_040251.1

SNP Detail

SNP ID Chromosome Location
rs977028069 Chr.14: 73932380-73932380 on Build GRCh38
rs552268260 Chr.14: 73932528-73932528 on Build GRCh38
rs762023993 Chr.14: 73932403-73932403 on Build GRCh38
rs148201672 Chr.14: 73932415-73932415 on Build GRCh38
rs6574155 Chr.14: 73932509-73932509 on Build GRCh38
rs899921786 Chr.14: 73932355-73932355 on Build GRCh38
rs532083626 Chr.14: 73932522-73932522 on Build GRCh38
rs751422334 Chr.14: 73932510-73932510 on Build GRCh38
rs760891694 Chr.14: 73932335-73932335 on Build GRCh38
rs958460553 Chr.14: 73932447-73932447 on Build GRCh38
rs971269731 Chr.14: 73932425-73932425 on Build GRCh38
rs576743789 Chr.14: 73932298-73932298 on Build GRCh38
rs754362748 Chr.14: 73932513-73932514 on Build GRCh38
rs966856592 Chr.14: 73932377-73932377 on Build GRCh38
rs550518747 Chr.14: 73932203-73932206 on Build GRCh38
rs375523054 Chr.14: 73932454-73932455 on Build GRCh38
rs2336547 Chr.14: 73932588-73932588 on Build GRCh38
rs909845561 Chr.14: 73932423-73932423 on Build GRCh38
rs1032357794 Chr.14: 73932386-73932386 on Build GRCh38
rs991225918 Chr.14: 73932491-73932491 on Build GRCh38
rs112276440 Chr.14: 73932449-73932450 on Build GRCh38
rs752155118 Chr.14: 73932474-73932474 on Build GRCh38
rs1010229771 Chr.14: 73932407-73932407 on Build GRCh38
rs1032643040 Chr.14: 73932252-73932252 on Build GRCh38
rs1021551592 Chr.14: 73932363-73932363 on Build GRCh38
rs554513909 Chr.14: 73932353-73932353 on Build GRCh38
rs908465918 Chr.14: 73932513-73932513 on Build GRCh38
rs931208466 Chr.14: 73932206-73932207 on Build GRCh38
rs543742383 Chr.14: 73932500-73932500 on Build GRCh38
rs762643833 Chr.14: 73932501-73932501 on Build GRCh38
rs932710606 Chr.14: 73932603-73932603 on Build GRCh38
rs954062847 Chr.14: 73932401-73932401 on Build GRCh38
rs772515614 Chr.14: 73932388-73932388 on Build GRCh38
rs1046999810 Chr.14: 73932214-73932214 on Build GRCh38
rs764659423 Chr.14: 73932511-73932513 on Build GRCh38
rs764759933 Chr.14: 73932412-73932412 on Build GRCh38
rs757059909 Chr.14: 73932527-73932527 on Build GRCh38
rs943922837 Chr.14: 73932348-73932348 on Build GRCh38
rs982245955 Chr.14: 73932437-73932437 on Build GRCh38
rs892878515 Chr.14: 73932258-73932258 on Build GRCh38
rs919577084 Chr.14: 73932493-73932493 on Build GRCh38
rs562412611 Chr.14: 73932396-73932396 on Build GRCh38
rs1035376895 Chr.14: 73932446-73932447 on Build GRCh38
rs773785291 Chr.14: 73932395-73932395 on Build GRCh38
rs539803860 Chr.14: 73932508-73932508 on Build GRCh38
rs536012299 Chr.14: 73932279-73932279 on Build GRCh38
rs1040918422 Chr.14: 73932560-73932560 on Build GRCh38
rs972814636 Chr.14: 73932503-73932503 on Build GRCh38
rs758601185 Chr.14: 73932509-73932509 on Build GRCh38
rs905795484 Chr.14: 73932604-73932604 on Build GRCh38

View Full Product Details