Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73933060-73933605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM161B
Entrez Gene ID
145483
Gene Name
family with sequence similarity 161 member B
Gene Aliases
C14orf44, c14_5547
UniGene
Hs.550547
Gene Chromosome Location
Chr.14: 73932031-73950414 on Build GRCh38
Transcripts
NM_152445.2 XM_011536475.1

SNP Detail

SNP ID Chromosome Location
rs570122003 Chr.14: 73933516-73933516 on Build GRCh38
rs763623510 Chr.14: 73933407-73933407 on Build GRCh38
rs953819315 Chr.14: 73933156-73933158 on Build GRCh38
rs72627137 Chr.14: 73933474-73933474 on Build GRCh38
rs527556060 Chr.14: 73933401-73933401 on Build GRCh38
rs886636456 Chr.14: 73933381-73933381 on Build GRCh38
rs562289703 Chr.14: 73933149-73933149 on Build GRCh38
rs188680991 Chr.14: 73933336-73933336 on Build GRCh38
rs958549746 Chr.14: 73933287-73933287 on Build GRCh38
rs927982317 Chr.14: 73933240-73933240 on Build GRCh38
rs67602711 Chr.14: 73933472-73933472 on Build GRCh38
rs191256921 Chr.14: 73933411-73933411 on Build GRCh38
rs752077577 Chr.14: 73933355-73933355 on Build GRCh38
rs750895647 Chr.14: 73933405-73933405 on Build GRCh38
rs965007335 Chr.14: 73933436-73933437 on Build GRCh38
rs1038908468 Chr.14: 73933498-73933498 on Build GRCh38
rs963067108 Chr.14: 73933170-73933170 on Build GRCh38
rs945922108 Chr.14: 73933369-73933369 on Build GRCh38
rs755574888 Chr.14: 73933398-73933398 on Build GRCh38
rs1055755 Chr.14: 73933518-73933518 on Build GRCh38
rs535954291 Chr.14: 73933528-73933528 on Build GRCh38
rs921188851 Chr.14: 73933133-73933134 on Build GRCh38
rs895407785 Chr.14: 73933328-73933329 on Build GRCh38
rs555971308 Chr.14: 73933532-73933532 on Build GRCh38
rs547942670 Chr.14: 73933266-73933266 on Build GRCh38
rs927155676 Chr.14: 73933283-73933283 on Build GRCh38
rs1002934072 Chr.14: 73933578-73933578 on Build GRCh38
rs749302516 Chr.14: 73933560-73933562 on Build GRCh38
rs1015792224 Chr.14: 73933432-73933432 on Build GRCh38
rs914488135 Chr.14: 73933368-73933368 on Build GRCh38
rs971355780 Chr.14: 73933551-73933551 on Build GRCh38
rs990515248 Chr.14: 73933338-73933338 on Build GRCh38
rs34239517 Chr.14: 73933230-73933230 on Build GRCh38
rs1007582325 Chr.14: 73933114-73933114 on Build GRCh38
rs989346897 Chr.14: 73933158-73933158 on Build GRCh38
rs989830271 Chr.14: 73933480-73933480 on Build GRCh38
rs945465099 Chr.14: 73933187-73933187 on Build GRCh38
rs763297857 Chr.14: 73933162-73933162 on Build GRCh38
rs898748995 Chr.14: 73933513-73933513 on Build GRCh38
rs981462029 Chr.14: 73933275-73933275 on Build GRCh38
rs757418571 Chr.14: 73933220-73933221 on Build GRCh38
rs562182414 Chr.14: 73933243-73933243 on Build GRCh38
rs965066469 Chr.14: 73933102-73933102 on Build GRCh38
rs997853683 Chr.14: 73933457-73933457 on Build GRCh38
rs1041386447 Chr.14: 73933374-73933374 on Build GRCh38
rs1017183259 Chr.14: 73933127-73933127 on Build GRCh38
rs994187219 Chr.14: 73933236-73933236 on Build GRCh38
rs142566901 Chr.14: 73933193-73933193 on Build GRCh38
rs1025611708 Chr.14: 73933537-73933537 on Build GRCh38
rs1050160220 Chr.14: 73933412-73933412 on Build GRCh38

View Full Product Details