Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73971681-73972099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD5
Entrez Gene ID
957
Gene Name
ectonucleoside triphosphate diphosphohydrolase 5
Gene Aliases
CD39L4, NTPDase-5, PCPH
UniGene
Hs.655070
Gene Chromosome Location
Chr.14: 73959420-74019326 on Build GRCh38
Transcripts
NM_001249.3 NM_001321984.1 NM_001321985.1 NM_001321986.1 NM_001321987.1 NM_001321988.1 XM_005268224.3 XM_006720325.3 XM_006720326.2 XM_017021813.1 XM_017021814.1 XM_017021815.1 XM_017021816.1 XM_017021817.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7122354 c.1049delT p.L350fs*1 Chr.14: 73971887-73971887 on Build GRCh38
7122356 c.1044T>A p.G348G Chr.14: 73971892-73971892 on Build GRCh38
6654307 c.1043delG p.G348fs*3 Chr.14: 73971893-73971893 on Build GRCh38
957710 c.1043G>C p.G348A Chr.14: 73971893-73971893 on Build GRCh38
7153293 c.1075G>C p.A359P Chr.14: 73971861-73971861 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562377025 Chr.14: 73971789-73971789 on Build GRCh38
rs777883629 Chr.14: 73971960-73971960 on Build GRCh38
rs762839921 Chr.14: 73972052-73972052 on Build GRCh38
rs750042321 Chr.14: 73971917-73971917 on Build GRCh38
rs192479842 Chr.14: 73972014-73972014 on Build GRCh38
rs143609351 Chr.14: 73971896-73971896 on Build GRCh38
rs371841301 Chr.14: 73971776-73971776 on Build GRCh38
rs199705274 Chr.14: 73971856-73971856 on Build GRCh38
rs376329214 Chr.14: 73971831-73971831 on Build GRCh38
rs748430316 Chr.14: 73971809-73971809 on Build GRCh38
rs761340054 Chr.14: 73971898-73971898 on Build GRCh38
rs756911549 Chr.14: 73971832-73971834 on Build GRCh38
rs60865479 Chr.14: 73971787-73971796 on Build GRCh38
rs1053202389 Chr.14: 73972003-73972003 on Build GRCh38
rs201347007 Chr.14: 73971802-73971802 on Build GRCh38
rs774010812 Chr.14: 73971897-73971897 on Build GRCh38
rs878887405 Chr.14: 73971985-73971985 on Build GRCh38
rs371633599 Chr.14: 73971938-73971938 on Build GRCh38
rs1014481651 Chr.14: 73971816-73971816 on Build GRCh38
rs904375575 Chr.14: 73971966-73971969 on Build GRCh38
rs79964629 Chr.14: 73971795-73971795 on Build GRCh38
rs1042921272 Chr.14: 73971965-73971966 on Build GRCh38
rs777041966 Chr.14: 73971872-73971872 on Build GRCh38
rs767307160 Chr.14: 73971903-73971903 on Build GRCh38
rs1014304889 Chr.14: 73972018-73972018 on Build GRCh38
rs750406098 Chr.14: 73971930-73971930 on Build GRCh38
rs752782819 Chr.14: 73971945-73971945 on Build GRCh38
rs533134769 Chr.14: 73972019-73972023 on Build GRCh38
rs71873615 Chr.14: 73971775-73971784 on Build GRCh38
rs1032427512 Chr.14: 73971925-73971925 on Build GRCh38
rs918892165 Chr.14: 73971921-73971921 on Build GRCh38
rs943186600 Chr.14: 73971955-73971955 on Build GRCh38
rs766424877 Chr.14: 73972032-73972032 on Build GRCh38
rs778768938 Chr.14: 73971944-73971944 on Build GRCh38
rs778449357 Chr.14: 73971858-73971858 on Build GRCh38
rs758435056 Chr.14: 73971952-73971952 on Build GRCh38
rs367801118 Chr.14: 73971920-73971920 on Build GRCh38
rs761718013 Chr.14: 73971886-73971886 on Build GRCh38
rs975240093 Chr.14: 73972068-73972068 on Build GRCh38
rs772299358 Chr.14: 73971845-73971845 on Build GRCh38
rs745394385 Chr.14: 73971960-73971964 on Build GRCh38
rs375138051 Chr.14: 73971943-73971943 on Build GRCh38
rs771301072 Chr.14: 73971859-73971859 on Build GRCh38
rs200541968 Chr.14: 73971788-73971797 on Build GRCh38
rs542071533 Chr.14: 73971785-73971785 on Build GRCh38
rs768492795 Chr.14: 73971784-73971784 on Build GRCh38
rs939703217 Chr.14: 73971833-73971833 on Build GRCh38
rs138341765 Chr.14: 73971800-73971800 on Build GRCh38
rs960041651 Chr.14: 73971811-73971811 on Build GRCh38
rs998922242 Chr.14: 73971990-73971990 on Build GRCh38

View Full Product Details