Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 74285965-74286513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCD4
Entrez Gene ID
5826
Gene Name
ATP binding cassette subfamily D member 4
Gene Aliases
ABC41, EST352188, MAHCJ, P70R, P79R, PMP69, PXMP1L
UniGene
Hs.94395
Gene Chromosome Location
Chr.14: 74285277-74303064 on Build GRCh38
Transcripts
NM_005050.3 NR_003256.2 XM_005267939.3 XM_005267940.3 XM_005267941.4 XM_005267942.4 XM_005267946.3 XM_005267949.3 XM_006720223.2 XM_011537041.2 XM_017021531.1 XM_017021532.1 XM_017021533.1 XM_017021534.1 XM_017021535.1 XM_017021536.1 XM_017021537.1 XM_017021538.1 XM_017021539.1 XM_017021540.1 XM_017021541.1 XM_017021542.1 XM_017021543.1 XM_017021544.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3983691 c.1804A>G p.R602G Chr.14: 74286478-74286478 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs987473967 Chr.14: 74286177-74286177 on Build GRCh38
rs907218862 Chr.14: 74286276-74286276 on Build GRCh38
rs4148081 Chr.14: 74286281-74286281 on Build GRCh38
rs529683757 Chr.14: 74286089-74286090 on Build GRCh38
rs777411039 Chr.14: 74286418-74286418 on Build GRCh38
rs746941180 Chr.14: 74286367-74286367 on Build GRCh38
rs994211890 Chr.14: 74285992-74285992 on Build GRCh38
rs575004843 Chr.14: 74285991-74285991 on Build GRCh38
rs776547619 Chr.14: 74286379-74286379 on Build GRCh38
rs1054509963 Chr.14: 74286069-74286069 on Build GRCh38
rs764083262 Chr.14: 74286393-74286393 on Build GRCh38
rs1024068495 Chr.14: 74286111-74286111 on Build GRCh38
rs768922040 Chr.14: 74286452-74286452 on Build GRCh38
rs568073928 Chr.14: 74286320-74286320 on Build GRCh38
rs1052022 Chr.14: 74286341-74286341 on Build GRCh38
rs368005846 Chr.14: 74286459-74286459 on Build GRCh38
rs749465661 Chr.14: 74286444-74286444 on Build GRCh38
rs958797889 Chr.14: 74286106-74286106 on Build GRCh38
rs967098451 Chr.14: 74286141-74286141 on Build GRCh38
rs777977935 Chr.14: 74286426-74286426 on Build GRCh38
rs773609274 Chr.14: 74286488-74286488 on Build GRCh38
rs747648965 Chr.14: 74286345-74286345 on Build GRCh38
rs745691939 Chr.14: 74286385-74286385 on Build GRCh38
rs1052008 Chr.14: 74286409-74286409 on Build GRCh38
rs758139648 Chr.14: 74286417-74286417 on Build GRCh38
rs975349818 Chr.14: 74286422-74286422 on Build GRCh38
rs774037012 Chr.14: 74286075-74286075 on Build GRCh38
rs1034245996 Chr.14: 74286094-74286094 on Build GRCh38
rs752301205 Chr.14: 74286412-74286412 on Build GRCh38
rs1054391763 Chr.14: 74286394-74286396 on Build GRCh38
rs930034094 Chr.14: 74286270-74286270 on Build GRCh38
rs757231099 Chr.14: 74286421-74286421 on Build GRCh38
rs1052012 Chr.14: 74286404-74286404 on Build GRCh38
rs1038535345 Chr.14: 74286230-74286230 on Build GRCh38
rs922923526 Chr.14: 74286158-74286158 on Build GRCh38
rs775713556 Chr.14: 74286443-74286443 on Build GRCh38
rs986806104 Chr.14: 74286294-74286294 on Build GRCh38
rs911694453 Chr.14: 74286007-74286007 on Build GRCh38
rs761437496 Chr.14: 74286089-74286089 on Build GRCh38
rs111792207 Chr.14: 74286402-74286403 on Build GRCh38
rs8618 Chr.14: 74286273-74286273 on Build GRCh38
rs2466 Chr.14: 74286199-74286199 on Build GRCh38
rs756462000 Chr.14: 74286407-74286407 on Build GRCh38
rs762291576 Chr.14: 74286403-74286403 on Build GRCh38
rs771052053 Chr.14: 74286378-74286378 on Build GRCh38
rs745627342 Chr.14: 74286431-74286431 on Build GRCh38
rs898617769 Chr.14: 74286232-74286232 on Build GRCh38
rs560403421 Chr.14: 74286192-74286192 on Build GRCh38
rs368652092 Chr.14: 74286425-74286425 on Build GRCh38
rs766334278 Chr.14: 74286353-74286353 on Build GRCh38
rs552887859 Chr.14: 74286386-74286386 on Build GRCh38
rs763031301 Chr.14: 74286388-74286388 on Build GRCh38
rs774479589 Chr.14: 74286397-74286397 on Build GRCh38
rs772523388 Chr.14: 74286468-74286468 on Build GRCh38
rs761917106 Chr.14: 74286464-74286464 on Build GRCh38
rs147254822 Chr.14: 74286025-74286025 on Build GRCh38
rs932965705 Chr.14: 74286176-74286176 on Build GRCh38
rs1052006 Chr.14: 74286438-74286438 on Build GRCh38
rs1038880773 Chr.14: 74286029-74286029 on Build GRCh38
rs141001008 Chr.14: 74286302-74286302 on Build GRCh38
rs989964856 Chr.14: 74286109-74286109 on Build GRCh38
rs191056843 Chr.14: 74286149-74286149 on Build GRCh38
rs371201802 Chr.14: 74286437-74286437 on Build GRCh38
rs773085830 Chr.14: 74286133-74286133 on Build GRCh38
rs761534929 Chr.14: 74286028-74286028 on Build GRCh38
rs775597361 Chr.14: 74286387-74286387 on Build GRCh38
rs755048805 Chr.14: 74286353-74286353 on Build GRCh38
rs935959840 Chr.14: 74286063-74286063 on Build GRCh38
rs746568976 Chr.14: 74286420-74286420 on Build GRCh38
rs900418412 Chr.14: 74286032-74286032 on Build GRCh38
rs764904616 Chr.14: 74286052-74286052 on Build GRCh38
rs552538428 Chr.14: 74286207-74286207 on Build GRCh38
rs111396442 Chr.14: 74286408-74286409 on Build GRCh38
rs911504189 Chr.14: 74286427-74286427 on Build GRCh38

View Full Product Details