Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 74714682-74715082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCF1
Entrez Gene ID
51077
Gene Name
FCF1 rRNA-processing protein
Gene Aliases
Bka, C14orf111, CGI-35, UTP24
UniGene
Hs.579828
Gene Chromosome Location
Chr.14: 74713118-74738620 on Build GRCh38
Transcripts
NM_001318508.1 NM_015962.4 XM_005267730.3 XM_011536815.2 XM_011536816.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6290768 c.100A>G p.K34E Chr.14: 74714900-74714900 on Build GRCh38
699414 c.112A>G p.K38E Chr.14: 74714912-74714912 on Build GRCh38
957803 c.139G>A p.E47K Chr.14: 74714939-74714939 on Build GRCh38
5027888 c.124G>A p.A42T Chr.14: 74714924-74714924 on Build GRCh38
4448908 c.143+2T>A p.? Chr.14: 74714945-74714945 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778267085 Chr.14: 74714918-74714918 on Build GRCh38
rs934216507 Chr.14: 74714741-74714741 on Build GRCh38
rs534499486 Chr.14: 74714982-74714982 on Build GRCh38
rs982001406 Chr.14: 74714953-74714953 on Build GRCh38
rs959601126 Chr.14: 74714972-74714972 on Build GRCh38
rs1051397714 Chr.14: 74714746-74714746 on Build GRCh38
rs371163318 Chr.14: 74714959-74714959 on Build GRCh38
rs199980632 Chr.14: 74714923-74714923 on Build GRCh38
rs920923400 Chr.14: 74715052-74715052 on Build GRCh38
rs893970465 Chr.14: 74714737-74714737 on Build GRCh38
rs397718730 Chr.14: 74714793-74714793 on Build GRCh38
rs1033681658 Chr.14: 74714877-74714877 on Build GRCh38
rs548157503 Chr.14: 74715011-74715011 on Build GRCh38
rs367728211 Chr.14: 74714954-74714954 on Build GRCh38
rs114089356 Chr.14: 74714716-74714716 on Build GRCh38
rs752861745 Chr.14: 74714833-74714833 on Build GRCh38
rs878994733 Chr.14: 74714778-74714779 on Build GRCh38
rs771901257 Chr.14: 74714939-74714939 on Build GRCh38
rs189323401 Chr.14: 74714820-74714820 on Build GRCh38
rs1015321980 Chr.14: 74715014-74715014 on Build GRCh38
rs757618989 Chr.14: 74714858-74714858 on Build GRCh38
rs1012837082 Chr.14: 74714932-74714932 on Build GRCh38
rs754668964 Chr.14: 74714907-74714907 on Build GRCh38
rs779619603 Chr.14: 74714876-74714876 on Build GRCh38
rs35842570 Chr.14: 74714897-74714897 on Build GRCh38
rs35901005 Chr.14: 74714779-74714779 on Build GRCh38
rs921479871 Chr.14: 74714715-74714715 on Build GRCh38
rs769009018 Chr.14: 74714753-74714753 on Build GRCh38
rs147067928 Chr.14: 74714924-74714924 on Build GRCh38
rs770872126 Chr.14: 74714955-74714955 on Build GRCh38
rs200030575 Chr.14: 74714853-74714853 on Build GRCh38
rs1042668748 Chr.14: 74714712-74714712 on Build GRCh38
rs185242089 Chr.14: 74714724-74714724 on Build GRCh38
rs775417448 Chr.14: 74714986-74714986 on Build GRCh38
rs976356327 Chr.14: 74715052-74715053 on Build GRCh38
rs10438165 Chr.14: 74715051-74715051 on Build GRCh38
rs1007319856 Chr.14: 74714798-74714798 on Build GRCh38
rs888745309 Chr.14: 74714756-74714756 on Build GRCh38
rs758807415 Chr.14: 74714836-74714836 on Build GRCh38
rs201603970 Chr.14: 74714777-74714777 on Build GRCh38
rs377716020 Chr.14: 74714881-74714881 on Build GRCh38

View Full Product Details