Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 75605384-75605894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLVCR2
Entrez Gene ID
55640
Gene Name
feline leukemia virus subgroup C cellular receptor family member 2
Gene Aliases
C14orf58, CCT, EPV, FLVCRL14q, MFSD7C, PVHH
UniGene
Hs.509966
Gene Chromosome Location
Chr.14: 75578597-75648169 on Build GRCh38
Transcripts
NM_001195283.1 NM_017791.2

SNP Detail

SNP ID Chromosome Location
rs192251413 Chr.14: 75605721-75605721 on Build GRCh38
rs894618049 Chr.14: 75605476-75605476 on Build GRCh38
rs971683809 Chr.14: 75605645-75605645 on Build GRCh38
rs370240449 Chr.14: 75605801-75605801 on Build GRCh38
rs774436302 Chr.14: 75605498-75605498 on Build GRCh38
rs1006284836 Chr.14: 75605530-75605530 on Build GRCh38
rs150356352 Chr.14: 75605832-75605832 on Build GRCh38
rs750038384 Chr.14: 75605670-75605670 on Build GRCh38
rs755611577 Chr.14: 75605618-75605618 on Build GRCh38
rs555934170 Chr.14: 75605812-75605812 on Build GRCh38
rs542102379 Chr.14: 75605661-75605661 on Build GRCh38
rs997871518 Chr.14: 75605786-75605786 on Build GRCh38
rs758795651 Chr.14: 75605554-75605554 on Build GRCh38
rs545404296 Chr.14: 75605648-75605648 on Build GRCh38
rs1040903189 Chr.14: 75605734-75605734 on Build GRCh38
rs965046555 Chr.14: 75605577-75605577 on Build GRCh38
rs938695911 Chr.14: 75605675-75605675 on Build GRCh38
rs924579675 Chr.14: 75605669-75605669 on Build GRCh38
rs1004404918 Chr.14: 75605556-75605556 on Build GRCh38
rs979802705 Chr.14: 75605663-75605663 on Build GRCh38
rs973097074 Chr.14: 75605606-75605606 on Build GRCh38
rs966951811 Chr.14: 75605835-75605835 on Build GRCh38
rs184860900 Chr.14: 75605738-75605738 on Build GRCh38
rs752957345 Chr.14: 75605523-75605523 on Build GRCh38
rs888652572 Chr.14: 75605718-75605718 on Build GRCh38
rs996801775 Chr.14: 75605800-75605800 on Build GRCh38
rs921197014 Chr.14: 75605608-75605608 on Build GRCh38
rs996194253 Chr.14: 75605407-75605407 on Build GRCh38
rs1003038363 Chr.14: 75605459-75605459 on Build GRCh38
rs989312347 Chr.14: 75605619-75605619 on Build GRCh38
rs781317401 Chr.14: 75605850-75605850 on Build GRCh38
rs965015492 Chr.14: 75605744-75605744 on Build GRCh38
rs576821425 Chr.14: 75605572-75605572 on Build GRCh38
rs751174563 Chr.14: 75605822-75605822 on Build GRCh38
rs189694618 Chr.14: 75605837-75605837 on Build GRCh38
rs4903337 Chr.14: 75605685-75605685 on Build GRCh38
rs538021444 Chr.14: 75605442-75605442 on Build GRCh38
rs924715918 Chr.14: 75605870-75605870 on Build GRCh38
rs545922571 Chr.14: 75605574-75605574 on Build GRCh38
rs374470470 Chr.14: 75605674-75605674 on Build GRCh38
rs1048547640 Chr.14: 75605708-75605708 on Build GRCh38
rs764392314 Chr.14: 75605648-75605648 on Build GRCh38
rs757991896 Chr.14: 75605677-75605677 on Build GRCh38
rs531958124 Chr.14: 75605613-75605613 on Build GRCh38
rs779500563 Chr.14: 75605640-75605640 on Build GRCh38
rs533260490 Chr.14: 75605802-75605802 on Build GRCh38
rs913431995 Chr.14: 75605705-75605705 on Build GRCh38
rs1057322652 Chr.14: 75605460-75605460 on Build GRCh38
rs748126517 Chr.14: 75605857-75605857 on Build GRCh38
rs952481324 Chr.14: 75605528-75605528 on Build GRCh38
rs1024525924 Chr.14: 75605865-75605865 on Build GRCh38
rs562934129 Chr.14: 75605612-75605612 on Build GRCh38
rs187221308 Chr.14: 75605512-75605512 on Build GRCh38
rs562010416 Chr.14: 75605678-75605678 on Build GRCh38
rs867275146 Chr.14: 75605424-75605424 on Build GRCh38

View Full Product Details