Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 75654807-75655308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf1
Entrez Gene ID
11161
Gene Name
chromosome 14 open reading frame 1
Gene Aliases
ERG28, NET51
UniGene
Hs.15106
Gene Chromosome Location
Chr.14: 75650890-75661195 on Build GRCh38
Transcripts
NM_007176.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
274206 c.190C>T p.R64C Chr.14: 75654920-75654920 on Build GRCh38
7140363 c.190C>A p.R64S Chr.14: 75654920-75654920 on Build GRCh38
6614277 c.170C>T p.T57M Chr.14: 75654940-75654940 on Build GRCh38
6076350 c.165C>T p.I55I Chr.14: 75654945-75654945 on Build GRCh38
5848562 c.134-1G>A p.? Chr.14: 75654977-75654977 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs931375581 Chr.14: 75655084-75655084 on Build GRCh38
rs959464074 Chr.14: 75654959-75654959 on Build GRCh38
rs1030739599 Chr.14: 75655219-75655219 on Build GRCh38
rs577298772 Chr.14: 75655039-75655039 on Build GRCh38
rs765206618 Chr.14: 75654989-75654990 on Build GRCh38
rs1056755001 Chr.14: 75655098-75655098 on Build GRCh38
rs912828852 Chr.14: 75655048-75655048 on Build GRCh38
rs530463866 Chr.14: 75655023-75655023 on Build GRCh38
rs1000552431 Chr.14: 75655212-75655212 on Build GRCh38
rs1047215093 Chr.14: 75655085-75655085 on Build GRCh38
rs995339305 Chr.14: 75655269-75655269 on Build GRCh38
rs750389148 Chr.14: 75655010-75655010 on Build GRCh38
rs746304122 Chr.14: 75654958-75654958 on Build GRCh38
rs533384833 Chr.14: 75655026-75655026 on Build GRCh38
rs757383277 Chr.14: 75654940-75654940 on Build GRCh38
rs139223174 Chr.14: 75655278-75655278 on Build GRCh38
rs903735611 Chr.14: 75655175-75655175 on Build GRCh38
rs765016161 Chr.14: 75654918-75654918 on Build GRCh38
rs1001297766 Chr.14: 75655200-75655200 on Build GRCh38
rs753886025 Chr.14: 75654900-75654900 on Build GRCh38
rs1023461486 Chr.14: 75655147-75655147 on Build GRCh38
rs912044230 Chr.14: 75655045-75655045 on Build GRCh38
rs976102227 Chr.14: 75655279-75655279 on Build GRCh38
rs1033754762 Chr.14: 75654943-75654943 on Build GRCh38
rs1028073893 Chr.14: 75655015-75655015 on Build GRCh38
rs770294325 Chr.14: 75654979-75654979 on Build GRCh38
rs987356084 Chr.14: 75655111-75655111 on Build GRCh38
rs907032700 Chr.14: 75655167-75655167 on Build GRCh38
rs767551614 Chr.14: 75655092-75655092 on Build GRCh38
rs766149602 Chr.14: 75654930-75654930 on Build GRCh38
rs986449516 Chr.14: 75655225-75655225 on Build GRCh38
rs146110609 Chr.14: 75655232-75655232 on Build GRCh38
rs975612203 Chr.14: 75655065-75655065 on Build GRCh38
rs759489109 Chr.14: 75655021-75655021 on Build GRCh38
rs766054988 Chr.14: 75655002-75655002 on Build GRCh38
rs749645990 Chr.14: 75654990-75654990 on Build GRCh38
rs777890454 Chr.14: 75654927-75654927 on Build GRCh38
rs987002528 Chr.14: 75655030-75655030 on Build GRCh38
rs1045648217 Chr.14: 75655157-75655157 on Build GRCh38
rs950987645 Chr.14: 75655018-75655018 on Build GRCh38
rs921656516 Chr.14: 75655071-75655071 on Build GRCh38
rs752452661 Chr.14: 75654919-75654919 on Build GRCh38
rs549970659 Chr.14: 75655061-75655061 on Build GRCh38
rs984146550 Chr.14: 75655043-75655043 on Build GRCh38
rs754817064 Chr.14: 75654904-75654904 on Build GRCh38
rs188936011 Chr.14: 75654981-75654981 on Build GRCh38
rs1020248203 Chr.14: 75655266-75655266 on Build GRCh38
rs370578358 Chr.14: 75654886-75654886 on Build GRCh38
rs974920289 Chr.14: 75655055-75655055 on Build GRCh38
rs772737266 Chr.14: 75654869-75654869 on Build GRCh38
rs115386117 Chr.14: 75654939-75654939 on Build GRCh38
rs896296788 Chr.14: 75654834-75654834 on Build GRCh38
rs372165750 Chr.14: 75654999-75654999 on Build GRCh38
rs1055573835 Chr.14: 75655220-75655220 on Build GRCh38
rs979186771 Chr.14: 75655003-75655003 on Build GRCh38
rs542477179 Chr.14: 75654835-75654835 on Build GRCh38
rs1012460330 Chr.14: 75655120-75655120 on Build GRCh38
rs781660674 Chr.14: 75654944-75654944 on Build GRCh38
rs956347264 Chr.14: 75655222-75655222 on Build GRCh38
rs760250098 Chr.14: 75654997-75654997 on Build GRCh38
rs942840432 Chr.14: 75655052-75655052 on Build GRCh38
rs181088204 Chr.14: 75655011-75655011 on Build GRCh38
rs1027652029 Chr.14: 75655276-75655276 on Build GRCh38
rs774804960 Chr.14: 75654889-75654891 on Build GRCh38
rs758600734 Chr.14: 75654923-75654923 on Build GRCh38
rs112516206 Chr.14: 75655104-75655104 on Build GRCh38
rs919969596 Chr.14: 75655067-75655067 on Build GRCh38
rs767097516 Chr.14: 75654847-75654847 on Build GRCh38
rs760299308 Chr.14: 75654872-75654872 on Build GRCh38
rs1023781709 Chr.14: 75654949-75654949 on Build GRCh38
rs374591682 Chr.14: 75654933-75654933 on Build GRCh38
rs201839359 Chr.14: 75654995-75654995 on Build GRCh38
rs184120611 Chr.14: 75655090-75655090 on Build GRCh38

View Full Product Details