Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 77434120-77434575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VIPAS39
Entrez Gene ID
63894
Gene Name
VPS33B interacting protein, apical-basolateral polarity regulator, spe-39 homolog
Gene Aliases
C14orf133, SPE-39, SPE39, VIPAR, VPS16B, hSPE-39
UniGene
Hs.16157
Gene Chromosome Location
Chr.14: 77426675-77457640 on Build GRCh38
Transcripts
NM_001193314.1 NM_001193315.1 NM_001193316.1 NM_001193317.1 NM_022067.3 XM_011537066.1 XM_017021579.1 XM_017021580.1 XM_017021581.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2139414 c.1076A>G p.K359R Chr.14: 77434275-77434275 on Build GRCh38
380880 c.1077G>C p.K359N Chr.14: 77434274-77434274 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757708480 Chr.14: 77434347-77434347 on Build GRCh38
rs758970093 Chr.14: 77434230-77434230 on Build GRCh38
rs367593503 Chr.14: 77434307-77434307 on Build GRCh38
rs556575006 Chr.14: 77434498-77434498 on Build GRCh38
rs1006925256 Chr.14: 77434473-77434473 on Build GRCh38
rs143951008 Chr.14: 77434285-77434285 on Build GRCh38
rs776875927 Chr.14: 77434291-77434291 on Build GRCh38
rs753248757 Chr.14: 77434223-77434223 on Build GRCh38
rs906069549 Chr.14: 77434363-77434363 on Build GRCh38
rs1048179424 Chr.14: 77434449-77434449 on Build GRCh38
rs138544284 Chr.14: 77434280-77434280 on Build GRCh38
rs193250575 Chr.14: 77434423-77434423 on Build GRCh38
rs889900577 Chr.14: 77434451-77434451 on Build GRCh38
rs377726527 Chr.14: 77434253-77434253 on Build GRCh38
rs749639513 Chr.14: 77434328-77434337 on Build GRCh38
rs758345667 Chr.14: 77434333-77434333 on Build GRCh38
rs770887106 Chr.14: 77434188-77434188 on Build GRCh38
rs774318839 Chr.14: 77434508-77434508 on Build GRCh38
rs550788204 Chr.14: 77434185-77434185 on Build GRCh38
rs979804206 Chr.14: 77434380-77434380 on Build GRCh38
rs374264167 Chr.14: 77434220-77434220 on Build GRCh38
rs187919311 Chr.14: 77434399-77434399 on Build GRCh38
rs772051008 Chr.14: 77434274-77434274 on Build GRCh38
rs371371767 Chr.14: 77434254-77434254 on Build GRCh38
rs759729606 Chr.14: 77434298-77434298 on Build GRCh38
rs118155418 Chr.14: 77434286-77434286 on Build GRCh38
rs768122521 Chr.14: 77434205-77434205 on Build GRCh38
rs566472910 Chr.14: 77434255-77434255 on Build GRCh38
rs113480488 Chr.14: 77434351-77434351 on Build GRCh38
rs867677264 Chr.14: 77434477-77434477 on Build GRCh38
rs752809458 Chr.14: 77434330-77434330 on Build GRCh38
rs530909420 Chr.14: 77434179-77434179 on Build GRCh38
rs536974743 Chr.14: 77434209-77434209 on Build GRCh38
rs773976661 Chr.14: 77434269-77434269 on Build GRCh38
rs201857750 Chr.14: 77434270-77434270 on Build GRCh38
rs567504201 Chr.14: 77434193-77434193 on Build GRCh38
rs537925244 Chr.14: 77434401-77434401 on Build GRCh38
rs554821410 Chr.14: 77434408-77434408 on Build GRCh38
rs182152260 Chr.14: 77434336-77434336 on Build GRCh38
rs974685752 Chr.14: 77434187-77434187 on Build GRCh38
rs67370165 Chr.14: 77434147-77434147 on Build GRCh38
rs931140180 Chr.14: 77434432-77434432 on Build GRCh38
rs768321436 Chr.14: 77434257-77434257 on Build GRCh38
rs141560550 Chr.14: 77434411-77434411 on Build GRCh38
rs778107170 Chr.14: 77434238-77434238 on Build GRCh38
rs950387752 Chr.14: 77434322-77434322 on Build GRCh38
rs538314467 Chr.14: 77434338-77434338 on Build GRCh38

View Full Product Details