Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 80761287-80761834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP128
Entrez Gene ID
145508
Gene Name
centrosomal protein 128
Gene Aliases
C14orf145, C14orf61, LEDP/132
UniGene
Hs.162889
Gene Chromosome Location
Chr.14: 80478321-80959501 on Build GRCh38
Transcripts
NM_152446.3 XM_006720056.3 XM_011536490.2 XM_011536491.2 XM_011536492.2 XM_011536493.2 XM_011536495.2 XM_017021042.1 XM_017021043.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3498355 c.2478C>T p.S826S Chr.14: 80761512-80761512 on Build GRCh38
6076297 c.2403G>C p.E801D Chr.14: 80761587-80761587 on Build GRCh38
324853 c.2414G>T p.R805M Chr.14: 80761576-80761576 on Build GRCh38
2249613 c.2536T>G p.S846A Chr.14: 80761454-80761454 on Build GRCh38
7209303 c.2472G>T p.Q824H Chr.14: 80761518-80761518 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017188483 Chr.14: 80761499-80761499 on Build GRCh38
rs1005903947 Chr.14: 80761792-80761792 on Build GRCh38
rs370527843 Chr.14: 80761397-80761397 on Build GRCh38
rs556816897 Chr.14: 80761795-80761795 on Build GRCh38
rs535542337 Chr.14: 80761764-80761764 on Build GRCh38
rs1025974758 Chr.14: 80761452-80761452 on Build GRCh38
rs372845477 Chr.14: 80761401-80761401 on Build GRCh38
rs758978018 Chr.14: 80761625-80761625 on Build GRCh38
rs747161258 Chr.14: 80761528-80761528 on Build GRCh38
rs746150830 Chr.14: 80761657-80761657 on Build GRCh38
rs749089110 Chr.14: 80761563-80761563 on Build GRCh38
rs766443016 Chr.14: 80761683-80761683 on Build GRCh38
rs199812257 Chr.14: 80761622-80761622 on Build GRCh38
rs758386822 Chr.14: 80761523-80761523 on Build GRCh38
rs1029066572 Chr.14: 80761489-80761489 on Build GRCh38
rs545967020 Chr.14: 80761363-80761363 on Build GRCh38
rs561720833 Chr.14: 80761477-80761477 on Build GRCh38
rs781392285 Chr.14: 80761654-80761654 on Build GRCh38
rs768404700 Chr.14: 80761658-80761658 on Build GRCh38
rs150745114 Chr.14: 80761464-80761464 on Build GRCh38
rs1038867801 Chr.14: 80761751-80761751 on Build GRCh38
rs766615722 Chr.14: 80761462-80761462 on Build GRCh38
rs766601515 Chr.14: 80761496-80761496 on Build GRCh38
rs774247120 Chr.14: 80761573-80761573 on Build GRCh38
rs771208848 Chr.14: 80761542-80761542 on Build GRCh38
rs775388270 Chr.14: 80761444-80761444 on Build GRCh38
rs982210653 Chr.14: 80761448-80761448 on Build GRCh38
rs773294254 Chr.14: 80761415-80761415 on Build GRCh38
rs189966484 Chr.14: 80761800-80761800 on Build GRCh38
rs761007663 Chr.14: 80761419-80761419 on Build GRCh38
rs78462419 Chr.14: 80761805-80761805 on Build GRCh38
rs377022081 Chr.14: 80761571-80761571 on Build GRCh38
rs763485591 Chr.14: 80761802-80761802 on Build GRCh38
rs528953617 Chr.14: 80761534-80761534 on Build GRCh38
rs755467958 Chr.14: 80761517-80761517 on Build GRCh38
rs755389996 Chr.14: 80761398-80761409 on Build GRCh38
rs545306987 Chr.14: 80761410-80761410 on Build GRCh38
rs986626187 Chr.14: 80761315-80761315 on Build GRCh38
rs755380273 Chr.14: 80761599-80761599 on Build GRCh38
rs374944803 Chr.14: 80761414-80761414 on Build GRCh38
rs747220286 Chr.14: 80761396-80761396 on Build GRCh38
rs893007599 Chr.14: 80761371-80761371 on Build GRCh38
rs536341934 Chr.14: 80761687-80761687 on Build GRCh38
rs1042287569 Chr.14: 80761336-80761336 on Build GRCh38
rs147331133 Chr.14: 80761758-80761758 on Build GRCh38
rs777667019 Chr.14: 80761638-80761638 on Build GRCh38
rs1047477160 Chr.14: 80761777-80761777 on Build GRCh38
rs934609870 Chr.14: 80761314-80761314 on Build GRCh38
rs990535131 Chr.14: 80761674-80761674 on Build GRCh38
rs369684181 Chr.14: 80761583-80761583 on Build GRCh38
rs754267133 Chr.14: 80761508-80761508 on Build GRCh38
rs372099803 Chr.14: 80761469-80761469 on Build GRCh38
rs201692060 Chr.14: 80761421-80761421 on Build GRCh38
rs936330851 Chr.14: 80761574-80761574 on Build GRCh38
rs1023123940 Chr.14: 80761549-80761549 on Build GRCh38
rs761775653 Chr.14: 80761579-80761579 on Build GRCh38
rs765747629 Chr.14: 80761608-80761608 on Build GRCh38
rs912408809 Chr.14: 80761321-80761321 on Build GRCh38
rs757357729 Chr.14: 80761649-80761649 on Build GRCh38
rs949090628 Chr.14: 80761587-80761587 on Build GRCh38
rs766895052 Chr.14: 80761581-80761581 on Build GRCh38
rs940881185 Chr.14: 80761694-80761694 on Build GRCh38
rs1029385580 Chr.14: 80761767-80761767 on Build GRCh38
rs749769931 Chr.14: 80761390-80761390 on Build GRCh38
rs781736180 Chr.14: 80761575-80761576 on Build GRCh38
rs145540490 Chr.14: 80761703-80761703 on Build GRCh38
rs546638217 Chr.14: 80761714-80761714 on Build GRCh38
rs750008544 Chr.14: 80761474-80761474 on Build GRCh38
rs139374154 Chr.14: 80761648-80761648 on Build GRCh38
rs766057980 Chr.14: 80761589-80761589 on Build GRCh38
rs775211405 Chr.14: 80761555-80761555 on Build GRCh38
rs886148766 Chr.14: 80761782-80761782 on Build GRCh38
rs527954840 Chr.14: 80761713-80761713 on Build GRCh38
rs61749305 Chr.14: 80761527-80761527 on Build GRCh38
rs752969645 Chr.14: 80761602-80761603 on Build GRCh38
rs748721479 Chr.14: 80761522-80761522 on Build GRCh38
rs768509138 Chr.14: 80761567-80761567 on Build GRCh38
rs767099184 Chr.14: 80761472-80761472 on Build GRCh38
rs185160947 Chr.14: 80761680-80761680 on Build GRCh38
rs759543468 Chr.14: 80761556-80761556 on Build GRCh38
rs762831918 Chr.14: 80761454-80761454 on Build GRCh38
rs533406401 Chr.14: 80761661-80761661 on Build GRCh38
rs977970537 Chr.14: 80761569-80761569 on Build GRCh38
rs74483141 Chr.14: 80761521-80761521 on Build GRCh38
rs774159202 Chr.14: 80761660-80761660 on Build GRCh38
rs753612391 Chr.14: 80761598-80761598 on Build GRCh38
rs780069846 Chr.14: 80761348-80761348 on Build GRCh38
rs894910550 Chr.14: 80761386-80761386 on Build GRCh38
rs945271542 Chr.14: 80761324-80761324 on Build GRCh38

View Full Product Details