Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 89160467-89161011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXN3
Entrez Gene ID
1112
Gene Name
forkhead box N3
Gene Aliases
C14orf116, CHES1, PRO1635
UniGene
Hs.434286
Gene Chromosome Location
Chr.14: 89156172-89619150 on Build GRCh38
Transcripts
NM_001085471.1 NM_005197.3

SNP Detail

SNP ID Chromosome Location
rs900791134 Chr.14: 89160918-89160923 on Build GRCh38
rs996821528 Chr.14: 89160926-89160926 on Build GRCh38
rs1015457180 Chr.14: 89160554-89160554 on Build GRCh38
rs545677812 Chr.14: 89160727-89160727 on Build GRCh38
rs775170146 Chr.14: 89160652-89160652 on Build GRCh38
rs980167670 Chr.14: 89160733-89160733 on Build GRCh38
rs891976431 Chr.14: 89160881-89160881 on Build GRCh38
rs928619462 Chr.14: 89160761-89160761 on Build GRCh38
rs372027650 Chr.14: 89160552-89160552 on Build GRCh38
rs185857691 Chr.14: 89160884-89160884 on Build GRCh38
rs530200860 Chr.14: 89160883-89160883 on Build GRCh38
rs190325774 Chr.14: 89160938-89160938 on Build GRCh38
rs1043291140 Chr.14: 89160900-89160905 on Build GRCh38
rs559533424 Chr.14: 89160559-89160559 on Build GRCh38
rs564227868 Chr.14: 89160562-89160562 on Build GRCh38
rs575209101 Chr.14: 89160525-89160525 on Build GRCh38
rs368205997 Chr.14: 89160502-89160502 on Build GRCh38
rs974001663 Chr.14: 89160623-89160623 on Build GRCh38
rs181882273 Chr.14: 89160536-89160536 on Build GRCh38
rs890765793 Chr.14: 89160956-89160958 on Build GRCh38
rs757326880 Chr.14: 89160674-89160675 on Build GRCh38
rs1053114456 Chr.14: 89160805-89160805 on Build GRCh38
rs568084596 Chr.14: 89160940-89160940 on Build GRCh38
rs948733000 Chr.14: 89160679-89160679 on Build GRCh38
rs528387086 Chr.14: 89160901-89160901 on Build GRCh38
rs938808951 Chr.14: 89160801-89160801 on Build GRCh38
rs947524065 Chr.14: 89160883-89160883 on Build GRCh38
rs866661398 Chr.14: 89160882-89160882 on Build GRCh38
rs961178869 Chr.14: 89160609-89160609 on Build GRCh38
rs532379675 Chr.14: 89160718-89160718 on Build GRCh38

View Full Product Details