Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 89160846-89161384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXN3
Entrez Gene ID
1112
Gene Name
forkhead box N3
Gene Aliases
C14orf116, CHES1, PRO1635
UniGene
Hs.434286
Gene Chromosome Location
Chr.14: 89156172-89619150 on Build GRCh38
Transcripts
NM_001085471.1 NM_005197.3

SNP Detail

SNP ID Chromosome Location
rs900791134 Chr.14: 89160918-89160923 on Build GRCh38
rs555620905 Chr.14: 89161296-89161296 on Build GRCh38
rs182571575 Chr.14: 89161163-89161163 on Build GRCh38
rs913279349 Chr.14: 89161233-89161233 on Build GRCh38
rs996821528 Chr.14: 89160926-89160926 on Build GRCh38
rs768035800 Chr.14: 89161067-89161069 on Build GRCh38
rs1043239043 Chr.14: 89161274-89161274 on Build GRCh38
rs980414239 Chr.14: 89161134-89161134 on Build GRCh38
rs988863116 Chr.14: 89161219-89161220 on Build GRCh38
rs891976431 Chr.14: 89160881-89160881 on Build GRCh38
rs201415250 Chr.14: 89161123-89161124 on Build GRCh38
rs185339454 Chr.14: 89161298-89161298 on Build GRCh38
rs961210389 Chr.14: 89161031-89161031 on Build GRCh38
rs535786672 Chr.14: 89161276-89161276 on Build GRCh38
rs185857691 Chr.14: 89160884-89160884 on Build GRCh38
rs530200860 Chr.14: 89160883-89160883 on Build GRCh38
rs190325774 Chr.14: 89160938-89160938 on Build GRCh38
rs1052420626 Chr.14: 89161306-89161306 on Build GRCh38
rs1043291140 Chr.14: 89160900-89160905 on Build GRCh38
rs763820430 Chr.14: 89161285-89161285 on Build GRCh38
rs546877994 Chr.14: 89161135-89161135 on Build GRCh38
rs938652930 Chr.14: 89161211-89161211 on Build GRCh38
rs202007563 Chr.14: 89161317-89161317 on Build GRCh38
rs530496409 Chr.14: 89160988-89160988 on Build GRCh38
rs890811987 Chr.14: 89161343-89161343 on Build GRCh38
rs947470305 Chr.14: 89161273-89161273 on Build GRCh38
rs201008151 Chr.14: 89161306-89161307 on Build GRCh38
rs890765793 Chr.14: 89160956-89160958 on Build GRCh38
rs570289133 Chr.14: 89161139-89161139 on Build GRCh38
rs1007865355 Chr.14: 89161355-89161355 on Build GRCh38
rs550729654 Chr.14: 89161029-89161029 on Build GRCh38
rs376979749 Chr.14: 89160990-89160990 on Build GRCh38
rs1027231548 Chr.14: 89161110-89161110 on Build GRCh38
rs973864348 Chr.14: 89161046-89161046 on Build GRCh38
rs540069988 Chr.14: 89161037-89161037 on Build GRCh38
rs769781326 Chr.14: 89161305-89161306 on Build GRCh38
rs762625374 Chr.14: 89161103-89161103 on Build GRCh38
rs571830408 Chr.14: 89161115-89161115 on Build GRCh38
rs568084596 Chr.14: 89160940-89160940 on Build GRCh38
rs71456683 Chr.14: 89161161-89161162 on Build GRCh38
rs1015326183 Chr.14: 89161010-89161010 on Build GRCh38
rs970035466 Chr.14: 89161113-89161113 on Build GRCh38
rs528387086 Chr.14: 89160901-89160901 on Build GRCh38
rs548476068 Chr.14: 89161131-89161131 on Build GRCh38
rs947524065 Chr.14: 89160883-89160883 on Build GRCh38
rs866661398 Chr.14: 89160882-89160882 on Build GRCh38
rs570444854 Chr.14: 89161032-89161032 on Build GRCh38
rs928676997 Chr.14: 89161196-89161196 on Build GRCh38

View Full Product Details