Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 90401167-90401642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALM1
Entrez Gene ID
801
Gene Name
calmodulin 1
Gene Aliases
CALML2, CAMI, CPVT4, DD132, LQT14, PHKD, caM
UniGene
Hs.282410
Gene Chromosome Location
Chr.14: 90396983-90408275 on Build GRCh38
Transcripts
NM_006888.4 XM_006720258.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1707695 c.110T>C p.M37T Chr.14: 90401334-90401334 on Build GRCh38
470327 c.178G>A p.G60S Chr.14: 90401402-90401402 on Build GRCh38
6035610 c.173C>T p.A58V Chr.14: 90401397-90401397 on Build GRCh38
39923 c.118C>T p.L40L Chr.14: 90401342-90401342 on Build GRCh38
416578 c.91A>G p.K31E Chr.14: 90401315-90401315 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767419328 Chr.14: 90401552-90401552 on Build GRCh38
rs779857950 Chr.14: 90401424-90401424 on Build GRCh38
rs34704743 Chr.14: 90401485-90401486 on Build GRCh38
rs1046712559 Chr.14: 90401235-90401235 on Build GRCh38
rs946306790 Chr.14: 90401249-90401249 on Build GRCh38
rs894919394 Chr.14: 90401434-90401434 on Build GRCh38
rs559624540 Chr.14: 90401474-90401474 on Build GRCh38
rs1029786659 Chr.14: 90401568-90401568 on Build GRCh38
rs755114167 Chr.14: 90401444-90401445 on Build GRCh38
rs368437284 Chr.14: 90401238-90401238 on Build GRCh38
rs376460656 Chr.14: 90401524-90401524 on Build GRCh38
rs781354745 Chr.14: 90401444-90401447 on Build GRCh38
rs755900663 Chr.14: 90401422-90401422 on Build GRCh38
rs976905497 Chr.14: 90401239-90401239 on Build GRCh38
rs144339242 Chr.14: 90401293-90401293 on Build GRCh38
rs976746114 Chr.14: 90401533-90401534 on Build GRCh38
rs964172968 Chr.14: 90401554-90401554 on Build GRCh38
rs759490368 Chr.14: 90401437-90401437 on Build GRCh38
rs1037442309 Chr.14: 90401213-90401213 on Build GRCh38
rs765418586 Chr.14: 90401241-90401241 on Build GRCh38
rs879185853 Chr.14: 90401255-90401255 on Build GRCh38
rs984527673 Chr.14: 90401226-90401226 on Build GRCh38
rs922742387 Chr.14: 90401245-90401245 on Build GRCh38
rs977602970 Chr.14: 90401278-90401278 on Build GRCh38
rs376789051 Chr.14: 90401450-90401450 on Build GRCh38
rs967558903 Chr.14: 90401570-90401570 on Build GRCh38
rs761006471 Chr.14: 90401225-90401225 on Build GRCh38
rs267607276 Chr.14: 90401385-90401385 on Build GRCh38
rs773804229 Chr.14: 90401208-90401208 on Build GRCh38
rs201018787 Chr.14: 90401420-90401420 on Build GRCh38
rs781374289 Chr.14: 90401557-90401557 on Build GRCh38
rs879058953 Chr.14: 90401257-90401257 on Build GRCh38
rs893962552 Chr.14: 90401468-90401468 on Build GRCh38
rs752946983 Chr.14: 90401499-90401499 on Build GRCh38
rs751867539 Chr.14: 90401434-90401436 on Build GRCh38
rs977551988 Chr.14: 90401602-90401602 on Build GRCh38
rs188486459 Chr.14: 90401494-90401494 on Build GRCh38
rs866819225 Chr.14: 90401607-90401607 on Build GRCh38
rs199950662 Chr.14: 90401343-90401343 on Build GRCh38
rs554372598 Chr.14: 90401365-90401365 on Build GRCh38
rs202194260 Chr.14: 90401373-90401373 on Build GRCh38
rs577729656 Chr.14: 90401234-90401234 on Build GRCh38
rs1005581729 Chr.14: 90401531-90401531 on Build GRCh38
rs199897752 Chr.14: 90401349-90401349 on Build GRCh38
rs936231727 Chr.14: 90401616-90401616 on Build GRCh38
rs267607278 Chr.14: 90401296-90401296 on Build GRCh38
rs746430354 Chr.14: 90401563-90401563 on Build GRCh38
rs754854631 Chr.14: 90401445-90401445 on Build GRCh38
rs373975440 Chr.14: 90401230-90401230 on Build GRCh38
rs911346105 Chr.14: 90401228-90401228 on Build GRCh38
rs766745492 Chr.14: 90401227-90401227 on Build GRCh38
rs28691160 Chr.14: 90401214-90401214 on Build GRCh38
rs747812283 Chr.14: 90401209-90401209 on Build GRCh38
rs543344514 Chr.14: 90401427-90401427 on Build GRCh38
rs1046277177 Chr.14: 90401463-90401463 on Build GRCh38
rs1030209550 Chr.14: 90401590-90401590 on Build GRCh38

View Full Product Details