Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 90403777-90404286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALM1
Entrez Gene ID
801
Gene Name
calmodulin 1
Gene Aliases
CALML2, CAMI, CPVT4, DD132, LQT14, PHKD, caM
UniGene
Hs.282410
Gene Chromosome Location
Chr.14: 90396983-90408275 on Build GRCh38
Transcripts
NM_006888.4 XM_006720258.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6076588 c.271C>T p.R91* Chr.14: 90403954-90403954 on Build GRCh38
6141515 c.219G>T p.M73I Chr.14: 90403902-90403902 on Build GRCh38
433463 c.201C>T p.P67P Chr.14: 90403884-90403884 on Build GRCh38
4404842 c.270C>T p.F90F Chr.14: 90403953-90403953 on Build GRCh38
4668913 c.191T>C p.I64T Chr.14: 90403874-90403874 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770548612 Chr.14: 90403990-90403990 on Build GRCh38
rs1027237323 Chr.14: 90403824-90403825 on Build GRCh38
rs371830608 Chr.14: 90403829-90403829 on Build GRCh38
rs11541171 Chr.14: 90403955-90403955 on Build GRCh38
rs763174765 Chr.14: 90403858-90403858 on Build GRCh38
rs147717934 Chr.14: 90403823-90403823 on Build GRCh38
rs138389360 Chr.14: 90404145-90404145 on Build GRCh38
rs748117698 Chr.14: 90403826-90403829 on Build GRCh38
rs191075238 Chr.14: 90404016-90404016 on Build GRCh38
rs563783047 Chr.14: 90404205-90404205 on Build GRCh38
rs746645163 Chr.14: 90403831-90403831 on Build GRCh38
rs762047997 Chr.14: 90403884-90403884 on Build GRCh38
rs556733562 Chr.14: 90403917-90403917 on Build GRCh38
rs771561540 Chr.14: 90403819-90403819 on Build GRCh38
rs536790792 Chr.14: 90404207-90404207 on Build GRCh38
rs183342142 Chr.14: 90404222-90404222 on Build GRCh38
rs776993939 Chr.14: 90403837-90403837 on Build GRCh38
rs182076004 Chr.14: 90403813-90403813 on Build GRCh38
rs757946033 Chr.14: 90403985-90403985 on Build GRCh38
rs548158380 Chr.14: 90404254-90404254 on Build GRCh38
rs776127091 Chr.14: 90403854-90403854 on Build GRCh38
rs1036766513 Chr.14: 90404118-90404118 on Build GRCh38
rs761927969 Chr.14: 90403818-90403818 on Build GRCh38
rs1020732375 Chr.14: 90403987-90403987 on Build GRCh38
rs780700616 Chr.14: 90403992-90403992 on Build GRCh38
rs759282337 Chr.14: 90404059-90404059 on Build GRCh38
rs765178797 Chr.14: 90404189-90404189 on Build GRCh38
rs771218827 Chr.14: 90403836-90403836 on Build GRCh38
rs770485969 Chr.14: 90403841-90403841 on Build GRCh38
rs761748981 Chr.14: 90404036-90404038 on Build GRCh38
rs778775777 Chr.14: 90403998-90403998 on Build GRCh38
rs185096080 Chr.14: 90403988-90403988 on Build GRCh38
rs764201432 Chr.14: 90403859-90403859 on Build GRCh38
rs368792536 Chr.14: 90404017-90404017 on Build GRCh38
rs1035830389 Chr.14: 90404231-90404233 on Build GRCh38
rs970666396 Chr.14: 90403825-90403825 on Build GRCh38
rs3213719 Chr.14: 90403812-90403812 on Build GRCh38
rs751759267 Chr.14: 90403866-90403866 on Build GRCh38
rs374780692 Chr.14: 90403989-90403989 on Build GRCh38
rs567763595 Chr.14: 90404259-90404259 on Build GRCh38
rs375506636 Chr.14: 90404117-90404117 on Build GRCh38
rs753602780 Chr.14: 90403929-90403929 on Build GRCh38
rs778883623 Chr.14: 90403977-90403977 on Build GRCh38
rs770904447 Chr.14: 90403830-90403840 on Build GRCh38
rs142658217 Chr.14: 90404078-90404078 on Build GRCh38
rs188126903 Chr.14: 90404230-90404230 on Build GRCh38
rs146899770 Chr.14: 90404136-90404136 on Build GRCh38
rs760121129 Chr.14: 90403840-90403840 on Build GRCh38
rs116807296 Chr.14: 90404238-90404238 on Build GRCh38
rs754764861 Chr.14: 90403975-90403975 on Build GRCh38
rs730882253 Chr.14: 90403951-90403951 on Build GRCh38
rs1049127619 Chr.14: 90404158-90404158 on Build GRCh38
rs200250055 Chr.14: 90403926-90403926 on Build GRCh38
rs1010349299 Chr.14: 90404245-90404245 on Build GRCh38
rs561167530 Chr.14: 90404133-90404133 on Build GRCh38
rs752613114 Chr.14: 90403980-90403980 on Build GRCh38
rs985237794 Chr.14: 90404036-90404036 on Build GRCh38

View Full Product Details