Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 91114196-91114696 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf159
Entrez Gene ID
80017
Gene Name
chromosome 14 open reading frame 159
Gene Aliases
-
UniGene
Hs.309849
Gene Chromosome Location
Chr.14: 91114013-91225396 on Build GRCh38
Transcripts
NM_001102366.2 NM_001102367.2 NM_001102368.2 NM_001102369.2 NM_001286470.1 NM_001286472.1 NM_001286473.1 NM_024952.7

SNP Detail

SNP ID Chromosome Location
rs577431885 Chr.14: 91114335-91114335 on Build GRCh38
rs1017817502 Chr.14: 91114385-91114385 on Build GRCh38
rs1054884664 Chr.14: 91114238-91114238 on Build GRCh38
rs117688873 Chr.14: 91114503-91114503 on Build GRCh38
rs934656759 Chr.14: 91114544-91114544 on Build GRCh38
rs909109210 Chr.14: 91114463-91114463 on Build GRCh38
rs957790612 Chr.14: 91114342-91114342 on Build GRCh38
rs939127098 Chr.14: 91114554-91114554 on Build GRCh38
rs1056329991 Chr.14: 91114555-91114555 on Build GRCh38
rs756086432 Chr.14: 91114483-91114483 on Build GRCh38
rs948846354 Chr.14: 91114261-91114261 on Build GRCh38
rs941677 Chr.14: 91114612-91114612 on Build GRCh38
rs148308026 Chr.14: 91114479-91114479 on Build GRCh38
rs557354442 Chr.14: 91114326-91114326 on Build GRCh38
rs775373018 Chr.14: 91114651-91114651 on Build GRCh38
rs1056359707 Chr.14: 91114354-91114354 on Build GRCh38
rs189165079 Chr.14: 91114262-91114262 on Build GRCh38
rs903272513 Chr.14: 91114527-91114527 on Build GRCh38
rs996542901 Chr.14: 91114673-91114673 on Build GRCh38
rs530506730 Chr.14: 91114558-91114558 on Build GRCh38
rs868093042 Chr.14: 91114282-91114282 on Build GRCh38
rs1054889584 Chr.14: 91114602-91114602 on Build GRCh38
rs980217362 Chr.14: 91114531-91114531 on Build GRCh38
rs770905802 Chr.14: 91114244-91114244 on Build GRCh38
rs887000919 Chr.14: 91114605-91114605 on Build GRCh38
rs965778592 Chr.14: 91114390-91114390 on Build GRCh38
rs948830087 Chr.14: 91114517-91114517 on Build GRCh38
rs559849205 Chr.14: 91114386-91114386 on Build GRCh38
rs766449561 Chr.14: 91114498-91114498 on Build GRCh38
rs868544905 Chr.14: 91114465-91114465 on Build GRCh38
rs887026458 Chr.14: 91114371-91114371 on Build GRCh38
rs999263359 Chr.14: 91114284-91114284 on Build GRCh38
rs1007716541 Chr.14: 91114367-91114367 on Build GRCh38
rs767694373 Chr.14: 91114259-91114259 on Build GRCh38
rs1054871185 Chr.14: 91114467-91114468 on Build GRCh38
rs997022082 Chr.14: 91114445-91114445 on Build GRCh38
rs528703989 Chr.14: 91114457-91114457 on Build GRCh38
rs919005436 Chr.14: 91114420-91114420 on Build GRCh38
rs1044902646 Chr.14: 91114269-91114269 on Build GRCh38
rs975896649 Chr.14: 91114399-91114399 on Build GRCh38
rs962475817 Chr.14: 91114421-91114421 on Build GRCh38
rs934641853 Chr.14: 91114222-91114222 on Build GRCh38
rs1031863181 Chr.14: 91114482-91114482 on Build GRCh38
rs957628750 Chr.14: 91114347-91114348 on Build GRCh38
rs928713528 Chr.14: 91114532-91114532 on Build GRCh38
rs939959250 Chr.14: 91114609-91114609 on Build GRCh38
rs905067454 Chr.14: 91114310-91114310 on Build GRCh38
rs894545903 Chr.14: 91114240-91114240 on Build GRCh38
rs570378413 Chr.14: 91114618-91114618 on Build GRCh38
rs181825270 Chr.14: 91114320-91114320 on Build GRCh38
rs1028442974 Chr.14: 91114448-91114448 on Build GRCh38
rs1017024755 Chr.14: 91114419-91114419 on Build GRCh38
rs546199834 Chr.14: 91114376-91114376 on Build GRCh38

View Full Product Details