Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 91175693-91176199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf159
Entrez Gene ID
80017
Gene Name
chromosome 14 open reading frame 159
Gene Aliases
-
UniGene
Hs.309849
Gene Chromosome Location
Chr.14: 91114013-91225396 on Build GRCh38
Transcripts
NM_001102366.2 NM_001102367.2 NM_001102368.2 NM_001102369.2 NM_001286470.1 NM_001286471.1 NM_001286472.1 NM_024952.7

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3987823 c.636T>A p.D212E Chr.14: 91175977-91175977 on Build GRCh38
5880820 c.710G>A p.S237N Chr.14: 91176051-91176051 on Build GRCh38
1608053 c.680_684delCTCCG p.P228fs*10 Chr.14: 91176021-91176025 on Build GRCh38
5580386 c.610G>A p.E204K Chr.14: 91175951-91175951 on Build GRCh38
5964999 c.621A>G p.K207K Chr.14: 91175962-91175962 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150109265 Chr.14: 91175746-91175746 on Build GRCh38
rs751597132 Chr.14: 91176005-91176005 on Build GRCh38
rs754989441 Chr.14: 91176030-91176030 on Build GRCh38
rs764043338 Chr.14: 91176102-91176102 on Build GRCh38
rs904734650 Chr.14: 91175816-91175816 on Build GRCh38
rs1055947561 Chr.14: 91176065-91176065 on Build GRCh38
rs888387168 Chr.14: 91176133-91176133 on Build GRCh38
rs758301013 Chr.14: 91176033-91176033 on Build GRCh38
rs778679270 Chr.14: 91176028-91176028 on Build GRCh38
rs1006624576 Chr.14: 91175805-91175805 on Build GRCh38
rs777154309 Chr.14: 91176035-91176035 on Build GRCh38
rs764201769 Chr.14: 91175932-91175932 on Build GRCh38
rs12323858 Chr.14: 91175733-91175733 on Build GRCh38
rs767469352 Chr.14: 91176070-91176070 on Build GRCh38
rs760125548 Chr.14: 91175884-91175884 on Build GRCh38
rs1055758500 Chr.14: 91175823-91175823 on Build GRCh38
rs1014310171 Chr.14: 91175875-91175875 on Build GRCh38
rs1042369627 Chr.14: 91175754-91175754 on Build GRCh38
rs993980523 Chr.14: 91176162-91176162 on Build GRCh38
rs778769291 Chr.14: 91175970-91175970 on Build GRCh38
rs142118168 Chr.14: 91175990-91175990 on Build GRCh38
rs532526240 Chr.14: 91175968-91175968 on Build GRCh38
rs149668017 Chr.14: 91176025-91176025 on Build GRCh38
rs775982099 Chr.14: 91176054-91176054 on Build GRCh38
rs770552780 Chr.14: 91175981-91175981 on Build GRCh38
rs530749288 Chr.14: 91175858-91175858 on Build GRCh38
rs35013049 Chr.14: 91176073-91176074 on Build GRCh38
rs559595227 Chr.14: 91175808-91175808 on Build GRCh38
rs550265359 Chr.14: 91175878-91175878 on Build GRCh38
rs770513216 Chr.14: 91176038-91176038 on Build GRCh38
rs761899157 Chr.14: 91176066-91176066 on Build GRCh38
rs45613242 Chr.14: 91175821-91175821 on Build GRCh38
rs556617649 Chr.14: 91176109-91176109 on Build GRCh38
rs759931341 Chr.14: 91175987-91175987 on Build GRCh38
rs753560430 Chr.14: 91175967-91175967 on Build GRCh38
rs754811826 Chr.14: 91176026-91176026 on Build GRCh38
rs75931548 Chr.14: 91175996-91175996 on Build GRCh38
rs952533205 Chr.14: 91175889-91175889 on Build GRCh38
rs377648382 Chr.14: 91176144-91176144 on Build GRCh38
rs776180503 Chr.14: 91175992-91175992 on Build GRCh38
rs757486874 Chr.14: 91175955-91175955 on Build GRCh38
rs748049947 Chr.14: 91175971-91175971 on Build GRCh38
rs201939538 Chr.14: 91176082-91176082 on Build GRCh38
rs761052329 Chr.14: 91175954-91175954 on Build GRCh38
rs376295292 Chr.14: 91175979-91175979 on Build GRCh38
rs781285360 Chr.14: 91176083-91176083 on Build GRCh38
rs751794100 Chr.14: 91175945-91175945 on Build GRCh38
rs774711853 Chr.14: 91175732-91175732 on Build GRCh38
rs561688369 Chr.14: 91175842-91175842 on Build GRCh38
rs757192393 Chr.14: 91176061-91176061 on Build GRCh38
rs373892481 Chr.14: 91175924-91175924 on Build GRCh38
rs777327159 Chr.14: 91175974-91175974 on Build GRCh38
rs1046758795 Chr.14: 91176115-91176115 on Build GRCh38
rs776890953 Chr.14: 91175982-91175982 on Build GRCh38
rs756195721 Chr.14: 91176021-91176021 on Build GRCh38
rs185548479 Chr.14: 91175800-91175800 on Build GRCh38
rs758300679 Chr.14: 91176100-91176100 on Build GRCh38
rs114448340 Chr.14: 91176002-91176002 on Build GRCh38
rs766548223 Chr.14: 91176024-91176024 on Build GRCh38
rs565144675 Chr.14: 91176136-91176136 on Build GRCh38
rs374050251 Chr.14: 91176062-91176062 on Build GRCh38
rs34302825 Chr.14: 91176051-91176051 on Build GRCh38
rs771553587 Chr.14: 91175972-91175972 on Build GRCh38
rs779723928 Chr.14: 91175965-91175965 on Build GRCh38
rs2295528 Chr.14: 91175890-91175890 on Build GRCh38
rs1025504382 Chr.14: 91176172-91176172 on Build GRCh38
rs779537404 Chr.14: 91176176-91176176 on Build GRCh38
rs752424839 Chr.14: 91176086-91176086 on Build GRCh38
rs148400411 Chr.14: 91176044-91176044 on Build GRCh38
rs141327847 Chr.14: 91176060-91176060 on Build GRCh38
rs769738774 Chr.14: 91176163-91176163 on Build GRCh38
rs889573794 Chr.14: 91175794-91175794 on Build GRCh38
rs193195256 Chr.14: 91175790-91175790 on Build GRCh38
rs1046235500 Chr.14: 91176121-91176121 on Build GRCh38
rs765800862 Chr.14: 91175886-91175886 on Build GRCh38
rs1041137841 Chr.14: 91175813-91175813 on Build GRCh38
rs115379791 Chr.14: 91175828-91175828 on Build GRCh38
rs115154303 Chr.14: 91176037-91176037 on Build GRCh38
rs761942648 Chr.14: 91176020-91176020 on Build GRCh38
rs768264755 Chr.14: 91175727-91175727 on Build GRCh38

View Full Product Details