Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 92932203-92932694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHGA
Entrez Gene ID
1113
Gene Name
chromogranin A
Gene Aliases
CGA
UniGene
Hs.150793
Gene Chromosome Location
Chr.14: 92922589-92935293 on Build GRCh38
Transcripts
NM_001275.3 NM_001301690.1 XM_011536370.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7361175 c.821C>G p.A274G Chr.14: 92932382-92932382 on Build GRCh38
5459903 c.1130G>A p.R377Q Chr.14: 92932691-92932691 on Build GRCh38
7361174 c.1105G>A p.D369N Chr.14: 92932666-92932666 on Build GRCh38
6458544 c.1082_1084delAGG p.E365delE Chr.14: 92932643-92932645 on Build GRCh38
6567935 c.1063G>T p.E355* Chr.14: 92932624-92932624 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs985223077 Chr.14: 92932575-92932575 on Build GRCh38
rs778247059 Chr.14: 92932528-92932528 on Build GRCh38
rs748718190 Chr.14: 92932500-92932500 on Build GRCh38
rs9658661 Chr.14: 92932251-92932251 on Build GRCh38
rs985275420 Chr.14: 92932428-92932428 on Build GRCh38
rs897140526 Chr.14: 92932533-92932533 on Build GRCh38
rs564902629 Chr.14: 92932397-92932397 on Build GRCh38
rs896522084 Chr.14: 92932513-92932513 on Build GRCh38
rs770895936 Chr.14: 92932470-92932471 on Build GRCh38
rs746161501 Chr.14: 92932664-92932664 on Build GRCh38
rs1019441332 Chr.14: 92932582-92932582 on Build GRCh38
rs556289170 Chr.14: 92932365-92932365 on Build GRCh38
rs371137176 Chr.14: 92932435-92932435 on Build GRCh38
rs201406253 Chr.14: 92932637-92932637 on Build GRCh38
rs760193216 Chr.14: 92932621-92932621 on Build GRCh38
rs778452104 Chr.14: 92932645-92932645 on Build GRCh38
rs764149977 Chr.14: 92932420-92932420 on Build GRCh38
rs912997626 Chr.14: 92932653-92932653 on Build GRCh38
rs747167515 Chr.14: 92932539-92932539 on Build GRCh38
rs767989572 Chr.14: 92932631-92932631 on Build GRCh38
rs764112390 Chr.14: 92932646-92932647 on Build GRCh38
rs750275509 Chr.14: 92932604-92932604 on Build GRCh38
rs554091679 Chr.14: 92932282-92932282 on Build GRCh38
rs199660196 Chr.14: 92932252-92932252 on Build GRCh38
rs775763691 Chr.14: 92932670-92932670 on Build GRCh38
rs572031425 Chr.14: 92932400-92932400 on Build GRCh38
rs532226129 Chr.14: 92932552-92932552 on Build GRCh38
rs757656588 Chr.14: 92932619-92932619 on Build GRCh38
rs9658663 Chr.14: 92932382-92932382 on Build GRCh38
rs574139130 Chr.14: 92932284-92932284 on Build GRCh38
rs1027191805 Chr.14: 92932614-92932614 on Build GRCh38
rs752136557 Chr.14: 92932554-92932554 on Build GRCh38
rs200580684 Chr.14: 92932355-92932355 on Build GRCh38
rs9658664 Chr.14: 92932504-92932504 on Build GRCh38
rs534568059 Chr.14: 92932527-92932527 on Build GRCh38
rs753407523 Chr.14: 92932460-92932460 on Build GRCh38
rs1044035713 Chr.14: 92932244-92932244 on Build GRCh38
rs374405768 Chr.14: 92932647-92932647 on Build GRCh38
rs779403860 Chr.14: 92932440-92932440 on Build GRCh38
rs767197817 Chr.14: 92932595-92932595 on Build GRCh38
rs776903552 Chr.14: 92932283-92932283 on Build GRCh38
rs761902208 Chr.14: 92932583-92932583 on Build GRCh38
rs2228575 Chr.14: 92932666-92932666 on Build GRCh38
rs912266066 Chr.14: 92932429-92932429 on Build GRCh38
rs961714095 Chr.14: 92932370-92932370 on Build GRCh38
rs980506099 Chr.14: 92932318-92932318 on Build GRCh38
rs761706024 Chr.14: 92932433-92932433 on Build GRCh38
rs960388876 Chr.14: 92932341-92932343 on Build GRCh38
rs185229400 Chr.14: 92932537-92932537 on Build GRCh38
rs1043983476 Chr.14: 92932497-92932497 on Build GRCh38
rs774428465 Chr.14: 92932557-92932557 on Build GRCh38
rs755578275 Chr.14: 92932341-92932341 on Build GRCh38
rs2236132 Chr.14: 92932325-92932325 on Build GRCh38
rs771000157 Chr.14: 92932659-92932659 on Build GRCh38
rs745320335 Chr.14: 92932328-92932328 on Build GRCh38
rs751728985 Chr.14: 92932643-92932645 on Build GRCh38
rs947065632 Chr.14: 92932485-92932485 on Build GRCh38
rs565635101 Chr.14: 92932422-92932422 on Build GRCh38
rs755086015 Chr.14: 92932643-92932651 on Build GRCh38
rs1016126585 Chr.14: 92932521-92932521 on Build GRCh38
rs147297806 Chr.14: 92932514-92932514 on Build GRCh38
rs747424184 Chr.14: 92932656-92932656 on Build GRCh38
rs755415831 Chr.14: 92932472-92932472 on Build GRCh38
rs763325622 Chr.14: 92932541-92932543 on Build GRCh38
rs570207965 Chr.14: 92932642-92932643 on Build GRCh38
rs781483407 Chr.14: 92932663-92932663 on Build GRCh38
rs552215557 Chr.14: 92932553-92932553 on Build GRCh38
rs763386106 Chr.14: 92932438-92932438 on Build GRCh38
rs779394961 Chr.14: 92932494-92932494 on Build GRCh38
rs200777605 Chr.14: 92932413-92932413 on Build GRCh38
rs761013910 Chr.14: 92932392-92932392 on Build GRCh38
rs753676670 Chr.14: 92932649-92932650 on Build GRCh38
rs779678001 Chr.14: 92932350-92932350 on Build GRCh38
rs869079468 Chr.14: 92932474-92932474 on Build GRCh38
rs9658665 Chr.14: 92932556-92932556 on Build GRCh38
rs113880894 Chr.14: 92932356-92932356 on Build GRCh38
rs749739890 Chr.14: 92932353-92932353 on Build GRCh38
rs748523969 Chr.14: 92932373-92932373 on Build GRCh38
rs9658662 Chr.14: 92932372-92932372 on Build GRCh38
rs755848437 Chr.14: 92932439-92932439 on Build GRCh38
rs772213323 Chr.14: 92932377-92932377 on Build GRCh38
rs1033740555 Chr.14: 92932255-92932255 on Build GRCh38
rs546878181 Chr.14: 92932630-92932630 on Build GRCh38
rs765455865 Chr.14: 92932471-92932473 on Build GRCh38
rs771215098 Chr.14: 92932549-92932549 on Build GRCh38
rs747999063 Chr.14: 92932245-92932245 on Build GRCh38
rs563400835 Chr.14: 92932551-92932551 on Build GRCh38
rs190999870 Chr.14: 92932434-92932434 on Build GRCh38
rs912890820 Chr.14: 92932442-92932442 on Build GRCh38
rs762272580 Chr.14: 92932323-92932323 on Build GRCh38
rs140089514 Chr.14: 92932477-92932477 on Build GRCh38
rs953024292 Chr.14: 92932308-92932308 on Build GRCh38
rs904123149 Chr.14: 92932499-92932499 on Build GRCh38
rs977945873 Chr.14: 92932390-92932390 on Build GRCh38

View Full Product Details