Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 92934562-92935068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHGA
Entrez Gene ID
1113
Gene Name
chromogranin A
Gene Aliases
CGA
UniGene
Hs.150793
Gene Chromosome Location
Chr.14: 92922589-92935293 on Build GRCh38
Transcripts
NM_001275.3 NM_001301690.1 XM_011536370.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4564688 c.1367_1368GG>AA p.R456Q Chr.14: 92934877-92934878 on Build GRCh38
1201042 c.1366C>T p.R456W Chr.14: 92934876-92934876 on Build GRCh38
232034 c.1298A>T p.E433V Chr.14: 92934808-92934808 on Build GRCh38
6042811 c.1310T>C p.L437P Chr.14: 92934820-92934820 on Build GRCh38
3754178 c.1323A>G p.E441E Chr.14: 92934833-92934833 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369835117 Chr.14: 92934775-92934775 on Build GRCh38
rs908929797 Chr.14: 92934661-92934661 on Build GRCh38
rs534157810 Chr.14: 92934874-92934874 on Build GRCh38
rs530828278 Chr.14: 92935026-92935026 on Build GRCh38
rs553435018 Chr.14: 92934981-92934981 on Build GRCh38
rs1023498822 Chr.14: 92934777-92934777 on Build GRCh38
rs781112577 Chr.14: 92934823-92934823 on Build GRCh38
rs1016046965 Chr.14: 92934845-92934845 on Build GRCh38
rs903452850 Chr.14: 92934724-92934724 on Build GRCh38
rs1803277 Chr.14: 92935017-92935017 on Build GRCh38
rs779009398 Chr.14: 92934784-92934784 on Build GRCh38
rs75909915 Chr.14: 92934599-92934599 on Build GRCh38
rs773837740 Chr.14: 92934876-92934876 on Build GRCh38
rs141005266 Chr.14: 92934877-92934877 on Build GRCh38
rs779875386 Chr.14: 92934829-92934829 on Build GRCh38
rs761247326 Chr.14: 92934870-92934870 on Build GRCh38
rs1026212391 Chr.14: 92934628-92934628 on Build GRCh38
rs757128358 Chr.14: 92934816-92934816 on Build GRCh38
rs974297819 Chr.14: 92934730-92934730 on Build GRCh38
rs963763202 Chr.14: 92934917-92934917 on Build GRCh38
rs907182537 Chr.14: 92934787-92934787 on Build GRCh38
rs373388498 Chr.14: 92935035-92935035 on Build GRCh38
rs948616088 Chr.14: 92934738-92934738 on Build GRCh38
rs535317688 Chr.14: 92934850-92934850 on Build GRCh38
rs933007387 Chr.14: 92934714-92934714 on Build GRCh38
rs981275445 Chr.14: 92934975-92934975 on Build GRCh38
rs7610 Chr.14: 92934971-92934971 on Build GRCh38
rs1047723713 Chr.14: 92935031-92935031 on Build GRCh38
rs915704824 Chr.14: 92934946-92934946 on Build GRCh38
rs187887130 Chr.14: 92935027-92935027 on Build GRCh38
rs375104177 Chr.14: 92934600-92934600 on Build GRCh38
rs570816459 Chr.14: 92934706-92934706 on Build GRCh38
rs1054747187 Chr.14: 92934906-92934906 on Build GRCh38
rs867997329 Chr.14: 92934857-92934857 on Build GRCh38
rs11555439 Chr.14: 92935029-92935029 on Build GRCh38
rs763939201 Chr.14: 92934755-92934755 on Build GRCh38
rs9658672 Chr.14: 92934980-92934980 on Build GRCh38
rs766816484 Chr.14: 92934885-92934885 on Build GRCh38
rs934782197 Chr.14: 92934728-92934728 on Build GRCh38
rs981368175 Chr.14: 92934608-92934608 on Build GRCh38
rs759752953 Chr.14: 92934891-92934891 on Build GRCh38
rs9658670 Chr.14: 92934729-92934729 on Build GRCh38
rs183255929 Chr.14: 92934632-92934632 on Build GRCh38
rs755193705 Chr.14: 92934776-92934776 on Build GRCh38
rs761113127 Chr.14: 92934760-92934760 on Build GRCh38
rs752740996 Chr.14: 92934813-92934813 on Build GRCh38
rs373342226 Chr.14: 92934890-92934890 on Build GRCh38
rs752332516 Chr.14: 92934716-92934716 on Build GRCh38
rs142980634 Chr.14: 92935037-92935037 on Build GRCh38
rs1016159807 Chr.14: 92935038-92935038 on Build GRCh38
rs745873520 Chr.14: 92934824-92934824 on Build GRCh38
rs142383456 Chr.14: 92934687-92934687 on Build GRCh38
rs577137764 Chr.14: 92934593-92934593 on Build GRCh38
rs769565118 Chr.14: 92934827-92934827 on Build GRCh38
rs1013294032 Chr.14: 92934596-92934596 on Build GRCh38
rs915758649 Chr.14: 92934737-92934737 on Build GRCh38
rs893290113 Chr.14: 92934937-92934937 on Build GRCh38
rs530993265 Chr.14: 92934662-92934662 on Build GRCh38
rs941581 Chr.14: 92934833-92934833 on Build GRCh38
rs550938426 Chr.14: 92935042-92935042 on Build GRCh38
rs150981613 Chr.14: 92934873-92934873 on Build GRCh38
rs1030314970 Chr.14: 92934633-92934633 on Build GRCh38
rs138998698 Chr.14: 92934904-92934904 on Build GRCh38
rs145505017 Chr.14: 92934839-92934839 on Build GRCh38
rs546620404 Chr.14: 92934736-92934736 on Build GRCh38
rs202023927 Chr.14: 92934896-92934896 on Build GRCh38
rs893348660 Chr.14: 92934772-92934772 on Build GRCh38
rs369458299 Chr.14: 92934637-92934637 on Build GRCh38
rs766906047 Chr.14: 92934768-92934768 on Build GRCh38
rs528718920 Chr.14: 92934616-92934616 on Build GRCh38
rs1017760007 Chr.14: 92934849-92934849 on Build GRCh38

View Full Product Details