Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 93203165-93203700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GON7
Entrez Gene ID
84520
Gene Name
GON7, KEOPS complex subunit homolog
Gene Aliases
C14orf142, PNAS-127
UniGene
Hs.20142
Gene Chromosome Location
Chr.14: 93202892-93207114 on Build GRCh38
Transcripts
NM_032490.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
958926 c.296C>T p.P99L Chr.14: 93203695-93203695 on Build GRCh38
1198064 c.303A>C p.*101Y Chr.14: 93203688-93203688 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs981235359 Chr.14: 93203559-93203559 on Build GRCh38
rs915359643 Chr.14: 93203277-93203277 on Build GRCh38
rs535902711 Chr.14: 93203645-93203645 on Build GRCh38
rs748761288 Chr.14: 93203656-93203656 on Build GRCh38
rs769420766 Chr.14: 93203643-93203643 on Build GRCh38
rs745946754 Chr.14: 93203289-93203289 on Build GRCh38
rs976583490 Chr.14: 93203469-93203469 on Build GRCh38
rs537110978 Chr.14: 93203492-93203492 on Build GRCh38
rs1022379880 Chr.14: 93203556-93203556 on Build GRCh38
rs968111995 Chr.14: 93203557-93203557 on Build GRCh38
rs768076439 Chr.14: 93203523-93203523 on Build GRCh38
rs961007887 Chr.14: 93203609-93203609 on Build GRCh38
rs146044377 Chr.14: 93203589-93203589 on Build GRCh38
rs762046820 Chr.14: 93203463-93203463 on Build GRCh38
rs895553012 Chr.14: 93203281-93203281 on Build GRCh38
rs769936878 Chr.14: 93203295-93203295 on Build GRCh38
rs951194600 Chr.14: 93203542-93203543 on Build GRCh38
rs752992257 Chr.14: 93203613-93203614 on Build GRCh38
rs768953664 Chr.14: 93203410-93203410 on Build GRCh38
rs781044407 Chr.14: 93203635-93203635 on Build GRCh38
rs1014448664 Chr.14: 93203324-93203324 on Build GRCh38
rs939296729 Chr.14: 93203345-93203345 on Build GRCh38
rs984005380 Chr.14: 93203564-93203564 on Build GRCh38
rs192133488 Chr.14: 93203359-93203359 on Build GRCh38
rs926583056 Chr.14: 93203330-93203330 on Build GRCh38
rs182594724 Chr.14: 93203660-93203660 on Build GRCh38
rs1007873768 Chr.14: 93203504-93203504 on Build GRCh38
rs973939467 Chr.14: 93203672-93203672 on Build GRCh38
rs1009532952 Chr.14: 93203203-93203203 on Build GRCh38
rs557001666 Chr.14: 93203581-93203581 on Build GRCh38
rs1049415699 Chr.14: 93203430-93203430 on Build GRCh38
rs1041465799 Chr.14: 93203674-93203674 on Build GRCh38
rs372634564 Chr.14: 93203223-93203223 on Build GRCh38
rs867427805 Chr.14: 93203378-93203378 on Build GRCh38
rs1007962957 Chr.14: 93203414-93203416 on Build GRCh38
rs552020762 Chr.14: 93203329-93203329 on Build GRCh38
rs34298427 Chr.14: 93203584-93203585 on Build GRCh38
rs752216558 Chr.14: 93203590-93203591 on Build GRCh38
rs528252356 Chr.14: 93203397-93203397 on Build GRCh38
rs756961982 Chr.14: 93203626-93203626 on Build GRCh38
rs142653634 Chr.14: 93203400-93203400 on Build GRCh38
rs908460781 Chr.14: 93203644-93203644 on Build GRCh38
rs1056761339 Chr.14: 93203347-93203347 on Build GRCh38
rs776760696 Chr.14: 93203619-93203620 on Build GRCh38
rs1009729182 Chr.14: 93203543-93203543 on Build GRCh38
rs1019127607 Chr.14: 93203204-93203204 on Build GRCh38
rs534536616 Chr.14: 93203428-93203428 on Build GRCh38
rs1041896118 Chr.14: 93203352-93203352 on Build GRCh38
rs957115123 Chr.14: 93203219-93203219 on Build GRCh38
rs774675117 Chr.14: 93203676-93203676 on Build GRCh38
rs939865368 Chr.14: 93203392-93203392 on Build GRCh38
rs745451666 Chr.14: 93203637-93203637 on Build GRCh38
rs779744808 Chr.14: 93203648-93203648 on Build GRCh38
rs749328506 Chr.14: 93203394-93203394 on Build GRCh38
rs139893467 Chr.14: 93203613-93203613 on Build GRCh38
rs892710223 Chr.14: 93203529-93203529 on Build GRCh38

View Full Product Details