Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 93240617-93241151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD7
Entrez Gene ID
55727
Gene Name
BTB domain containing 7
Gene Aliases
FUP1
UniGene
Hs.525549
Gene Chromosome Location
Chr.14: 93237550-93333092 on Build GRCh38
Transcripts
NM_001002860.3 NM_001289133.1

SNP Detail

SNP ID Chromosome Location
rs891629865 Chr.14: 93240859-93240859 on Build GRCh38
rs567254858 Chr.14: 93240856-93240856 on Build GRCh38
rs547541397 Chr.14: 93240694-93240694 on Build GRCh38
rs919957162 Chr.14: 93240748-93240748 on Build GRCh38
rs750302390 Chr.14: 93240664-93240664 on Build GRCh38
rs955319514 Chr.14: 93241015-93241016 on Build GRCh38
rs778886165 Chr.14: 93240913-93240913 on Build GRCh38
rs947971599 Chr.14: 93241090-93241090 on Build GRCh38
rs533952082 Chr.14: 93240718-93240718 on Build GRCh38
rs974111879 Chr.14: 93240729-93240729 on Build GRCh38
rs1030684045 Chr.14: 93240922-93240922 on Build GRCh38
rs558385506 Chr.14: 93241041-93241041 on Build GRCh38
rs1027047170 Chr.14: 93241082-93241082 on Build GRCh38
rs896738370 Chr.14: 93240970-93240970 on Build GRCh38
rs985456102 Chr.14: 93240793-93240793 on Build GRCh38
rs912554317 Chr.14: 93241108-93241108 on Build GRCh38
rs576163016 Chr.14: 93240953-93240953 on Build GRCh38
rs79912934 Chr.14: 93241089-93241089 on Build GRCh38
rs934056488 Chr.14: 93240735-93240736 on Build GRCh38
rs200065053 Chr.14: 93240654-93240654 on Build GRCh38
rs907713063 Chr.14: 93240762-93240762 on Build GRCh38
rs932659391 Chr.14: 93240758-93240758 on Build GRCh38
rs528824289 Chr.14: 93240832-93240833 on Build GRCh38
rs982884084 Chr.14: 93241031-93241031 on Build GRCh38
rs1056760713 Chr.14: 93240943-93240943 on Build GRCh38
rs1015738893 Chr.14: 93240983-93240983 on Build GRCh38
rs1022568423 Chr.14: 93240962-93240962 on Build GRCh38
rs922607373 Chr.14: 93241009-93241009 on Build GRCh38
rs1003247963 Chr.14: 93240858-93240858 on Build GRCh38
rs45600236 Chr.14: 93240938-93240938 on Build GRCh38
rs961512548 Chr.14: 93240991-93240991 on Build GRCh38
rs964105802 Chr.14: 93240667-93240667 on Build GRCh38
rs756025781 Chr.14: 93240878-93240878 on Build GRCh38
rs772524352 Chr.14: 93240756-93240764 on Build GRCh38
rs189552173 Chr.14: 93240857-93240857 on Build GRCh38
rs545150110 Chr.14: 93240968-93240968 on Build GRCh38
rs368227972 Chr.14: 93241085-93241085 on Build GRCh38
rs1013834946 Chr.14: 93240971-93240971 on Build GRCh38
rs943937844 Chr.14: 93240831-93240831 on Build GRCh38
rs781227171 Chr.14: 93240647-93240647 on Build GRCh38
rs947142441 Chr.14: 93240925-93240925 on Build GRCh38
rs553724849 Chr.14: 93240830-93240830 on Build GRCh38
rs1037639064 Chr.14: 93240800-93240800 on Build GRCh38
rs912473797 Chr.14: 93240795-93240795 on Build GRCh38
rs1042813090 Chr.14: 93240931-93240931 on Build GRCh38
rs867245956 Chr.14: 93240940-93240940 on Build GRCh38
rs940484351 Chr.14: 93240786-93240786 on Build GRCh38
rs539713864 Chr.14: 93240954-93240954 on Build GRCh38
rs867372423 Chr.14: 93241032-93241032 on Build GRCh38
rs772435044 Chr.14: 93241067-93241067 on Build GRCh38

View Full Product Details