Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 93245719-93246216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD7
Entrez Gene ID
55727
Gene Name
BTB domain containing 7
Gene Aliases
FUP1
UniGene
Hs.525549
Gene Chromosome Location
Chr.14: 93237550-93333092 on Build GRCh38
Transcripts
NM_001002860.3 NM_001289133.1 XM_011536939.2 XM_017021438.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2252261 c.2473C>T p.P825S Chr.14: 93245935-93245935 on Build GRCh38
5903067 c.1333C>T p.P445S Chr.14: 93246022-93246022 on Build GRCh38
6613398 c.1364C>T p.T455I Chr.14: 93245991-93245991 on Build GRCh38
5691868 c.2481T>A p.C827* Chr.14: 93245927-93245927 on Build GRCh38
5936049 c.1384C>T p.P462S Chr.14: 93245971-93245971 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775872289 Chr.14: 93246135-93246136 on Build GRCh38
rs376812979 Chr.14: 93245955-93245955 on Build GRCh38
rs774851734 Chr.14: 93246126-93246131 on Build GRCh38
rs964348401 Chr.14: 93245845-93245845 on Build GRCh38
rs769760612 Chr.14: 93246027-93246027 on Build GRCh38
rs764801597 Chr.14: 93245909-93245909 on Build GRCh38
rs772691463 Chr.14: 93246121-93246122 on Build GRCh38
rs372008556 Chr.14: 93246016-93246016 on Build GRCh38
rs367904111 Chr.14: 93246013-93246013 on Build GRCh38
rs775861811 Chr.14: 93245881-93245881 on Build GRCh38
rs752984452 Chr.14: 93246098-93246098 on Build GRCh38
rs768007931 Chr.14: 93246133-93246134 on Build GRCh38
rs769535893 Chr.14: 93246105-93246118 on Build GRCh38
rs772836761 Chr.14: 93246124-93246124 on Build GRCh38
rs756799672 Chr.14: 93245997-93245997 on Build GRCh38
rs535882603 Chr.14: 93245862-93245862 on Build GRCh38
rs370817738 Chr.14: 93245897-93245897 on Build GRCh38
rs555463968 Chr.14: 93245882-93245882 on Build GRCh38
rs762559046 Chr.14: 93245975-93245975 on Build GRCh38
rs376218202 Chr.14: 93245803-93245803 on Build GRCh38
rs140197697 Chr.14: 93245945-93245945 on Build GRCh38
rs757362672 Chr.14: 93246161-93246161 on Build GRCh38
rs763144352 Chr.14: 93245778-93245778 on Build GRCh38
rs118092506 Chr.14: 93246130-93246130 on Build GRCh38
rs375634897 Chr.14: 93246038-93246038 on Build GRCh38
rs773784616 Chr.14: 93245753-93245753 on Build GRCh38
rs534774411 Chr.14: 93245801-93245801 on Build GRCh38
rs778587077 Chr.14: 93245816-93245816 on Build GRCh38
rs760922085 Chr.14: 93246141-93246141 on Build GRCh38
rs748317296 Chr.14: 93245964-93245964 on Build GRCh38
rs746888000 Chr.14: 93245867-93245868 on Build GRCh38
rs757695688 Chr.14: 93245934-93245934 on Build GRCh38
rs150117172 Chr.14: 93246009-93246009 on Build GRCh38
rs775310218 Chr.14: 93245974-93245974 on Build GRCh38
rs12890823 Chr.14: 93245838-93245838 on Build GRCh38
rs756424655 Chr.14: 93245840-93245842 on Build GRCh38
rs774086947 Chr.14: 93246046-93246046 on Build GRCh38
rs754955079 Chr.14: 93245869-93245870 on Build GRCh38
rs760091724 Chr.14: 93246131-93246132 on Build GRCh38
rs145489029 Chr.14: 93246019-93246019 on Build GRCh38
rs774622619 Chr.14: 93245888-93245888 on Build GRCh38
rs763799229 Chr.14: 93245987-93245987 on Build GRCh38
rs780168711 Chr.14: 93245857-93245858 on Build GRCh38
rs781203655 Chr.14: 93246099-93246100 on Build GRCh38
rs767519544 Chr.14: 93245785-93245785 on Build GRCh38
rs746155999 Chr.14: 93245937-93245937 on Build GRCh38
rs758594827 Chr.14: 93246101-93246101 on Build GRCh38
rs747867992 Chr.14: 93246103-93246114 on Build GRCh38
rs778188665 Chr.14: 93245843-93245843 on Build GRCh38
rs201407664 Chr.14: 93245968-93245968 on Build GRCh38
rs767033525 Chr.14: 93246060-93246060 on Build GRCh38
rs771690645 Chr.14: 93246123-93246123 on Build GRCh38
rs553522189 Chr.14: 93245790-93245790 on Build GRCh38
rs769701963 Chr.14: 93246107-93246107 on Build GRCh38
rs558241574 Chr.14: 93245936-93245936 on Build GRCh38
rs564361018 Chr.14: 93245916-93245916 on Build GRCh38
rs761182084 Chr.14: 93246058-93246058 on Build GRCh38
rs753561085 Chr.14: 93245906-93245906 on Build GRCh38
rs148508605 Chr.14: 93245898-93245898 on Build GRCh38
rs878989992 Chr.14: 93246128-93246128 on Build GRCh38
rs757937511 Chr.14: 93245827-93245827 on Build GRCh38
rs759769618 Chr.14: 93246132-93246132 on Build GRCh38
rs770087269 Chr.14: 93245879-93245879 on Build GRCh38
rs774065129 Chr.14: 93246118-93246118 on Build GRCh38
rs559843145 Chr.14: 93245965-93245965 on Build GRCh38
rs144235721 Chr.14: 93245931-93245931 on Build GRCh38
rs780536598 Chr.14: 93246012-93246012 on Build GRCh38
rs550998075 Chr.14: 93246074-93246074 on Build GRCh38
rs777912493 Chr.14: 93246147-93246147 on Build GRCh38
rs755398350 Chr.14: 93246090-93246090 on Build GRCh38
rs768521978 Chr.14: 93246117-93246117 on Build GRCh38
rs762754430 Chr.14: 93246037-93246037 on Build GRCh38
rs749943034 Chr.14: 93246063-93246063 on Build GRCh38
rs946709827 Chr.14: 93245744-93245744 on Build GRCh38
rs759387545 Chr.14: 93245907-93245907 on Build GRCh38
rs751576449 Chr.14: 93246149-93246149 on Build GRCh38
rs189215614 Chr.14: 93245913-93245913 on Build GRCh38
rs753732824 Chr.14: 93245795-93245795 on Build GRCh38
rs778961670 Chr.14: 93246094-93246094 on Build GRCh38
rs575536203 Chr.14: 93245885-93245885 on Build GRCh38
rs573349419 Chr.14: 93245860-93245860 on Build GRCh38
rs61991737 Chr.14: 93245944-93245944 on Build GRCh38
rs368369249 Chr.14: 93245928-93245928 on Build GRCh38
rs760459841 Chr.14: 93245791-93245791 on Build GRCh38
rs770363546 Chr.14: 93246126-93246130 on Build GRCh38
rs748862996 Chr.14: 93246124-93246125 on Build GRCh38
rs754257757 Chr.14: 93246084-93246084 on Build GRCh38
rs756685745 Chr.14: 93246066-93246066 on Build GRCh38
rs946708889 Chr.14: 93245986-93245986 on Build GRCh38
rs1053070194 Chr.14: 93245754-93245754 on Build GRCh38
rs765611364 Chr.14: 93246138-93246138 on Build GRCh38
rs776633399 Chr.14: 93245967-93245967 on Build GRCh38
rs777239659 Chr.14: 93245920-93245920 on Build GRCh38
rs962707312 Chr.14: 93245925-93245925 on Build GRCh38
rs757232177 Chr.14: 93245854-93245855 on Build GRCh38
rs768880925 Chr.14: 93245883-93245883 on Build GRCh38
rs758639318 Chr.14: 93246144-93246144 on Build GRCh38
rs200009046 Chr.14: 93245954-93245954 on Build GRCh38
rs143877217 Chr.14: 93245971-93245971 on Build GRCh38
rs766073836 Chr.14: 93245794-93245794 on Build GRCh38
rs770316248 Chr.14: 93245938-93245938 on Build GRCh38
rs140532239 Chr.14: 93246008-93246008 on Build GRCh38
rs146853662 Chr.14: 93245991-93245991 on Build GRCh38
rs779975627 Chr.14: 93246110-93246110 on Build GRCh38
rs369767057 Chr.14: 93246105-93246105 on Build GRCh38
rs746305889 Chr.14: 93245877-93245877 on Build GRCh38
rs752155280 Chr.14: 93245825-93245825 on Build GRCh38
rs761498218 Chr.14: 93246120-93246120 on Build GRCh38
rs199912370 Chr.14: 93246106-93246106 on Build GRCh38
rs764187778 Chr.14: 93245839-93245840 on Build GRCh38
rs775258347 Chr.14: 93246036-93246036 on Build GRCh38
rs745781357 Chr.14: 93246022-93246022 on Build GRCh38
rs867790115 Chr.14: 93246136-93246136 on Build GRCh38

View Full Product Details