Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 93918707-93919217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM181A
Entrez Gene ID
90050
Gene Name
family with sequence similarity 181 member A
Gene Aliases
C14orf152
UniGene
Hs.525550
Gene Chromosome Location
Chr.14: 93918894-93929608 on Build GRCh38
Transcripts
NM_001207071.1 NM_138344.4 NR_027004.1

Gene Symbol
FAM181A-AS1
Entrez Gene ID
283592
Gene Name
FAM181A antisense RNA 1
Gene Aliases
C14orf86
UniGene
Hs.585648
Gene Chromosome Location
Chr.14: 93904730-93926372 on Build GRCh38
Transcripts
NM_001207071.1 NM_138344.4 NR_027004.1

SNP Detail

SNP ID Chromosome Location
rs891574266 Chr.14: 93919042-93919042 on Build GRCh38
rs912682115 Chr.14: 93918941-93918941 on Build GRCh38
rs117720558 Chr.14: 93919036-93919036 on Build GRCh38
rs1035402537 Chr.14: 93918869-93918869 on Build GRCh38
rs537781447 Chr.14: 93918890-93918890 on Build GRCh38
rs940107031 Chr.14: 93918983-93918983 on Build GRCh38
rs924338293 Chr.14: 93919027-93919027 on Build GRCh38
rs181549272 Chr.14: 93919064-93919064 on Build GRCh38
rs976645591 Chr.14: 93918795-93918795 on Build GRCh38
rs186428515 Chr.14: 93919172-93919172 on Build GRCh38
rs904332138 Chr.14: 93919059-93919059 on Build GRCh38
rs923523530 Chr.14: 93918984-93918984 on Build GRCh38
rs770711587 Chr.14: 93918883-93918883 on Build GRCh38
rs965325259 Chr.14: 93918971-93918971 on Build GRCh38
rs954007455 Chr.14: 93918929-93918929 on Build GRCh38
rs952151101 Chr.14: 93918925-93918925 on Build GRCh38
rs774527029 Chr.14: 93919037-93919037 on Build GRCh38
rs887852192 Chr.14: 93919113-93919113 on Build GRCh38
rs372243852 Chr.14: 93919071-93919071 on Build GRCh38
rs1026360923 Chr.14: 93918760-93918760 on Build GRCh38
rs931462684 Chr.14: 93919183-93919183 on Build GRCh38
rs1010110719 Chr.14: 93918888-93918888 on Build GRCh38
rs1026329426 Chr.14: 93918923-93918923 on Build GRCh38
rs905501811 Chr.14: 93918857-93918857 on Build GRCh38
rs972816411 Chr.14: 93919167-93919167 on Build GRCh38
rs746546808 Chr.14: 93918882-93918882 on Build GRCh38
rs1044485492 Chr.14: 93918832-93918832 on Build GRCh38
rs1054150219 Chr.14: 93918891-93918891 on Build GRCh38
rs945527984 Chr.14: 93918878-93918878 on Build GRCh38
rs996499272 Chr.14: 93919060-93919060 on Build GRCh38
rs745650679 Chr.14: 93918930-93918930 on Build GRCh38
rs570647741 Chr.14: 93918881-93918881 on Build GRCh38
rs978272497 Chr.14: 93919022-93919022 on Build GRCh38
rs1006342533 Chr.14: 93919115-93919115 on Build GRCh38
rs771884091 Chr.14: 93918766-93918766 on Build GRCh38
rs556445837 Chr.14: 93918903-93918903 on Build GRCh38
rs977340582 Chr.14: 93918877-93918877 on Build GRCh38
rs902946606 Chr.14: 93918884-93918884 on Build GRCh38
rs754430703 Chr.14: 93919068-93919068 on Build GRCh38
rs1051106236 Chr.14: 93918732-93918732 on Build GRCh38
rs1037630773 Chr.14: 93918994-93918994 on Build GRCh38
rs995757066 Chr.14: 93918823-93918823 on Build GRCh38
rs560183069 Chr.14: 93919182-93919182 on Build GRCh38
rs79737218 Chr.14: 93919100-93919100 on Build GRCh38
rs907142607 Chr.14: 93919078-93919078 on Build GRCh38
rs1012870093 Chr.14: 93918901-93918901 on Build GRCh38
rs45571237 Chr.14: 93919049-93919049 on Build GRCh38
rs777101316 Chr.14: 93918789-93918789 on Build GRCh38
rs1017635434 Chr.14: 93918962-93918962 on Build GRCh38
rs1045618349 Chr.14: 93919050-93919050 on Build GRCh38
rs920094906 Chr.14: 93919002-93919002 on Build GRCh38
rs116169098 Chr.14: 93919029-93919029 on Build GRCh38
rs993300192 Chr.14: 93918912-93918912 on Build GRCh38
rs188951658 Chr.14: 93918780-93918780 on Build GRCh38
rs984824416 Chr.14: 93918776-93918776 on Build GRCh38
rs966974134 Chr.14: 93919188-93919188 on Build GRCh38
rs907006684 Chr.14: 93918904-93918904 on Build GRCh38
rs894370163 Chr.14: 93918897-93918897 on Build GRCh38
rs142137032 Chr.14: 93918863-93918863 on Build GRCh38
rs921282320 Chr.14: 93918914-93918914 on Build GRCh38
rs151137248 Chr.14: 93919083-93919083 on Build GRCh38
rs12894357 Chr.14: 93919080-93919080 on Build GRCh38
rs986854788 Chr.14: 93918937-93918937 on Build GRCh38
rs139058031 Chr.14: 93919153-93919153 on Build GRCh38
rs115104454 Chr.14: 93919189-93919189 on Build GRCh38
rs964705537 Chr.14: 93919160-93919160 on Build GRCh38
rs1037154246 Chr.14: 93918788-93918788 on Build GRCh38
rs574606917 Chr.14: 93919021-93919021 on Build GRCh38
rs976319163 Chr.14: 93918975-93918975 on Build GRCh38
rs17129352 Chr.14: 93919162-93919162 on Build GRCh38

View Full Product Details