Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 93924707-93925246 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM181A
Entrez Gene ID
90050
Gene Name
family with sequence similarity 181 member A
Gene Aliases
C14orf152
UniGene
Hs.525550
Gene Chromosome Location
Chr.14: 93918894-93929608 on Build GRCh38
Transcripts
NM_001207071.1 NM_138344.4 NR_027004.1

Gene Symbol
FAM181A-AS1
Entrez Gene ID
283592
Gene Name
FAM181A antisense RNA 1
Gene Aliases
C14orf86
UniGene
Hs.585648
Gene Chromosome Location
Chr.14: 93904730-93926372 on Build GRCh38
Transcripts
NM_001207071.1 NM_138344.4 NR_027004.1

SNP Detail

SNP ID Chromosome Location
rs1024336440 Chr.14: 93924758-93924758 on Build GRCh38
rs1057360522 Chr.14: 93924896-93924896 on Build GRCh38
rs1017860042 Chr.14: 93925191-93925191 on Build GRCh38
rs73339799 Chr.14: 93925134-93925134 on Build GRCh38
rs779440600 Chr.14: 93925204-93925204 on Build GRCh38
rs535432095 Chr.14: 93925127-93925127 on Build GRCh38
rs952366807 Chr.14: 93924787-93924787 on Build GRCh38
rs938397879 Chr.14: 93925156-93925156 on Build GRCh38
rs533983925 Chr.14: 93925002-93925002 on Build GRCh38
rs141007020 Chr.14: 93925157-93925160 on Build GRCh38
rs1002751315 Chr.14: 93924892-93924892 on Build GRCh38
rs972954245 Chr.14: 93925095-93925095 on Build GRCh38
rs141654229 Chr.14: 93925183-93925183 on Build GRCh38
rs1000810584 Chr.14: 93925003-93925006 on Build GRCh38
rs755058711 Chr.14: 93925087-93925087 on Build GRCh38
rs955268492 Chr.14: 93925224-93925224 on Build GRCh38
rs1020930980 Chr.14: 93924958-93924958 on Build GRCh38
rs984965541 Chr.14: 93924856-93924856 on Build GRCh38
rs572169180 Chr.14: 93925140-93925140 on Build GRCh38
rs182577474 Chr.14: 93924819-93924819 on Build GRCh38
rs912851688 Chr.14: 93924753-93924753 on Build GRCh38
rs931066835 Chr.14: 93924825-93924825 on Build GRCh38
rs570769323 Chr.14: 93925067-93925067 on Build GRCh38
rs977465735 Chr.14: 93924972-93924972 on Build GRCh38
rs891601685 Chr.14: 93925179-93925179 on Build GRCh38
rs752192073 Chr.14: 93925149-93925149 on Build GRCh38
rs879531005 Chr.14: 93924925-93924925 on Build GRCh38
rs568094145 Chr.14: 93925118-93925118 on Build GRCh38
rs1010532356 Chr.14: 93924955-93924955 on Build GRCh38
rs922327860 Chr.14: 93924944-93924944 on Build GRCh38
rs112693564 Chr.14: 93925057-93925057 on Build GRCh38
rs919603300 Chr.14: 93925103-93925103 on Build GRCh38
rs1010168182 Chr.14: 93925181-93925181 on Build GRCh38
rs748563370 Chr.14: 93925219-93925219 on Build GRCh38
rs987357361 Chr.14: 93924749-93924749 on Build GRCh38
rs749371336 Chr.14: 93925047-93925047 on Build GRCh38
rs140263980 Chr.14: 93924766-93924766 on Build GRCh38
rs894348032 Chr.14: 93925084-93925084 on Build GRCh38
rs562336309 Chr.14: 93924843-93924843 on Build GRCh38
rs756348703 Chr.14: 93924786-93924786 on Build GRCh38
rs186159453 Chr.14: 93924906-93924906 on Build GRCh38
rs954267086 Chr.14: 93925064-93925064 on Build GRCh38
rs190153359 Chr.14: 93924729-93924729 on Build GRCh38
rs1054298509 Chr.14: 93925072-93925072 on Build GRCh38
rs748148539 Chr.14: 93925222-93925231 on Build GRCh38
rs191419877 Chr.14: 93925184-93925184 on Build GRCh38
rs967993321 Chr.14: 93924987-93924987 on Build GRCh38
rs529849420 Chr.14: 93924847-93924847 on Build GRCh38
rs576713689 Chr.14: 93925190-93925190 on Build GRCh38
rs149937597 Chr.14: 93924940-93924940 on Build GRCh38
rs933691777 Chr.14: 93924971-93924971 on Build GRCh38
rs910681938 Chr.14: 93924885-93924885 on Build GRCh38
rs971452620 Chr.14: 93924759-93924759 on Build GRCh38
rs780187863 Chr.14: 93925156-93925157 on Build GRCh38
rs924562406 Chr.14: 93924996-93924996 on Build GRCh38
rs537736717 Chr.14: 93925094-93925094 on Build GRCh38
rs552038418 Chr.14: 93925063-93925063 on Build GRCh38
rs543744088 Chr.14: 93925206-93925206 on Build GRCh38
rs888035955 Chr.14: 93925152-93925152 on Build GRCh38
rs562915160 Chr.14: 93925166-93925166 on Build GRCh38
rs556552170 Chr.14: 93925104-93925104 on Build GRCh38
rs376691319 Chr.14: 93924891-93924891 on Build GRCh38
rs370583616 Chr.14: 93925119-93925119 on Build GRCh38

View Full Product Details