Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 93933999-93934494 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB2
Entrez Gene ID
51676
Gene Name
ankyrin repeat and SOCS box containing 2
Gene Aliases
ASB-2
UniGene
Hs.510327
Gene Chromosome Location
Chr.14: 93934153-93976796 on Build GRCh38
Transcripts
NM_001202429.1 NM_016150.4 XM_005267758.3 XM_011536834.2 XM_011536835.2 XM_017021369.1

SNP Detail

SNP ID Chromosome Location
rs894999906 Chr.14: 93934233-93934233 on Build GRCh38
rs946425392 Chr.14: 93934189-93934189 on Build GRCh38
rs542180188 Chr.14: 93934181-93934181 on Build GRCh38
rs113402958 Chr.14: 93934048-93934048 on Build GRCh38
rs1002370222 Chr.14: 93934339-93934339 on Build GRCh38
rs899690119 Chr.14: 93934201-93934201 on Build GRCh38
rs548889336 Chr.14: 93934092-93934092 on Build GRCh38
rs969571576 Chr.14: 93934338-93934338 on Build GRCh38
rs751639585 Chr.14: 93934219-93934219 on Build GRCh38
rs753758396 Chr.14: 93934332-93934332 on Build GRCh38
rs755984274 Chr.14: 93934224-93934224 on Build GRCh38
rs1043592981 Chr.14: 93934200-93934200 on Build GRCh38
rs975814769 Chr.14: 93934095-93934095 on Build GRCh38
rs976848510 Chr.14: 93934443-93934443 on Build GRCh38
rs996738247 Chr.14: 93934030-93934030 on Build GRCh38
rs766316044 Chr.14: 93934331-93934331 on Build GRCh38
rs981015276 Chr.14: 93934101-93934101 on Build GRCh38
rs1029561861 Chr.14: 93934034-93934034 on Build GRCh38
rs8004028 Chr.14: 93934022-93934022 on Build GRCh38
rs985692552 Chr.14: 93934406-93934406 on Build GRCh38
rs939050897 Chr.14: 93934115-93934115 on Build GRCh38
rs952724660 Chr.14: 93934387-93934387 on Build GRCh38
rs528083197 Chr.14: 93934306-93934306 on Build GRCh38
rs936603671 Chr.14: 93934217-93934217 on Build GRCh38
rs76565163 Chr.14: 93934234-93934234 on Build GRCh38
rs765399533 Chr.14: 93934371-93934371 on Build GRCh38
rs945062836 Chr.14: 93934141-93934141 on Build GRCh38
rs80335391 Chr.14: 93934087-93934087 on Build GRCh38
rs554112222 Chr.14: 93934290-93934290 on Build GRCh38
rs778620666 Chr.14: 93934260-93934260 on Build GRCh38
rs776242935 Chr.14: 93934181-93934183 on Build GRCh38
rs368105566 Chr.14: 93934447-93934447 on Build GRCh38
rs558626819 Chr.14: 93934070-93934070 on Build GRCh38
rs541905372 Chr.14: 93934279-93934279 on Build GRCh38
rs985664535 Chr.14: 93934086-93934086 on Build GRCh38
rs1051485592 Chr.14: 93934220-93934220 on Build GRCh38
rs928236924 Chr.14: 93934103-93934103 on Build GRCh38
rs960949800 Chr.14: 93934451-93934451 on Build GRCh38
rs180871500 Chr.14: 93934313-93934313 on Build GRCh38
rs973111595 Chr.14: 93934054-93934054 on Build GRCh38
rs754770459 Chr.14: 93934244-93934244 on Build GRCh38
rs766322917 Chr.14: 93934231-93934231 on Build GRCh38
rs568690618 Chr.14: 93934215-93934215 on Build GRCh38
rs1018450828 Chr.14: 93934431-93934431 on Build GRCh38
rs1027678939 Chr.14: 93934336-93934336 on Build GRCh38
rs760160423 Chr.14: 93934280-93934280 on Build GRCh38
rs965524692 Chr.14: 93934432-93934432 on Build GRCh38
rs11625548 Chr.14: 93934085-93934085 on Build GRCh38
rs545522636 Chr.14: 93934248-93934248 on Build GRCh38
rs188300208 Chr.14: 93934093-93934093 on Build GRCh38
rs994254297 Chr.14: 93934303-93934303 on Build GRCh38
rs565020887 Chr.14: 93934379-93934379 on Build GRCh38
rs566775699 Chr.14: 93934044-93934044 on Build GRCh38
rs922978409 Chr.14: 93934108-93934108 on Build GRCh38
rs1042487823 Chr.14: 93934172-93934172 on Build GRCh38
rs1008891210 Chr.14: 93934049-93934049 on Build GRCh38

View Full Product Details