Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 93956919-93957457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB2
Entrez Gene ID
51676
Gene Name
ankyrin repeat and SOCS box containing 2
Gene Aliases
ASB-2
UniGene
Hs.510327
Gene Chromosome Location
Chr.14: 93934153-93976796 on Build GRCh38
Transcripts
NM_001202429.1 NM_016150.4 XM_005267758.3 XM_011536834.2 XM_017021369.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
959095 c.9C>A p.R3R Chr.14: 93956924-93956924 on Build GRCh38
4663652 c.8G>A p.R3H Chr.14: 93956925-93956925 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs3765494 Chr.14: 93956948-93956948 on Build GRCh38
rs749348691 Chr.14: 93956964-93956964 on Build GRCh38
rs572588380 Chr.14: 93957018-93957018 on Build GRCh38
rs898891715 Chr.14: 93957139-93957139 on Build GRCh38
rs199867477 Chr.14: 93956952-93956952 on Build GRCh38
rs976545618 Chr.14: 93957144-93957144 on Build GRCh38
rs756416347 Chr.14: 93956957-93956957 on Build GRCh38
rs574288545 Chr.14: 93957429-93957429 on Build GRCh38
rs939584848 Chr.14: 93957073-93957073 on Build GRCh38
rs935153596 Chr.14: 93957007-93957007 on Build GRCh38
rs915346064 Chr.14: 93957234-93957234 on Build GRCh38
rs999391904 Chr.14: 93957351-93957351 on Build GRCh38
rs371389301 Chr.14: 93957016-93957016 on Build GRCh38
rs535354318 Chr.14: 93957289-93957289 on Build GRCh38
rs923462000 Chr.14: 93957146-93957146 on Build GRCh38
rs954798110 Chr.14: 93957193-93957193 on Build GRCh38
rs537872513 Chr.14: 93957378-93957378 on Build GRCh38
rs909347596 Chr.14: 93957419-93957419 on Build GRCh38
rs574432696 Chr.14: 93957411-93957411 on Build GRCh38
rs368137954 Chr.14: 93957317-93957317 on Build GRCh38
rs1047331380 Chr.14: 93957279-93957279 on Build GRCh38
rs576672194 Chr.14: 93957053-93957053 on Build GRCh38
rs3835252 Chr.14: 93956981-93956982 on Build GRCh38
rs1017619043 Chr.14: 93957366-93957366 on Build GRCh38
rs554160033 Chr.14: 93957292-93957292 on Build GRCh38
rs3814823 Chr.14: 93957381-93957381 on Build GRCh38
rs981187741 Chr.14: 93957252-93957252 on Build GRCh38
rs779967090 Chr.14: 93956958-93956958 on Build GRCh38
rs888012505 Chr.14: 93957014-93957014 on Build GRCh38
rs987716055 Chr.14: 93957001-93957001 on Build GRCh38
rs543804213 Chr.14: 93957054-93957054 on Build GRCh38
rs552114914 Chr.14: 93957356-93957356 on Build GRCh38
rs3814824 Chr.14: 93957067-93957067 on Build GRCh38
rs551967973 Chr.14: 93956949-93956949 on Build GRCh38
rs777409065 Chr.14: 93957326-93957326 on Build GRCh38
rs923409442 Chr.14: 93956991-93956991 on Build GRCh38
rs559682923 Chr.14: 93957140-93957140 on Build GRCh38
rs568779744 Chr.14: 93957358-93957358 on Build GRCh38
rs764620347 Chr.14: 93956941-93956941 on Build GRCh38
rs957930791 Chr.14: 93957096-93957096 on Build GRCh38
rs568329449 Chr.14: 93957281-93957281 on Build GRCh38
rs928021364 Chr.14: 93957261-93957261 on Build GRCh38
rs989604543 Chr.14: 93957010-93957010 on Build GRCh38
rs574787594 Chr.14: 93957002-93957002 on Build GRCh38
rs141689443 Chr.14: 93957223-93957223 on Build GRCh38
rs572425647 Chr.14: 93957331-93957331 on Build GRCh38
rs570322001 Chr.14: 93957217-93957217 on Build GRCh38
rs902250744 Chr.14: 93957350-93957351 on Build GRCh38
rs929576382 Chr.14: 93956994-93956994 on Build GRCh38
rs79398428 Chr.14: 93957410-93957410 on Build GRCh38
rs377378577 Chr.14: 93956943-93956943 on Build GRCh38
rs146176608 Chr.14: 93957363-93957363 on Build GRCh38
rs138445914 Chr.14: 93957222-93957222 on Build GRCh38
rs548152148 Chr.14: 93957129-93957129 on Build GRCh38
rs371596976 Chr.14: 93957359-93957359 on Build GRCh38
rs546292922 Chr.14: 93957038-93957038 on Build GRCh38
rs373669765 Chr.14: 93957274-93957274 on Build GRCh38
rs551807005 Chr.14: 93957216-93957216 on Build GRCh38
rs192814902 Chr.14: 93957081-93957081 on Build GRCh38
rs969421248 Chr.14: 93957245-93957245 on Build GRCh38
rs376105153 Chr.14: 93956951-93956951 on Build GRCh38
rs537751076 Chr.14: 93956966-93956966 on Build GRCh38
rs527405661 Chr.14: 93957147-93957147 on Build GRCh38
rs964834110 Chr.14: 93957137-93957137 on Build GRCh38
rs1008850382 Chr.14: 93957211-93957211 on Build GRCh38
rs1028620056 Chr.14: 93957163-93957163 on Build GRCh38
rs756777623 Chr.14: 93957154-93957154 on Build GRCh38
rs774799897 Chr.14: 93957092-93957093 on Build GRCh38
rs1046203292 Chr.14: 93957342-93957342 on Build GRCh38
rs929502674 Chr.14: 93957161-93957161 on Build GRCh38
rs939343819 Chr.14: 93957278-93957278 on Build GRCh38
rs66925826 Chr.14: 93956981-93956981 on Build GRCh38
rs529681327 Chr.14: 93957104-93957104 on Build GRCh38
rs1005656016 Chr.14: 93957105-93957105 on Build GRCh38
rs150248187 Chr.14: 93957017-93957017 on Build GRCh38
rs564674889 Chr.14: 93957039-93957039 on Build GRCh38
rs1045062723 Chr.14: 93957047-93957047 on Build GRCh38
rs751980728 Chr.14: 93956988-93956988 on Build GRCh38
rs990205276 Chr.14: 93957103-93957103 on Build GRCh38
rs866413902 Chr.14: 93957336-93957336 on Build GRCh38
rs995818866 Chr.14: 93957151-93957151 on Build GRCh38
rs1013717271 Chr.14: 93957022-93957022 on Build GRCh38
rs777796672 Chr.14: 93956969-93956969 on Build GRCh38
rs535591863 Chr.14: 93957134-93957134 on Build GRCh38
rs1040552357 Chr.14: 93956990-93956990 on Build GRCh38
rs139130377 Chr.14: 93957402-93957402 on Build GRCh38
rs758447951 Chr.14: 93956974-93956977 on Build GRCh38

View Full Product Details